Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73681622-73682166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC30
Entrez Gene ID
84277
Gene Name
DnaJ heat shock protein family (Hsp40) member C30
Gene Aliases
WBSCR18
UniGene
Hs.647046
Gene Chromosome Location
Chr.7: 73680918-73683451 on Build GRCh38
Transcripts
NM_032317.2

SNP Detail

SNP ID Chromosome Location
rs550609527 Chr.7: 73682129-73682129 on Build GRCh38
rs937699841 Chr.7: 73681844-73681844 on Build GRCh38
rs965381862 Chr.7: 73681719-73681719 on Build GRCh38
rs975418487 Chr.7: 73681978-73681978 on Build GRCh38
rs186916259 Chr.7: 73681742-73681742 on Build GRCh38
rs575755110 Chr.7: 73681874-73681874 on Build GRCh38
rs3828974 Chr.7: 73681676-73681676 on Build GRCh38
rs535944485 Chr.7: 73681784-73681784 on Build GRCh38
rs782597836 Chr.7: 73681940-73681940 on Build GRCh38
rs921361216 Chr.7: 73681725-73681725 on Build GRCh38
rs1019894613 Chr.7: 73681946-73681946 on Build GRCh38
rs545224762 Chr.7: 73682091-73682091 on Build GRCh38
rs569143383 Chr.7: 73681724-73681724 on Build GRCh38
rs747044210 Chr.7: 73681751-73681751 on Build GRCh38
rs1046285917 Chr.7: 73681812-73681812 on Build GRCh38
rs539465403 Chr.7: 73682063-73682063 on Build GRCh38
rs564734050 Chr.7: 73681987-73681987 on Build GRCh38
rs939871416 Chr.7: 73682035-73682035 on Build GRCh38
rs1054681121 Chr.7: 73681863-73681863 on Build GRCh38
rs1028733122 Chr.7: 73681984-73681984 on Build GRCh38
rs573591952 Chr.7: 73681989-73681989 on Build GRCh38
rs529019464 Chr.7: 73682086-73682086 on Build GRCh38
rs965580145 Chr.7: 73681954-73681954 on Build GRCh38
rs906431839 Chr.7: 73681831-73681831 on Build GRCh38
rs192291794 Chr.7: 73682062-73682062 on Build GRCh38
rs1054834965 Chr.7: 73682075-73682075 on Build GRCh38
rs782362722 Chr.7: 73681887-73681887 on Build GRCh38
rs984594774 Chr.7: 73682003-73682003 on Build GRCh38
rs368070429 Chr.7: 73682140-73682140 on Build GRCh38
rs557315406 Chr.7: 73681869-73681869 on Build GRCh38
rs903336114 Chr.7: 73681909-73681909 on Build GRCh38
rs908400453 Chr.7: 73682008-73682008 on Build GRCh38
rs782189251 Chr.7: 73681785-73681785 on Build GRCh38
rs908368912 Chr.7: 73681769-73681769 on Build GRCh38
rs946513341 Chr.7: 73682111-73682111 on Build GRCh38
rs1011066179 Chr.7: 73681893-73681893 on Build GRCh38
rs536224511 Chr.7: 73682030-73682030 on Build GRCh38
rs115086105 Chr.7: 73682051-73682051 on Build GRCh38
rs573560468 Chr.7: 73682037-73682037 on Build GRCh38
rs1021816228 Chr.7: 73681895-73681895 on Build GRCh38
rs3763431 Chr.7: 73681991-73681991 on Build GRCh38
rs139533931 Chr.7: 73681782-73681782 on Build GRCh38

View Full Product Details