Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100573737-100574270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRCH4
Entrez Gene ID
4034
Gene Name
leucine rich repeats and calponin homology domain containing 4
Gene Aliases
LRN, LRRN1, LRRN4, PP14183
UniGene
Hs.125742
Gene Chromosome Location
Chr.7: 100574011-100586188 on Build GRCh38
Transcripts
NM_001289934.1 NM_002319.4 XM_005250096.3 XM_006715822.3 NM_001289933.1

Gene Symbol
SAP25
Entrez Gene ID
100316904
Gene Name
Sin3A associated protein 25
Gene Aliases
-
UniGene
Hs.633863
Gene Chromosome Location
Chr.7: 100572225-100573864 on Build GRCh38
Transcripts
NM_001289934.1 NM_002319.4 XM_005250096.3 XM_006715822.3 NM_001289933.1

Gene Symbol
ZASP
Entrez Gene ID
101927655
Gene Name
ZO-2 associated speckle protein
Gene Aliases
-
UniGene
Hs.125742
Gene Chromosome Location
Chr.7: 100574035-100574733 on Build GRCh38
Transcripts
NM_001289934.1 NM_002319.4 XM_005250096.3 XM_006715822.3 NM_001289933.1

SNP Detail

SNP ID Chromosome Location
rs1053886326 Chr.7: 100573793-100573793 on Build GRCh38
rs887380954 Chr.7: 100574016-100574016 on Build GRCh38
rs764374013 Chr.7: 100574209-100574209 on Build GRCh38
rs562168405 Chr.7: 100573887-100573887 on Build GRCh38
rs939461174 Chr.7: 100574227-100574227 on Build GRCh38
rs911591895 Chr.7: 100574147-100574147 on Build GRCh38
rs566462396 Chr.7: 100574005-100574006 on Build GRCh38
rs957189381 Chr.7: 100574244-100574244 on Build GRCh38
rs920845222 Chr.7: 100574178-100574178 on Build GRCh38
rs887730270 Chr.7: 100574065-100574073 on Build GRCh38
rs1003755634 Chr.7: 100573842-100573842 on Build GRCh38
rs922436488 Chr.7: 100573911-100573911 on Build GRCh38
rs930875667 Chr.7: 100574179-100574179 on Build GRCh38
rs538341896 Chr.7: 100574129-100574129 on Build GRCh38
rs902514505 Chr.7: 100573788-100573788 on Build GRCh38
rs899518101 Chr.7: 100574192-100574192 on Build GRCh38
rs1047719203 Chr.7: 100574143-100574143 on Build GRCh38
rs928081279 Chr.7: 100574125-100574125 on Build GRCh38
rs1026295396 Chr.7: 100573852-100573852 on Build GRCh38
rs939420329 Chr.7: 100574130-100574130 on Build GRCh38
rs564123554 Chr.7: 100574139-100574139 on Build GRCh38
rs964175384 Chr.7: 100573898-100573898 on Build GRCh38
rs761611927 Chr.7: 100574238-100574238 on Build GRCh38
rs201992851 Chr.7: 100574048-100574048 on Build GRCh38
rs1029394482 Chr.7: 100574059-100574059 on Build GRCh38
rs974380433 Chr.7: 100574167-100574167 on Build GRCh38
rs539063846 Chr.7: 100574249-100574249 on Build GRCh38
rs924492290 Chr.7: 100573949-100573949 on Build GRCh38
rs539244317 Chr.7: 100573969-100573970 on Build GRCh38
rs1045090053 Chr.7: 100573974-100573974 on Build GRCh38
rs3173634 Chr.7: 100574174-100574174 on Build GRCh38
rs887276724 Chr.7: 100574145-100574145 on Build GRCh38
rs1017044552 Chr.7: 100574034-100574034 on Build GRCh38
rs1016583052 Chr.7: 100573894-100573894 on Build GRCh38
rs975151446 Chr.7: 100573899-100573899 on Build GRCh38
rs1029317522 Chr.7: 100574210-100574210 on Build GRCh38
rs895131773 Chr.7: 100574046-100574046 on Build GRCh38
rs908032362 Chr.7: 100574196-100574196 on Build GRCh38
rs112460186 Chr.7: 100574035-100574035 on Build GRCh38
rs578058484 Chr.7: 100574245-100574245 on Build GRCh38
rs1003640889 Chr.7: 100573998-100573999 on Build GRCh38
rs1045141822 Chr.7: 100574111-100574111 on Build GRCh38
rs1012265784 Chr.7: 100573819-100573819 on Build GRCh38
rs772723592 Chr.7: 100574118-100574118 on Build GRCh38
rs1038577576 Chr.7: 100574184-100574184 on Build GRCh38
rs933928954 Chr.7: 100573781-100573781 on Build GRCh38
rs534127904 Chr.7: 100574047-100574047 on Build GRCh38
rs924448630 Chr.7: 100574066-100574066 on Build GRCh38
rs754201608 Chr.7: 100574213-100574213 on Build GRCh38
rs893910694 Chr.7: 100573813-100573813 on Build GRCh38
rs935918683 Chr.7: 100573950-100573950 on Build GRCh38
rs1048394954 Chr.7: 100574195-100574195 on Build GRCh38
rs148046169 Chr.7: 100573821-100573821 on Build GRCh38
rs1005799532 Chr.7: 100574019-100574019 on Build GRCh38
rs1000446188 Chr.7: 100574090-100574090 on Build GRCh38
rs899067785 Chr.7: 100573969-100573969 on Build GRCh38
rs1054219574 Chr.7: 100573951-100573951 on Build GRCh38
rs77221373 Chr.7: 100574110-100574110 on Build GRCh38
rs995131834 Chr.7: 100573970-100573970 on Build GRCh38
rs1041622396 Chr.7: 100573784-100573785 on Build GRCh38
rs551645318 Chr.7: 100573948-100573948 on Build GRCh38
rs533414365 Chr.7: 100573917-100573917 on Build GRCh38
rs867357135 Chr.7: 100574065-100574065 on Build GRCh38
rs906731428 Chr.7: 100573990-100573990 on Build GRCh38
rs998611400 Chr.7: 100574212-100574212 on Build GRCh38
rs968626099 Chr.7: 100574084-100574084 on Build GRCh38
rs987203210 Chr.7: 100574138-100574138 on Build GRCh38
rs370122417 Chr.7: 100573963-100573964 on Build GRCh38
rs1031954208 Chr.7: 100574215-100574215 on Build GRCh38
rs915232723 Chr.7: 100574104-100574104 on Build GRCh38
rs996486244 Chr.7: 100574194-100574194 on Build GRCh38
rs73711136 Chr.7: 100574165-100574165 on Build GRCh38
rs922426550 Chr.7: 100573775-100573775 on Build GRCh38
rs867932608 Chr.7: 100573973-100573973 on Build GRCh38
rs951093172 Chr.7: 100573874-100573874 on Build GRCh38

View Full Product Details