Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 116953261-116953759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST7
Entrez Gene ID
7982
Gene Name
suppression of tumorigenicity 7
Gene Aliases
ETS7q, FAM4A, FAM4A1, HELG, RAY1, SEN4, TSG7
UniGene
Hs.368131
Gene Chromosome Location
Chr.7: 116953327-117230023 on Build GRCh38
Transcripts
NM_018412.3 NM_021908.2 NR_002330.1

Gene Symbol
ST7-AS1
Entrez Gene ID
93653
Gene Name
ST7 antisense RNA 1
Gene Aliases
ST7AS1, ST7OT1
UniGene
Hs.597516
Gene Chromosome Location
Chr.7: 116952446-116954334 on Build GRCh38
Transcripts
NM_018412.3 NM_021908.2 NR_002330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1488151 c.36C>T p.L12L Chr.7: 116953576-116953576 on Build GRCh38
1316564 c.104T>C p.V35A Chr.7: 116953644-116953644 on Build GRCh38
4482382 c.25C>T p.L9L Chr.7: 116953565-116953565 on Build GRCh38
5762545 c.151+6G>A p.? Chr.7: 116953697-116953697 on Build GRCh38
1227690 c.129A>C p.K43N Chr.7: 116953669-116953669 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572638275 Chr.7: 116953361-116953361 on Build GRCh38
rs901545537 Chr.7: 116953532-116953532 on Build GRCh38
rs1056592348 Chr.7: 116953447-116953448 on Build GRCh38
rs754753980 Chr.7: 116953597-116953597 on Build GRCh38
rs750009066 Chr.7: 116953580-116953580 on Build GRCh38
rs939369155 Chr.7: 116953739-116953739 on Build GRCh38
rs763423090 Chr.7: 116953568-116953568 on Build GRCh38
rs748711364 Chr.7: 116953733-116953733 on Build GRCh38
rs765077451 Chr.7: 116953684-116953684 on Build GRCh38
rs200586105 Chr.7: 116953739-116953739 on Build GRCh38
rs947922096 Chr.7: 116953473-116953473 on Build GRCh38
rs186337335 Chr.7: 116953375-116953375 on Build GRCh38
rs564015259 Chr.7: 116953474-116953474 on Build GRCh38
rs370235468 Chr.7: 116953574-116953574 on Build GRCh38
rs778870159 Chr.7: 116953508-116953508 on Build GRCh38
rs753464201 Chr.7: 116953590-116953590 on Build GRCh38
rs764988892 Chr.7: 116953599-116953599 on Build GRCh38
rs144972398 Chr.7: 116953557-116953557 on Build GRCh38
rs952110890 Chr.7: 116953296-116953296 on Build GRCh38
rs775781428 Chr.7: 116953666-116953666 on Build GRCh38
rs1014524936 Chr.7: 116953490-116953490 on Build GRCh38
rs968310354 Chr.7: 116953681-116953681 on Build GRCh38
rs769078832 Chr.7: 116953471-116953471 on Build GRCh38
rs946703262 Chr.7: 116953433-116953433 on Build GRCh38
rs922760299 Chr.7: 116953373-116953373 on Build GRCh38
rs535830871 Chr.7: 116953723-116953723 on Build GRCh38
rs770174891 Chr.7: 116953658-116953658 on Build GRCh38
rs760270648 Chr.7: 116953581-116953581 on Build GRCh38
rs752647523 Chr.7: 116953606-116953606 on Build GRCh38
rs770533098 Chr.7: 116953735-116953735 on Build GRCh38
rs559871671 Chr.7: 116953451-116953451 on Build GRCh38
rs777819624 Chr.7: 116953628-116953628 on Build GRCh38
rs866422448 Chr.7: 116953324-116953324 on Build GRCh38
rs924001346 Chr.7: 116953439-116953439 on Build GRCh38
rs747229687 Chr.7: 116953520-116953520 on Build GRCh38
rs549430851 Chr.7: 116953488-116953488 on Build GRCh38
rs564113475 Chr.7: 116953719-116953719 on Build GRCh38
rs978102335 Chr.7: 116953435-116953435 on Build GRCh38
rs530902566 Chr.7: 116953487-116953487 on Build GRCh38
rs776853279 Chr.7: 116953521-116953521 on Build GRCh38
rs774949259 Chr.7: 116953732-116953732 on Build GRCh38
rs752493455 Chr.7: 116953694-116953694 on Build GRCh38
rs571378922 Chr.7: 116953677-116953677 on Build GRCh38
rs753625139 Chr.7: 116953495-116953495 on Build GRCh38
rs759881657 Chr.7: 116953531-116953531 on Build GRCh38
rs533367944 Chr.7: 116953453-116953453 on Build GRCh38
rs538497080 Chr.7: 116953682-116953682 on Build GRCh38
rs769171042 Chr.7: 116953730-116953730 on Build GRCh38
rs915479803 Chr.7: 116953383-116953383 on Build GRCh38
rs965786026 Chr.7: 116953321-116953321 on Build GRCh38
rs535089804 Chr.7: 116953519-116953519 on Build GRCh38
rs746104078 Chr.7: 116953653-116953653 on Build GRCh38
rs567678176 Chr.7: 116953509-116953509 on Build GRCh38
rs757281468 Chr.7: 116953706-116953706 on Build GRCh38
rs997213503 Chr.7: 116953539-116953539 on Build GRCh38
rs756345421 Chr.7: 116953718-116953718 on Build GRCh38
rs139362546 Chr.7: 116953639-116953639 on Build GRCh38
rs774998721 Chr.7: 116953673-116953673 on Build GRCh38
rs939400201 Chr.7: 116953441-116953441 on Build GRCh38
rs866589596 Chr.7: 116953485-116953485 on Build GRCh38
rs754732919 Chr.7: 116953496-116953496 on Build GRCh38
rs766059066 Chr.7: 116953586-116953586 on Build GRCh38
rs770090404 Chr.7: 116953551-116953551 on Build GRCh38
rs781183397 Chr.7: 116953709-116953709 on Build GRCh38
rs758205368 Chr.7: 116953698-116953698 on Build GRCh38
rs181383891 Chr.7: 116953305-116953305 on Build GRCh38
rs769224047 Chr.7: 116953671-116953671 on Build GRCh38
rs557219322 Chr.7: 116953702-116953702 on Build GRCh38
rs552038902 Chr.7: 116953454-116953454 on Build GRCh38
rs764027499 Chr.7: 116953522-116953523 on Build GRCh38
rs747033623 Chr.7: 116953631-116953631 on Build GRCh38
rs779978105 Chr.7: 116953493-116953493 on Build GRCh38
rs376240267 Chr.7: 116953670-116953670 on Build GRCh38
rs746076144 Chr.7: 116953711-116953711 on Build GRCh38
rs369531523 Chr.7: 116953705-116953705 on Build GRCh38
rs147927478 Chr.7: 116953616-116953616 on Build GRCh38

View Full Product Details