Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 155303484-155303977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INSIG1
Entrez Gene ID
3638
Gene Name
insulin induced gene 1
Gene Aliases
CL-6, CL6
UniGene
Hs.520819
Gene Chromosome Location
Chr.7: 155297776-155310235 on Build GRCh38
Transcripts
NM_005542.4 NM_198336.2 NM_198337.2 XM_005249542.1 XM_005249543.1 XM_017012177.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6224478 c.865A>C p.S289R Chr.7: 155303885-155303885 on Build GRCh38
7235925 c.875+10G>T p.? Chr.7: 155303905-155303905 on Build GRCh38
5701648 c.875_875+1GG>TT p.? Chr.7: 155303895-155303896 on Build GRCh38
4838698 c.816C>T p.F272F Chr.7: 155303836-155303836 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs9692436 Chr.7: 155303576-155303576 on Build GRCh38
rs143701174 Chr.7: 155303660-155303660 on Build GRCh38
rs9768687 Chr.7: 155303726-155303726 on Build GRCh38
rs923714218 Chr.7: 155303519-155303519 on Build GRCh38
rs994877844 Chr.7: 155303616-155303616 on Build GRCh38
rs899384491 Chr.7: 155303615-155303615 on Build GRCh38
rs542433506 Chr.7: 155303894-155303894 on Build GRCh38
rs201635926 Chr.7: 155303896-155303896 on Build GRCh38
rs955808269 Chr.7: 155303561-155303561 on Build GRCh38
rs370013374 Chr.7: 155303794-155303794 on Build GRCh38
rs986515148 Chr.7: 155303728-155303728 on Build GRCh38
rs148946308 Chr.7: 155303575-155303575 on Build GRCh38
rs904405014 Chr.7: 155303673-155303673 on Build GRCh38
rs758407859 Chr.7: 155303782-155303782 on Build GRCh38
rs781269534 Chr.7: 155303941-155303941 on Build GRCh38
rs746866191 Chr.7: 155303931-155303932 on Build GRCh38
rs764266272 Chr.7: 155303559-155303559 on Build GRCh38
rs1044338528 Chr.7: 155303668-155303668 on Build GRCh38
rs754911057 Chr.7: 155303915-155303915 on Build GRCh38
rs750947072 Chr.7: 155303956-155303956 on Build GRCh38
rs991029253 Chr.7: 155303658-155303658 on Build GRCh38
rs187142732 Chr.7: 155303936-155303936 on Build GRCh38
rs778796614 Chr.7: 155303920-155303920 on Build GRCh38
rs373521398 Chr.7: 155303810-155303810 on Build GRCh38
rs777538409 Chr.7: 155303927-155303928 on Build GRCh38
rs576787621 Chr.7: 155303718-155303718 on Build GRCh38
rs181063437 Chr.7: 155303656-155303656 on Build GRCh38
rs895205055 Chr.7: 155303954-155303954 on Build GRCh38
rs765920985 Chr.7: 155303886-155303886 on Build GRCh38
rs779533807 Chr.7: 155303780-155303780 on Build GRCh38
rs774540117 Chr.7: 155303787-155303787 on Build GRCh38
rs1017861242 Chr.7: 155303730-155303730 on Build GRCh38
rs558544668 Chr.7: 155303680-155303680 on Build GRCh38
rs536438378 Chr.7: 155303634-155303634 on Build GRCh38
rs532863999 Chr.7: 155303564-155303564 on Build GRCh38
rs895681081 Chr.7: 155303661-155303661 on Build GRCh38
rs891450439 Chr.7: 155303714-155303714 on Build GRCh38
rs369053471 Chr.7: 155303866-155303866 on Build GRCh38
rs757728813 Chr.7: 155303617-155303617 on Build GRCh38
rs555080709 Chr.7: 155303651-155303651 on Build GRCh38
rs200710774 Chr.7: 155303845-155303845 on Build GRCh38
rs948530714 Chr.7: 155303664-155303664 on Build GRCh38
rs750005581 Chr.7: 155303847-155303847 on Build GRCh38
rs977764955 Chr.7: 155303700-155303700 on Build GRCh38
rs929294986 Chr.7: 155303857-155303857 on Build GRCh38
rs202092473 Chr.7: 155303717-155303718 on Build GRCh38
rs954879204 Chr.7: 155303726-155303729 on Build GRCh38
rs73178614 Chr.7: 155303542-155303542 on Build GRCh38
rs148094376 Chr.7: 155303776-155303776 on Build GRCh38
rs754364743 Chr.7: 155303586-155303586 on Build GRCh38
rs1023290917 Chr.7: 155303538-155303538 on Build GRCh38
rs923599412 Chr.7: 155303712-155303712 on Build GRCh38
rs60860323 Chr.7: 155303910-155303910 on Build GRCh38
rs758738320 Chr.7: 155303778-155303778 on Build GRCh38
rs377100490 Chr.7: 155303934-155303934 on Build GRCh38
rs576294359 Chr.7: 155303632-155303632 on Build GRCh38
rs764466138 Chr.7: 155303784-155303784 on Build GRCh38
rs200321841 Chr.7: 155303726-155303726 on Build GRCh38
rs1003622390 Chr.7: 155303646-155303646 on Build GRCh38
rs79984271 Chr.7: 155303742-155303742 on Build GRCh38
rs145773058 Chr.7: 155303541-155303541 on Build GRCh38
rs372824115 Chr.7: 155303869-155303869 on Build GRCh38
rs189166944 Chr.7: 155303538-155303538 on Build GRCh38

View Full Product Details