Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161160920-161161416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP21
Entrez Gene ID
27005
Gene Name
ubiquitin specific peptidase 21
Gene Aliases
USP16, USP23
UniGene
Hs.8015
Gene Chromosome Location
Chr.1: 161159458-161165726 on Build GRCh38
Transcripts
NM_001014443.2 NM_001319848.1 NM_012475.4 XM_011509414.1 XM_017001036.1 XM_017001037.1 XM_017001038.1 XM_017001039.1 XM_017001040.1 XM_017001041.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
249153 c.365T>C p.M122T Chr.1: 161161005-161161005 on Build GRCh38
7241853 c.433C>T p.R145W Chr.1: 161161073-161161073 on Build GRCh38
286385 c.535T>C p.S179P Chr.1: 161161175-161161175 on Build GRCh38
7241852 c.432C>T p.L144L Chr.1: 161161072-161161072 on Build GRCh38
1295354 c.366G>A p.M122I Chr.1: 161161006-161161006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140569144 Chr.1: 161161140-161161140 on Build GRCh38
rs755353846 Chr.1: 161161261-161161261 on Build GRCh38
rs760197993 Chr.1: 161161205-161161205 on Build GRCh38
rs367921038 Chr.1: 161161351-161161351 on Build GRCh38
rs763201934 Chr.1: 161160969-161160969 on Build GRCh38
rs529747030 Chr.1: 161161071-161161071 on Build GRCh38
rs777352567 Chr.1: 161161070-161161070 on Build GRCh38
rs554465394 Chr.1: 161161232-161161232 on Build GRCh38
rs759274012 Chr.1: 161161211-161161211 on Build GRCh38
rs377456378 Chr.1: 161161209-161161209 on Build GRCh38
rs748611022 Chr.1: 161161275-161161275 on Build GRCh38
rs755520314 Chr.1: 161161157-161161157 on Build GRCh38
rs897868457 Chr.1: 161161111-161161111 on Build GRCh38
rs753499138 Chr.1: 161161236-161161236 on Build GRCh38
rs950084465 Chr.1: 161161316-161161316 on Build GRCh38
rs752222593 Chr.1: 161161109-161161109 on Build GRCh38
rs763740106 Chr.1: 161161115-161161115 on Build GRCh38
rs149224884 Chr.1: 161161234-161161234 on Build GRCh38
rs372425350 Chr.1: 161160965-161160965 on Build GRCh38
rs758241325 Chr.1: 161160953-161160953 on Build GRCh38
rs905160197 Chr.1: 161161324-161161324 on Build GRCh38
rs375923326 Chr.1: 161161036-161161036 on Build GRCh38
rs1002071132 Chr.1: 161161325-161161325 on Build GRCh38
rs755738229 Chr.1: 161161063-161161063 on Build GRCh38
rs375476473 Chr.1: 161160967-161160967 on Build GRCh38
rs371692736 Chr.1: 161161073-161161073 on Build GRCh38
rs201884202 Chr.1: 161161260-161161260 on Build GRCh38
rs1044357580 Chr.1: 161161319-161161319 on Build GRCh38
rs751053740 Chr.1: 161160966-161160966 on Build GRCh38
rs149047051 Chr.1: 161161099-161161099 on Build GRCh38
rs1009741486 Chr.1: 161161345-161161345 on Build GRCh38
rs966602309 Chr.1: 161161168-161161168 on Build GRCh38
rs748273705 Chr.1: 161161174-161161174 on Build GRCh38
rs994821674 Chr.1: 161161112-161161112 on Build GRCh38
rs151044234 Chr.1: 161161074-161161074 on Build GRCh38
rs370393340 Chr.1: 161161342-161161342 on Build GRCh38
rs374162328 Chr.1: 161161285-161161285 on Build GRCh38
rs1008016886 Chr.1: 161161143-161161143 on Build GRCh38
rs369085261 Chr.1: 161161105-161161105 on Build GRCh38
rs769907409 Chr.1: 161160946-161160946 on Build GRCh38
rs760671742 Chr.1: 161161018-161161018 on Build GRCh38
rs367894011 Chr.1: 161160961-161160961 on Build GRCh38
rs762124989 Chr.1: 161161032-161161032 on Build GRCh38
rs754462761 Chr.1: 161161061-161161061 on Build GRCh38
rs775338931 Chr.1: 161161226-161161226 on Build GRCh38
rs746436438 Chr.1: 161161005-161161005 on Build GRCh38
rs200067760 Chr.1: 161161022-161161022 on Build GRCh38
rs191180673 Chr.1: 161160998-161160998 on Build GRCh38
rs762760965 Chr.1: 161161048-161161048 on Build GRCh38
rs751449721 Chr.1: 161161059-161161059 on Build GRCh38
rs373371747 Chr.1: 161161169-161161169 on Build GRCh38
rs747512665 Chr.1: 161161147-161161147 on Build GRCh38
rs930518506 Chr.1: 161161390-161161390 on Build GRCh38
rs772474679 Chr.1: 161161007-161161007 on Build GRCh38
rs917251451 Chr.1: 161161315-161161315 on Build GRCh38
rs767469808 Chr.1: 161161219-161161219 on Build GRCh38
rs750167338 Chr.1: 161161037-161161037 on Build GRCh38
rs752768895 Chr.1: 161161152-161161152 on Build GRCh38
rs766704023 Chr.1: 161161249-161161249 on Build GRCh38
rs375288621 Chr.1: 161161091-161161091 on Build GRCh38
rs868304743 Chr.1: 161161235-161161235 on Build GRCh38
rs773447985 Chr.1: 161161098-161161098 on Build GRCh38
rs369956021 Chr.1: 161161141-161161141 on Build GRCh38
rs143084354 Chr.1: 161161374-161161374 on Build GRCh38
rs1013474720 Chr.1: 161161049-161161049 on Build GRCh38
rs369057043 Chr.1: 161161057-161161057 on Build GRCh38
rs753471183 Chr.1: 161161116-161161116 on Build GRCh38
rs373262739 Chr.1: 161161113-161161113 on Build GRCh38
rs148485578 Chr.1: 161161062-161161062 on Build GRCh38
rs1019119107 Chr.1: 161161166-161161166 on Build GRCh38
rs762176011 Chr.1: 161161086-161161086 on Build GRCh38
rs376196870 Chr.1: 161161127-161161127 on Build GRCh38
rs566090442 Chr.1: 161161077-161161077 on Build GRCh38
rs777112723 Chr.1: 161161216-161161216 on Build GRCh38
rs766259751 Chr.1: 161161050-161161050 on Build GRCh38
rs750456510 Chr.1: 161160944-161160944 on Build GRCh38
rs751515613 Chr.1: 161161104-161161104 on Build GRCh38
rs1054037829 Chr.1: 161161331-161161331 on Build GRCh38
rs756904192 Chr.1: 161161238-161161238 on Build GRCh38
rs540693926 Chr.1: 161161262-161161262 on Build GRCh38
rs538169930 Chr.1: 161161201-161161201 on Build GRCh38
rs1036305623 Chr.1: 161161360-161161360 on Build GRCh38
rs779217194 Chr.1: 161160999-161160999 on Build GRCh38
rs980897029 Chr.1: 161161296-161161296 on Build GRCh38
rs554313844 Chr.1: 161161300-161161300 on Build GRCh38
rs776021910 Chr.1: 161161017-161161017 on Build GRCh38
rs749514583 Chr.1: 161161206-161161206 on Build GRCh38
rs759816778 Chr.1: 161161243-161161243 on Build GRCh38
rs372271090 Chr.1: 161160979-161160979 on Build GRCh38
rs768795185 Chr.1: 161161028-161161028 on Build GRCh38
rs143739061 Chr.1: 161161078-161161078 on Build GRCh38
rs766142083 Chr.1: 161161100-161161100 on Build GRCh38
rs3813616 Chr.1: 161161029-161161029 on Build GRCh38

View Full Product Details