Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 74362804-74363302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDAP1
Entrez Gene ID
54332
Gene Name
ganglioside induced differentiation associated protein 1
Gene Aliases
CMT4, CMT4A, CMTRIA
UniGene
Hs.168950
Gene Chromosome Location
Chr.8: 74349251-74488867 on Build GRCh38
Transcripts
NM_001040875.2 NM_018972.2 NR_046346.1 XM_011517551.2 XM_011517552.2 XM_017013585.1 XM_017013586.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6393864 c.593A>C p.D198A Chr.8: 74362952-74362952 on Build GRCh38
751813 c.625C>G p.L209V Chr.8: 74362984-74362984 on Build GRCh38
751812 c.683A>G p.E228G Chr.8: 74363042-74363042 on Build GRCh38
1193667 c.677G>A p.R226K Chr.8: 74363036-74363036 on Build GRCh38
3835014 c.591T>G p.L197L Chr.8: 74362950-74362950 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs4463412 Chr.8: 74363077-74363077 on Build GRCh38
rs769449440 Chr.8: 74363027-74363027 on Build GRCh38
rs759443843 Chr.8: 74363044-74363044 on Build GRCh38
rs200390101 Chr.8: 74362988-74362988 on Build GRCh38
rs181157785 Chr.8: 74363052-74363052 on Build GRCh38
rs866655204 Chr.8: 74362909-74362909 on Build GRCh38
rs776580087 Chr.8: 74363040-74363041 on Build GRCh38
rs549461011 Chr.8: 74363256-74363256 on Build GRCh38
rs565901784 Chr.8: 74362953-74362953 on Build GRCh38
rs751633302 Chr.8: 74363051-74363052 on Build GRCh38
rs762473236 Chr.8: 74363040-74363040 on Build GRCh38
rs546886921 Chr.8: 74363106-74363106 on Build GRCh38
rs371325136 Chr.8: 74362963-74362963 on Build GRCh38
rs121908114 Chr.8: 74363051-74363051 on Build GRCh38
rs776973989 Chr.8: 74362999-74362999 on Build GRCh38
rs559771647 Chr.8: 74363133-74363133 on Build GRCh38
rs377712744 Chr.8: 74362958-74362958 on Build GRCh38
rs758365842 Chr.8: 74362968-74362968 on Build GRCh38
rs772784810 Chr.8: 74363028-74363028 on Build GRCh38
rs16938895 Chr.8: 74363128-74363128 on Build GRCh38
rs878855054 Chr.8: 74362973-74362973 on Build GRCh38
rs563865282 Chr.8: 74363198-74363198 on Build GRCh38
rs946530965 Chr.8: 74363184-74363184 on Build GRCh38
rs561388116 Chr.8: 74363279-74363279 on Build GRCh38
rs981619328 Chr.8: 74363093-74363093 on Build GRCh38
rs567670511 Chr.8: 74362860-74362860 on Build GRCh38
rs188057103 Chr.8: 74363227-74363227 on Build GRCh38
rs765785596 Chr.8: 74363042-74363042 on Build GRCh38
rs928888846 Chr.8: 74363103-74363103 on Build GRCh38
rs754181609 Chr.8: 74362923-74362923 on Build GRCh38
rs752550394 Chr.8: 74363058-74363058 on Build GRCh38
rs780629692 Chr.8: 74362977-74362977 on Build GRCh38
rs5892453 Chr.8: 74363172-74363172 on Build GRCh38
rs897061820 Chr.8: 74363249-74363249 on Build GRCh38
rs1053034169 Chr.8: 74363111-74363111 on Build GRCh38
rs1035267651 Chr.8: 74362830-74362830 on Build GRCh38
rs1040936430 Chr.8: 74363244-74363244 on Build GRCh38
rs921778868 Chr.8: 74363138-74363138 on Build GRCh38
rs267606842 Chr.8: 74363037-74363037 on Build GRCh38
rs549373705 Chr.8: 74362854-74362854 on Build GRCh38
rs960940180 Chr.8: 74362880-74362880 on Build GRCh38
rs765583343 Chr.8: 74363099-74363099 on Build GRCh38
rs532104160 Chr.8: 74363131-74363131 on Build GRCh38
rs116176011 Chr.8: 74363191-74363191 on Build GRCh38
rs372147594 Chr.8: 74363079-74363079 on Build GRCh38
rs999518442 Chr.8: 74363237-74363237 on Build GRCh38
rs765198796 Chr.8: 74363049-74363050 on Build GRCh38
rs140384868 Chr.8: 74362979-74362979 on Build GRCh38
rs535407407 Chr.8: 74362896-74362896 on Build GRCh38
rs185600292 Chr.8: 74363174-74363174 on Build GRCh38
rs1056380664 Chr.8: 74363248-74363248 on Build GRCh38
rs1012447756 Chr.8: 74362912-74362912 on Build GRCh38
rs761939180 Chr.8: 74363057-74363057 on Build GRCh38
rs1019548212 Chr.8: 74362852-74362852 on Build GRCh38
rs1026680509 Chr.8: 74363039-74363039 on Build GRCh38
rs104894075 Chr.8: 74362940-74362940 on Build GRCh38
rs111269356 Chr.8: 74362915-74362915 on Build GRCh38
rs531271794 Chr.8: 74362838-74362838 on Build GRCh38
rs541778476 Chr.8: 74363132-74363132 on Build GRCh38
rs774388702 Chr.8: 74363049-74363049 on Build GRCh38
rs121908113 Chr.8: 74363011-74363011 on Build GRCh38
rs373127785 Chr.8: 74363014-74363014 on Build GRCh38
rs556827873 Chr.8: 74363012-74363012 on Build GRCh38
rs952552386 Chr.8: 74363043-74363043 on Build GRCh38
rs1008061756 Chr.8: 74363219-74363219 on Build GRCh38
rs146768052 Chr.8: 74362976-74362976 on Build GRCh38
rs769102513 Chr.8: 74362996-74362996 on Build GRCh38

View Full Product Details