Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 93754980-93755422 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM67
Entrez Gene ID
91147
Gene Name
transmembrane protein 67
Gene Aliases
JBTS6, MECKELIN, MKS3, NPHP11, TNEM67
UniGene
Hs.116240
Gene Chromosome Location
Chr.8: 93754844-93832653 on Build GRCh38
Transcripts
NM_001142301.1 NM_153704.5 NR_024522.1 XM_006716686.3 XM_011517363.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6181760 c.221G>A p.R74Q Chr.8: 93755135-93755135 on Build GRCh38
3779413 c.207G>C p.Q69H Chr.8: 93755121-93755121 on Build GRCh38
3835143 c.94C>T p.R32C Chr.8: 93755008-93755008 on Build GRCh38
6181762 c.171C>A p.I57I Chr.8: 93755085-93755085 on Build GRCh38
1102476 c.93T>G p.P31P Chr.8: 93755007-93755007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764236301 Chr.8: 93755177-93755177 on Build GRCh38
rs1054088508 Chr.8: 93755006-93755006 on Build GRCh38
rs386834188 Chr.8: 93755075-93755075 on Build GRCh38
rs748980606 Chr.8: 93755176-93755177 on Build GRCh38
rs961425275 Chr.8: 93755182-93755182 on Build GRCh38
rs927200189 Chr.8: 93755207-93755207 on Build GRCh38
rs375493644 Chr.8: 93755104-93755104 on Build GRCh38
rs575077207 Chr.8: 93755011-93755011 on Build GRCh38
rs746906232 Chr.8: 93755096-93755096 on Build GRCh38
rs1016976015 Chr.8: 93755145-93755145 on Build GRCh38
rs754102562 Chr.8: 93755057-93755057 on Build GRCh38
rs945777500 Chr.8: 93755255-93755255 on Build GRCh38
rs998413541 Chr.8: 93755073-93755073 on Build GRCh38
rs983850660 Chr.8: 93755264-93755264 on Build GRCh38
rs552116341 Chr.8: 93755364-93755364 on Build GRCh38
rs767999682 Chr.8: 93755034-93755034 on Build GRCh38
rs199708882 Chr.8: 93755051-93755051 on Build GRCh38
rs138079446 Chr.8: 93755131-93755131 on Build GRCh38
rs747100363 Chr.8: 93755005-93755005 on Build GRCh38
rs986133637 Chr.8: 93755200-93755200 on Build GRCh38
rs912914559 Chr.8: 93755246-93755246 on Build GRCh38
rs953365166 Chr.8: 93755180-93755181 on Build GRCh38
rs146569149 Chr.8: 93755202-93755202 on Build GRCh38
rs369696315 Chr.8: 93755152-93755152 on Build GRCh38
rs897160550 Chr.8: 93755389-93755389 on Build GRCh38
rs774755725 Chr.8: 93755132-93755132 on Build GRCh38
rs267607118 Chr.8: 93755044-93755044 on Build GRCh38
rs890027936 Chr.8: 93755347-93755347 on Build GRCh38
rs115660279 Chr.8: 93755100-93755100 on Build GRCh38
rs976242729 Chr.8: 93755260-93755260 on Build GRCh38
rs994270943 Chr.8: 93755192-93755192 on Build GRCh38
rs752813601 Chr.8: 93755038-93755038 on Build GRCh38
rs772188678 Chr.8: 93755091-93755091 on Build GRCh38
rs886063171 Chr.8: 93755044-93755045 on Build GRCh38
rs755801680 Chr.8: 93755063-93755065 on Build GRCh38
rs934148900 Chr.8: 93755272-93755272 on Build GRCh38
rs561049390 Chr.8: 93755109-93755109 on Build GRCh38
rs780111733 Chr.8: 93755094-93755094 on Build GRCh38
rs144253777 Chr.8: 93755062-93755062 on Build GRCh38
rs532314661 Chr.8: 93755327-93755327 on Build GRCh38
rs74671511 Chr.8: 93755210-93755210 on Build GRCh38
rs759339094 Chr.8: 93755317-93755317 on Build GRCh38
rs760103712 Chr.8: 93755028-93755028 on Build GRCh38
rs1044782419 Chr.8: 93755340-93755340 on Build GRCh38
rs151047825 Chr.8: 93755088-93755088 on Build GRCh38
rs947695516 Chr.8: 93755321-93755321 on Build GRCh38
rs34779331 Chr.8: 93755021-93755021 on Build GRCh38
rs989763965 Chr.8: 93755238-93755238 on Build GRCh38
rs909664649 Chr.8: 93755296-93755296 on Build GRCh38
rs753854958 Chr.8: 93755185-93755185 on Build GRCh38
rs766782158 Chr.8: 93755001-93755001 on Build GRCh38
rs759862721 Chr.8: 93755147-93755147 on Build GRCh38
rs976534512 Chr.8: 93755204-93755204 on Build GRCh38
rs1049947488 Chr.8: 93755314-93755314 on Build GRCh38
rs923077679 Chr.8: 93755263-93755263 on Build GRCh38
rs1024101692 Chr.8: 93755199-93755199 on Build GRCh38
rs1005535447 Chr.8: 93755113-93755113 on Build GRCh38
rs979994320 Chr.8: 93755203-93755203 on Build GRCh38
rs112258105 Chr.8: 93755325-93755325 on Build GRCh38
rs779709768 Chr.8: 93755125-93755126 on Build GRCh38
rs941607037 Chr.8: 93755351-93755351 on Build GRCh38
rs142873030 Chr.8: 93755198-93755198 on Build GRCh38
rs1010962851 Chr.8: 93755162-93755162 on Build GRCh38
rs150383995 Chr.8: 93755106-93755106 on Build GRCh38
rs182449103 Chr.8: 93755380-93755380 on Build GRCh38
rs761028521 Chr.8: 93755166-93755166 on Build GRCh38
rs778831425 Chr.8: 93755078-93755078 on Build GRCh38
rs753224153 Chr.8: 93755153-93755153 on Build GRCh38
rs762188098 Chr.8: 93755150-93755150 on Build GRCh38

View Full Product Details