Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 96784783-96785281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPQ
Entrez Gene ID
10404
Gene Name
carboxypeptidase Q
Gene Aliases
LDP, PGCP
UniGene
Hs.156178
Gene Chromosome Location
Chr.8: 96645227-97143503 on Build GRCh38
Transcripts
NM_016134.3 XM_017012953.1 XM_017012954.1 XM_017012955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1674019 c.308T>G p.V103G Chr.8: 96785205-96785205 on Build GRCh38
4839358 c.202G>A p.A68T Chr.8: 96785099-96785099 on Build GRCh38
5951922 c.27C>T p.F9F Chr.8: 96784924-96784924 on Build GRCh38
6634757 c.232C>A p.L78M Chr.8: 96785129-96785129 on Build GRCh38
160725 c.333C>G p.P111P Chr.8: 96785230-96785230 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971389109 Chr.8: 96785105-96785105 on Build GRCh38
rs763907056 Chr.8: 96784882-96784882 on Build GRCh38
rs746213268 Chr.8: 96785065-96785065 on Build GRCh38
rs145221540 Chr.8: 96785257-96785257 on Build GRCh38
rs565595387 Chr.8: 96784964-96784964 on Build GRCh38
rs1046210621 Chr.8: 96784808-96784808 on Build GRCh38
rs756399625 Chr.8: 96785026-96785027 on Build GRCh38
rs773178240 Chr.8: 96785109-96785109 on Build GRCh38
rs748190900 Chr.8: 96785205-96785205 on Build GRCh38
rs916933272 Chr.8: 96784950-96784960 on Build GRCh38
rs751689686 Chr.8: 96785184-96785184 on Build GRCh38
rs372650907 Chr.8: 96785243-96785243 on Build GRCh38
rs776801209 Chr.8: 96785094-96785094 on Build GRCh38
rs769931993 Chr.8: 96785213-96785213 on Build GRCh38
rs903283339 Chr.8: 96784831-96784831 on Build GRCh38
rs534614802 Chr.8: 96784967-96784967 on Build GRCh38
rs761812940 Chr.8: 96784969-96784969 on Build GRCh38
rs768943731 Chr.8: 96785175-96785175 on Build GRCh38
rs780125284 Chr.8: 96785088-96785088 on Build GRCh38
rs191258208 Chr.8: 96784900-96784900 on Build GRCh38
rs774811444 Chr.8: 96785208-96785209 on Build GRCh38
rs781314881 Chr.8: 96785203-96785203 on Build GRCh38
rs766899037 Chr.8: 96785142-96785142 on Build GRCh38
rs761498745 Chr.8: 96784898-96784898 on Build GRCh38
rs758074689 Chr.8: 96784912-96784912 on Build GRCh38
rs765988969 Chr.8: 96785000-96785000 on Build GRCh38
rs752275268 Chr.8: 96785144-96785144 on Build GRCh38
rs751024063 Chr.8: 96785170-96785170 on Build GRCh38
rs373100921 Chr.8: 96785002-96785002 on Build GRCh38
rs769693736 Chr.8: 96785097-96785097 on Build GRCh38
rs770568459 Chr.8: 96785222-96785222 on Build GRCh38
rs959156328 Chr.8: 96784938-96784938 on Build GRCh38
rs900644565 Chr.8: 96784824-96784824 on Build GRCh38
rs747760951 Chr.8: 96785093-96785093 on Build GRCh38
rs900697169 Chr.8: 96785030-96785030 on Build GRCh38
rs199912422 Chr.8: 96785145-96785145 on Build GRCh38
rs11538936 Chr.8: 96785197-96785197 on Build GRCh38
rs114593053 Chr.8: 96784985-96784985 on Build GRCh38
rs907534090 Chr.8: 96785250-96785250 on Build GRCh38
rs61734395 Chr.8: 96785003-96785003 on Build GRCh38
rs764969018 Chr.8: 96784974-96784974 on Build GRCh38
rs745946672 Chr.8: 96784919-96784919 on Build GRCh38
rs150845839 Chr.8: 96784836-96784836 on Build GRCh38
rs759062668 Chr.8: 96785255-96785255 on Build GRCh38
rs920409961 Chr.8: 96785192-96785192 on Build GRCh38
rs747267124 Chr.8: 96785092-96785092 on Build GRCh38
rs760611528 Chr.8: 96784879-96784879 on Build GRCh38
rs760311225 Chr.8: 96785161-96785161 on Build GRCh38
rs201861702 Chr.8: 96784935-96784935 on Build GRCh38
rs908913270 Chr.8: 96785004-96785004 on Build GRCh38
rs1034414828 Chr.8: 96785143-96785143 on Build GRCh38
rs758899042 Chr.8: 96785076-96785076 on Build GRCh38
rs760249789 Chr.8: 96785202-96785202 on Build GRCh38
rs754233930 Chr.8: 96785019-96785019 on Build GRCh38
rs142316711 Chr.8: 96784816-96784816 on Build GRCh38
rs893380227 Chr.8: 96784842-96784842 on Build GRCh38
rs748555287 Chr.8: 96785253-96785254 on Build GRCh38
rs183172032 Chr.8: 96784916-96784916 on Build GRCh38
rs755247748 Chr.8: 96785191-96785191 on Build GRCh38
rs1026470145 Chr.8: 96784995-96784995 on Build GRCh38
rs1007730288 Chr.8: 96784901-96784901 on Build GRCh38
rs200887903 Chr.8: 96784894-96784894 on Build GRCh38
rs759233915 Chr.8: 96785127-96785127 on Build GRCh38
rs747054913 Chr.8: 96784956-96784956 on Build GRCh38
rs780846696 Chr.8: 96785089-96785089 on Build GRCh38
rs773819729 Chr.8: 96785123-96785123 on Build GRCh38
rs765027440 Chr.8: 96784899-96784899 on Build GRCh38
rs772446810 Chr.8: 96784944-96784944 on Build GRCh38
rs145622173 Chr.8: 96785045-96785045 on Build GRCh38
rs34330149 Chr.8: 96785227-96785227 on Build GRCh38
rs762736003 Chr.8: 96785112-96785112 on Build GRCh38
rs765822863 Chr.8: 96784915-96784915 on Build GRCh38
rs780470025 Chr.8: 96785183-96785183 on Build GRCh38
rs781070792 Chr.8: 96784890-96784890 on Build GRCh38
rs575239324 Chr.8: 96784832-96784832 on Build GRCh38
rs746048796 Chr.8: 96784939-96784939 on Build GRCh38
rs148891768 Chr.8: 96785214-96785214 on Build GRCh38
rs149981793 Chr.8: 96785171-96785171 on Build GRCh38
rs770820512 Chr.8: 96785113-96785113 on Build GRCh38
rs369831003 Chr.8: 96784925-96784925 on Build GRCh38
rs983351964 Chr.8: 96785231-96785231 on Build GRCh38
rs866659130 Chr.8: 96785025-96785025 on Build GRCh38
rs780479623 Chr.8: 96785033-96785033 on Build GRCh38
rs748027162 Chr.8: 96785096-96785096 on Build GRCh38
rs775821667 Chr.8: 96784951-96784951 on Build GRCh38
rs746815323 Chr.8: 96784924-96784924 on Build GRCh38
rs773010520 Chr.8: 96784977-96784977 on Build GRCh38
rs543781934 Chr.8: 96785059-96785060 on Build GRCh38
rs767070225 Chr.8: 96785008-96785008 on Build GRCh38
rs201719687 Chr.8: 96784889-96784890 on Build GRCh38
rs569818798 Chr.8: 96784825-96784825 on Build GRCh38
rs765833957 Chr.8: 96785165-96785165 on Build GRCh38
rs749240539 Chr.8: 96785218-96785218 on Build GRCh38
rs757870995 Chr.8: 96785041-96785041 on Build GRCh38

View Full Product Details