Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 103377570-103378109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTHRC1
Entrez Gene ID
115908
Gene Name
collagen triple helix repeat containing 1
Gene Aliases
-
UniGene
Hs.405614
Gene Chromosome Location
Chr.8: 103371515-103383005 on Build GRCh38
Transcripts
NM_001256099.1 NM_138455.3 XM_011516824.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6637429 c.353G>A p.R118H Chr.8: 103378049-103378049 on Build GRCh38
3412621 c.394C>T p.R132W Chr.8: 103378090-103378090 on Build GRCh38
1202577 c.376T>C p.C126R Chr.8: 103378030-103378030 on Build GRCh38
6637431 c.400A>C p.K134Q Chr.8: 103378096-103378096 on Build GRCh38
5408496 c.396G>A p.R132R Chr.8: 103378092-103378092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746538268 Chr.8: 103378001-103378001 on Build GRCh38
rs765386910 Chr.8: 103378026-103378026 on Build GRCh38
rs369875487 Chr.8: 103377982-103377982 on Build GRCh38
rs1017160947 Chr.8: 103377753-103377753 on Build GRCh38
rs754161448 Chr.8: 103378015-103378015 on Build GRCh38
rs147914468 Chr.8: 103377605-103377609 on Build GRCh38
rs1047906999 Chr.8: 103377868-103377868 on Build GRCh38
rs1012218154 Chr.8: 103377909-103377909 on Build GRCh38
rs62525557 Chr.8: 103377729-103377729 on Build GRCh38
rs754897420 Chr.8: 103378074-103378074 on Build GRCh38
rs7820071 Chr.8: 103377707-103377707 on Build GRCh38
rs573348995 Chr.8: 103377930-103377930 on Build GRCh38
rs183927233 Chr.8: 103377900-103377900 on Build GRCh38
rs1042466591 Chr.8: 103377705-103377705 on Build GRCh38
rs914504755 Chr.8: 103377826-103377826 on Build GRCh38
rs546285910 Chr.8: 103377749-103377749 on Build GRCh38
rs757475049 Chr.8: 103378018-103378018 on Build GRCh38
rs964686051 Chr.8: 103377636-103377636 on Build GRCh38
rs986864277 Chr.8: 103377792-103377792 on Build GRCh38
rs374472802 Chr.8: 103378078-103378078 on Build GRCh38
rs924989138 Chr.8: 103377824-103377824 on Build GRCh38
rs570961809 Chr.8: 103377752-103377752 on Build GRCh38
rs549284598 Chr.8: 103377715-103377715 on Build GRCh38
rs188720010 Chr.8: 103377901-103377901 on Build GRCh38
rs376856799 Chr.8: 103378002-103378002 on Build GRCh38
rs750559913 Chr.8: 103378048-103378048 on Build GRCh38
rs747347935 Chr.8: 103377658-103377658 on Build GRCh38
rs556503633 Chr.8: 103377853-103377853 on Build GRCh38
rs962824408 Chr.8: 103377760-103377760 on Build GRCh38
rs911127968 Chr.8: 103377633-103377633 on Build GRCh38
rs747332572 Chr.8: 103378013-103378014 on Build GRCh38
rs368630680 Chr.8: 103377821-103377821 on Build GRCh38
rs371151606 Chr.8: 103377986-103377986 on Build GRCh38
rs562882412 Chr.8: 103377691-103377691 on Build GRCh38
rs1050787190 Chr.8: 103377628-103377628 on Build GRCh38
rs955191171 Chr.8: 103377780-103377780 on Build GRCh38
rs1054686021 Chr.8: 103377846-103377846 on Build GRCh38
rs370183649 Chr.8: 103378063-103378063 on Build GRCh38
rs78792460 Chr.8: 103377643-103377643 on Build GRCh38
rs746901634 Chr.8: 103378069-103378069 on Build GRCh38
rs571309198 Chr.8: 103377899-103377899 on Build GRCh38
rs146824354 Chr.8: 103377716-103377716 on Build GRCh38
rs367864557 Chr.8: 103377680-103377680 on Build GRCh38
rs962935829 Chr.8: 103377934-103377934 on Build GRCh38
rs538355519 Chr.8: 103377776-103377776 on Build GRCh38
rs541621041 Chr.8: 103377885-103377885 on Build GRCh38
rs986652836 Chr.8: 103377735-103377735 on Build GRCh38
rs892997499 Chr.8: 103377672-103377672 on Build GRCh38
rs1030298342 Chr.8: 103377979-103377979 on Build GRCh38
rs112907293 Chr.8: 103377801-103377801 on Build GRCh38
rs189716470 Chr.8: 103377855-103377855 on Build GRCh38
rs972884295 Chr.8: 103377768-103377768 on Build GRCh38
rs1010090950 Chr.8: 103377673-103377673 on Build GRCh38
rs2687364 Chr.8: 103377996-103377996 on Build GRCh38
rs1016300129 Chr.8: 103377635-103377635 on Build GRCh38
rs963678117 Chr.8: 103378049-103378049 on Build GRCh38
rs937672024 Chr.8: 103377845-103377845 on Build GRCh38
rs902591688 Chr.8: 103377870-103377870 on Build GRCh38
rs753750190 Chr.8: 103377958-103377958 on Build GRCh38
rs74728199 Chr.8: 103377966-103377966 on Build GRCh38
rs910917971 Chr.8: 103377800-103377800 on Build GRCh38
rs933618953 Chr.8: 103377618-103377618 on Build GRCh38
rs1025846410 Chr.8: 103377962-103377962 on Build GRCh38
rs1054349943 Chr.8: 103377819-103377819 on Build GRCh38
rs913700667 Chr.8: 103377757-103377757 on Build GRCh38
rs78566953 Chr.8: 103377642-103377642 on Build GRCh38
rs900535519 Chr.8: 103377946-103377946 on Build GRCh38
rs141335365 Chr.8: 103378059-103378059 on Build GRCh38
rs1054276849 Chr.8: 103377666-103377666 on Build GRCh38
rs966945057 Chr.8: 103378031-103378031 on Build GRCh38
rs896242095 Chr.8: 103377851-103377851 on Build GRCh38

View Full Product Details