Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 106684168-106684676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OXR1
Entrez Gene ID
55074
Gene Name
oxidation resistance 1
Gene Aliases
Nbla00307, TLDC3
UniGene
Hs.127286
Gene Chromosome Location
Chr.8: 106270178-106752694 on Build GRCh38
Transcripts
NM_001198532.1 NM_001198533.1 NM_018002.3 NM_181354.4 XM_006716595.2 XM_017013589.1 XM_017013590.1 XM_017013591.1 XM_017013592.1 XM_017013593.1 XM_017013594.1 XM_017013595.1 XM_017013596.1 XM_017013597.1 XM_017013598.1 XM_017013599.1 XM_017013600.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5904583 c.278G>A p.S93N Chr.8: 106684313-106684313 on Build GRCh38
5889055 c.217C>T p.P73S Chr.8: 106684315-106684315 on Build GRCh38
5408588 c.316_317AA>GG p.K106G Chr.8: 106684351-106684352 on Build GRCh38
5408585 c.517_518AA>GG p.K173G Chr.8: 106684351-106684352 on Build GRCh38
5904582 c.215G>A p.S72N Chr.8: 106684313-106684313 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs983695247 Chr.8: 106684599-106684599 on Build GRCh38
rs564838320 Chr.8: 106684273-106684273 on Build GRCh38
rs760710638 Chr.8: 106684279-106684279 on Build GRCh38
rs531264087 Chr.8: 106684416-106684416 on Build GRCh38
rs189539780 Chr.8: 106684224-106684224 on Build GRCh38
rs1051710949 Chr.8: 106684418-106684418 on Build GRCh38
rs774113408 Chr.8: 106684264-106684264 on Build GRCh38
rs1033542502 Chr.8: 106684520-106684520 on Build GRCh38
rs773003380 Chr.8: 106684277-106684277 on Build GRCh38
rs756577195 Chr.8: 106684327-106684327 on Build GRCh38
rs549711491 Chr.8: 106684421-106684421 on Build GRCh38
rs779484745 Chr.8: 106684388-106684388 on Build GRCh38
rs368799864 Chr.8: 106684243-106684243 on Build GRCh38
rs563947626 Chr.8: 106684413-106684413 on Build GRCh38
rs991811203 Chr.8: 106684219-106684219 on Build GRCh38
rs1039601303 Chr.8: 106684614-106684614 on Build GRCh38
rs571699162 Chr.8: 106684535-106684535 on Build GRCh38
rs967883551 Chr.8: 106684518-106684518 on Build GRCh38
rs547064249 Chr.8: 106684501-106684501 on Build GRCh38
rs944542725 Chr.8: 106684449-106684449 on Build GRCh38
rs767313116 Chr.8: 106684567-106684567 on Build GRCh38
rs1001089605 Chr.8: 106684519-106684519 on Build GRCh38
rs752009772 Chr.8: 106684368-106684368 on Build GRCh38
rs959104185 Chr.8: 106684522-106684522 on Build GRCh38
rs750783682 Chr.8: 106684251-106684251 on Build GRCh38
rs917550418 Chr.8: 106684554-106684554 on Build GRCh38
rs866707745 Chr.8: 106684325-106684325 on Build GRCh38
rs1012383424 Chr.8: 106684642-106684642 on Build GRCh38
rs918572840 Chr.8: 106684377-106684377 on Build GRCh38
rs757788659 Chr.8: 106684387-106684387 on Build GRCh38
rs1035187665 Chr.8: 106684612-106684612 on Build GRCh38
rs765362819 Chr.8: 106684341-106684341 on Build GRCh38
rs768314744 Chr.8: 106684260-106684260 on Build GRCh38
rs138903311 Chr.8: 106684309-106684309 on Build GRCh38
rs764565606 Chr.8: 106684366-106684366 on Build GRCh38
rs768079163 Chr.8: 106684308-106684308 on Build GRCh38
rs142837359 Chr.8: 106684358-106684358 on Build GRCh38
rs1484416 Chr.8: 106684245-106684245 on Build GRCh38
rs538629559 Chr.8: 106684637-106684637 on Build GRCh38
rs760605302 Chr.8: 106684367-106684368 on Build GRCh38
rs766527237 Chr.8: 106684298-106684298 on Build GRCh38
rs747552984 Chr.8: 106684215-106684215 on Build GRCh38
rs776854403 Chr.8: 106684305-106684305 on Build GRCh38
rs942324835 Chr.8: 106684613-106684613 on Build GRCh38
rs896203017 Chr.8: 106684640-106684640 on Build GRCh38
rs76510007 Chr.8: 106684411-106684411 on Build GRCh38
rs867205087 Chr.8: 106684492-106684492 on Build GRCh38
rs778135940 Chr.8: 106684198-106684198 on Build GRCh38
rs183420156 Chr.8: 106684473-106684473 on Build GRCh38
rs999324479 Chr.8: 106684576-106684576 on Build GRCh38
rs946596898 Chr.8: 106684493-106684493 on Build GRCh38
rs1009451797 Chr.8: 106684472-106684472 on Build GRCh38
rs766745028 Chr.8: 106684389-106684389 on Build GRCh38
rs974064549 Chr.8: 106684360-106684360 on Build GRCh38
rs929543399 Chr.8: 106684226-106684226 on Build GRCh38
rs747894407 Chr.8: 106684271-106684271 on Build GRCh38
rs148571201 Chr.8: 106684462-106684462 on Build GRCh38
rs900437137 Chr.8: 106684469-106684469 on Build GRCh38
rs1484417 Chr.8: 106684338-106684338 on Build GRCh38
rs902171656 Chr.8: 106684561-106684561 on Build GRCh38
rs753029341 Chr.8: 106684357-106684357 on Build GRCh38
rs1023735758 Chr.8: 106684645-106684645 on Build GRCh38

View Full Product Details