Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 119758067-119758573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAF2
Entrez Gene ID
6873
Gene Name
TATA-box binding protein associated factor 2
Gene Aliases
CIF150, MRT40, TAF2B, TAFII150
UniGene
Hs.122752
Gene Chromosome Location
Chr.8: 119730774-119832858 on Build GRCh38
Transcripts
NM_003184.3 XM_006716621.3 XM_011517259.2 XM_017013779.1 XM_017013780.1 XM_017013781.1 XM_017013782.1 XM_017013783.1 XM_017013784.1 XM_017013785.1 XM_017013786.1 XM_017013787.1 XM_017013788.1 XM_017013789.1 XM_017013790.1 XM_017013791.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3778859 c.2739G>A p.M913I Chr.8: 119758102-119758102 on Build GRCh38
749562 c.2762A>C p.Y921S Chr.8: 119758079-119758079 on Build GRCh38
486016 c.2724A>G p.Q908Q Chr.8: 119758117-119758117 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs137940227 Chr.8: 119758132-119758132 on Build GRCh38
rs998635863 Chr.8: 119758354-119758354 on Build GRCh38
rs10106425 Chr.8: 119758303-119758303 on Build GRCh38
rs374659090 Chr.8: 119758131-119758131 on Build GRCh38
rs942733390 Chr.8: 119758116-119758116 on Build GRCh38
rs538306795 Chr.8: 119758285-119758285 on Build GRCh38
rs368912229 Chr.8: 119758150-119758150 on Build GRCh38
rs754407747 Chr.8: 119758151-119758155 on Build GRCh38
rs1024437485 Chr.8: 119758323-119758323 on Build GRCh38
rs766100451 Chr.8: 119758099-119758099 on Build GRCh38
rs907003201 Chr.8: 119758342-119758342 on Build GRCh38
rs776027646 Chr.8: 119758178-119758178 on Build GRCh38
rs954848163 Chr.8: 119758471-119758471 on Build GRCh38
rs540458680 Chr.8: 119758361-119758361 on Build GRCh38
rs562698998 Chr.8: 119758546-119758546 on Build GRCh38
rs770114326 Chr.8: 119758162-119758162 on Build GRCh38
rs1013436240 Chr.8: 119758253-119758253 on Build GRCh38
rs943361377 Chr.8: 119758161-119758161 on Build GRCh38
rs368245311 Chr.8: 119758251-119758251 on Build GRCh38
rs753170885 Chr.8: 119758316-119758316 on Build GRCh38
rs150316992 Chr.8: 119758117-119758117 on Build GRCh38
rs544584998 Chr.8: 119758480-119758480 on Build GRCh38
rs1031493458 Chr.8: 119758356-119758356 on Build GRCh38
rs990448166 Chr.8: 119758376-119758377 on Build GRCh38
rs753619496 Chr.8: 119758103-119758103 on Build GRCh38
rs574041212 Chr.8: 119758408-119758408 on Build GRCh38
rs747203981 Chr.8: 119758139-119758139 on Build GRCh38
rs958311355 Chr.8: 119758409-119758409 on Build GRCh38
rs751864705 Chr.8: 119758159-119758159 on Build GRCh38
rs771131068 Chr.8: 119758144-119758144 on Build GRCh38
rs777998983 Chr.8: 119758137-119758137 on Build GRCh38
rs113998740 Chr.8: 119758207-119758207 on Build GRCh38
rs886904032 Chr.8: 119758236-119758236 on Build GRCh38
rs1009735615 Chr.8: 119758424-119758424 on Build GRCh38
rs377232078 Chr.8: 119758413-119758413 on Build GRCh38
rs1038132131 Chr.8: 119758167-119758167 on Build GRCh38
rs746203810 Chr.8: 119758160-119758160 on Build GRCh38
rs778508682 Chr.8: 119758190-119758191 on Build GRCh38
rs975440967 Chr.8: 119758522-119758522 on Build GRCh38
rs573066312 Chr.8: 119758313-119758313 on Build GRCh38
rs374603580 Chr.8: 119758194-119758194 on Build GRCh38
rs987618504 Chr.8: 119758478-119758478 on Build GRCh38
rs1021170158 Chr.8: 119758428-119758428 on Build GRCh38
rs756692926 Chr.8: 119758548-119758548 on Build GRCh38
rs755324125 Chr.8: 119758302-119758302 on Build GRCh38
rs373917557 Chr.8: 119758104-119758104 on Build GRCh38
rs965187676 Chr.8: 119758514-119758514 on Build GRCh38
rs957265258 Chr.8: 119758363-119758363 on Build GRCh38
rs555618777 Chr.8: 119758404-119758404 on Build GRCh38
rs749803836 Chr.8: 119758129-119758130 on Build GRCh38
rs964355340 Chr.8: 119758509-119758509 on Build GRCh38
rs1001863221 Chr.8: 119758339-119758339 on Build GRCh38
rs758606267 Chr.8: 119758133-119758133 on Build GRCh38
rs760306871 Chr.8: 119758098-119758098 on Build GRCh38

View Full Product Details