Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 141422116-141422616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTP4A3
Entrez Gene ID
11156
Gene Name
protein tyrosine phosphatase type IVA, member 3
Gene Aliases
PRL-3, PRL-R, PRL3
UniGene
Hs.43666
Gene Chromosome Location
Chr.8: 141391993-141432454 on Build GRCh38
Transcripts
NM_007079.3 NM_032611.2 XM_005250764.2 XM_006716500.2 XM_011516803.1 XM_011516805.1 XM_017012998.1 XM_017012999.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5041861 c.41A>T p.Y14F Chr.8: 141422281-141422281 on Build GRCh38
3897252 c.53G>A p.R18H Chr.8: 141422293-141422293 on Build GRCh38
3319951 c.27G>A p.P9P Chr.8: 141422267-141422267 on Build GRCh38
749865 c.100A>C p.I34L Chr.8: 141422340-141422340 on Build GRCh38
1096795 c.20C>T p.P7L Chr.8: 141422260-141422260 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540849350 Chr.8: 141422421-141422421 on Build GRCh38
rs267601805 Chr.8: 141422339-141422339 on Build GRCh38
rs1014035876 Chr.8: 141422406-141422406 on Build GRCh38
rs773026145 Chr.8: 141422203-141422203 on Build GRCh38
rs767688987 Chr.8: 141422270-141422270 on Build GRCh38
rs201900679 Chr.8: 141422358-141422358 on Build GRCh38
rs368266833 Chr.8: 141422191-141422191 on Build GRCh38
rs777497023 Chr.8: 141422312-141422312 on Build GRCh38
rs890683359 Chr.8: 141422413-141422413 on Build GRCh38
rs146539294 Chr.8: 141422562-141422562 on Build GRCh38
rs183258262 Chr.8: 141422141-141422141 on Build GRCh38
rs758182631 Chr.8: 141422311-141422311 on Build GRCh38
rs764412569 Chr.8: 141422264-141422264 on Build GRCh38
rs199642324 Chr.8: 141422230-141422230 on Build GRCh38
rs199731261 Chr.8: 141422273-141422273 on Build GRCh38
rs373678370 Chr.8: 141422215-141422215 on Build GRCh38
rs780282009 Chr.8: 141422292-141422292 on Build GRCh38
rs980347201 Chr.8: 141422455-141422455 on Build GRCh38
rs961006533 Chr.8: 141422403-141422403 on Build GRCh38
rs925802988 Chr.8: 141422578-141422578 on Build GRCh38
rs766410960 Chr.8: 141422389-141422389 on Build GRCh38
rs762472480 Chr.8: 141422267-141422267 on Build GRCh38
rs777756341 Chr.8: 141422442-141422442 on Build GRCh38
rs957298867 Chr.8: 141422489-141422489 on Build GRCh38
rs1054232230 Chr.8: 141422399-141422399 on Build GRCh38
rs765575889 Chr.8: 141422217-141422217 on Build GRCh38
rs367738731 Chr.8: 141422231-141422231 on Build GRCh38
rs557905567 Chr.8: 141422435-141422435 on Build GRCh38
rs372301414 Chr.8: 141422228-141422228 on Build GRCh38
rs7018018 Chr.8: 141422512-141422512 on Build GRCh38
rs773886596 Chr.8: 141422366-141422366 on Build GRCh38
rs144048289 Chr.8: 141422260-141422260 on Build GRCh38
rs945868380 Chr.8: 141422341-141422341 on Build GRCh38
rs992702314 Chr.8: 141422313-141422313 on Build GRCh38
rs200609580 Chr.8: 141422261-141422261 on Build GRCh38
rs376532003 Chr.8: 141422257-141422257 on Build GRCh38
rs917176250 Chr.8: 141422541-141422541 on Build GRCh38
rs888584983 Chr.8: 141422590-141422590 on Build GRCh38
rs1005771161 Chr.8: 141422361-141422361 on Build GRCh38
rs879787096 Chr.8: 141422420-141422420 on Build GRCh38
rs201548699 Chr.8: 141422238-141422238 on Build GRCh38
rs963465322 Chr.8: 141422423-141422423 on Build GRCh38
rs752072890 Chr.8: 141422266-141422266 on Build GRCh38
rs201532208 Chr.8: 141422221-141422221 on Build GRCh38
rs888702341 Chr.8: 141422317-141422317 on Build GRCh38
rs932711618 Chr.8: 141422558-141422558 on Build GRCh38
rs1041169271 Chr.8: 141422526-141422526 on Build GRCh38
rs746936080 Chr.8: 141422319-141422319 on Build GRCh38
rs1057370260 Chr.8: 141422157-141422157 on Build GRCh38
rs187792729 Chr.8: 141422241-141422241 on Build GRCh38
rs572703439 Chr.8: 141422494-141422494 on Build GRCh38
rs371438862 Chr.8: 141422233-141422233 on Build GRCh38
rs949509610 Chr.8: 141422164-141422164 on Build GRCh38
rs771985683 Chr.8: 141422202-141422202 on Build GRCh38
rs763970522 Chr.8: 141422183-141422183 on Build GRCh38
rs1015062680 Chr.8: 141422418-141422418 on Build GRCh38
rs754058110 Chr.8: 141422300-141422300 on Build GRCh38
rs755214303 Chr.8: 141422395-141422395 on Build GRCh38
rs922674340 Chr.8: 141422557-141422557 on Build GRCh38
rs28712476 Chr.8: 141422159-141422159 on Build GRCh38
rs780153774 Chr.8: 141422235-141422235 on Build GRCh38
rs945355822 Chr.8: 141422517-141422517 on Build GRCh38
rs750471229 Chr.8: 141422279-141422279 on Build GRCh38
rs937234422 Chr.8: 141422148-141422148 on Build GRCh38
rs752082490 Chr.8: 141422263-141422263 on Build GRCh38
rs753052041 Chr.8: 141422219-141422219 on Build GRCh38
rs769055803 Chr.8: 141422357-141422357 on Build GRCh38
rs769213289 Chr.8: 141422335-141422335 on Build GRCh38
rs749069884 Chr.8: 141422485-141422485 on Build GRCh38
rs992729950 Chr.8: 141422514-141422514 on Build GRCh38
rs924044983 Chr.8: 141422364-141422366 on Build GRCh38
rs765923893 Chr.8: 141422214-141422214 on Build GRCh38
rs781116529 Chr.8: 141422321-141422321 on Build GRCh38
rs1005218527 Chr.8: 141422142-141422142 on Build GRCh38
rs992957141 Chr.8: 141422220-141422220 on Build GRCh38
rs762551334 Chr.8: 141422347-141422347 on Build GRCh38
rs994587522 Chr.8: 141422441-141422441 on Build GRCh38
rs376464093 Chr.8: 141422239-141422239 on Build GRCh38
rs1005074407 Chr.8: 141422416-141422416 on Build GRCh38
rs760787537 Chr.8: 141422386-141422386 on Build GRCh38
rs571742056 Chr.8: 141422320-141422320 on Build GRCh38
rs745344665 Chr.8: 141422322-141422322 on Build GRCh38
rs752025617 Chr.8: 141422227-141422227 on Build GRCh38
rs1024470047 Chr.8: 141422417-141422417 on Build GRCh38
rs775183329 Chr.8: 141422340-141422340 on Build GRCh38
rs1052004880 Chr.8: 141422410-141422410 on Build GRCh38
rs867220700 Chr.8: 141422326-141422326 on Build GRCh38
rs371648082 Chr.8: 141422247-141422247 on Build GRCh38
rs1030546962 Chr.8: 141422238-141422238 on Build GRCh38
rs764624749 Chr.8: 141422222-141422222 on Build GRCh38
rs369508086 Chr.8: 141422364-141422364 on Build GRCh38
rs750117379 Chr.8: 141422591-141422591 on Build GRCh38
rs945482557 Chr.8: 141422256-141422256 on Build GRCh38
rs754086412 Chr.8: 141422392-141422392 on Build GRCh38
rs760184262 Chr.8: 141422207-141422207 on Build GRCh38
rs374921000 Chr.8: 141422237-141422237 on Build GRCh38

View Full Product Details