Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 14735246-14735754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FREM1
Entrez Gene ID
158326
Gene Name
FRAS1 related extracellular matrix 1
Gene Aliases
BNAR, C9orf143, C9orf145, C9orf154, MOTA, TILRR, TRIGNO2
UniGene
Hs.50850
Gene Chromosome Location
Chr.9: 14734666-14911653 on Build GRCh38
Transcripts
NM_001177704.1 NM_144966.5 XM_005251382.3 XM_005251384.4 XM_006716729.3 XM_011517758.2 XM_017014316.1 XM_017014317.1 XM_017014318.1 XM_017014319.1 XM_017014320.1 XM_017014321.1 XM_017014322.1 XM_017014323.1 XM_017014324.1 XM_017014325.1 XM_017014326.1 XM_017014327.1

SNP Detail

SNP ID Chromosome Location
rs1023480059 Chr.9: 14735289-14735289 on Build GRCh38
rs964292307 Chr.9: 14735636-14735636 on Build GRCh38
rs763298549 Chr.9: 14735471-14735474 on Build GRCh38
rs576323077 Chr.9: 14735386-14735386 on Build GRCh38
rs557523204 Chr.9: 14735329-14735329 on Build GRCh38
rs769884958 Chr.9: 14735271-14735271 on Build GRCh38
rs555549310 Chr.9: 14735499-14735499 on Build GRCh38
rs7033520 Chr.9: 14735284-14735284 on Build GRCh38
rs565124897 Chr.9: 14735328-14735328 on Build GRCh38
rs960343292 Chr.9: 14735324-14735324 on Build GRCh38
rs1010341159 Chr.9: 14735569-14735569 on Build GRCh38
rs949704953 Chr.9: 14735476-14735476 on Build GRCh38
rs953704987 Chr.9: 14735595-14735595 on Build GRCh38
rs529192766 Chr.9: 14735653-14735653 on Build GRCh38
rs912025555 Chr.9: 14735604-14735604 on Build GRCh38
rs368893723 Chr.9: 14735348-14735348 on Build GRCh38
rs951515340 Chr.9: 14735680-14735680 on Build GRCh38
rs978783060 Chr.9: 14735368-14735368 on Build GRCh38
rs886063753 Chr.9: 14735572-14735573 on Build GRCh38
rs891124248 Chr.9: 14735590-14735590 on Build GRCh38
rs974271928 Chr.9: 14735659-14735659 on Build GRCh38
rs907540710 Chr.9: 14735441-14735441 on Build GRCh38
rs930412604 Chr.9: 14735392-14735392 on Build GRCh38
rs994567285 Chr.9: 14735277-14735277 on Build GRCh38
rs1494342 Chr.9: 14735531-14735531 on Build GRCh38
rs1009868514 Chr.9: 14735546-14735546 on Build GRCh38
rs10961688 Chr.9: 14735552-14735552 on Build GRCh38
rs1035371761 Chr.9: 14735543-14735543 on Build GRCh38
rs1020724195 Chr.9: 14735295-14735295 on Build GRCh38
rs907654735 Chr.9: 14735716-14735716 on Build GRCh38
rs575605478 Chr.9: 14735454-14735454 on Build GRCh38
rs892259614 Chr.9: 14735729-14735729 on Build GRCh38
rs776754648 Chr.9: 14735576-14735576 on Build GRCh38
rs528248069 Chr.9: 14735730-14735730 on Build GRCh38
rs751524710 Chr.9: 14735727-14735727 on Build GRCh38
rs10756607 Chr.9: 14735612-14735612 on Build GRCh38
rs549260371 Chr.9: 14735692-14735692 on Build GRCh38
rs1044831988 Chr.9: 14735503-14735503 on Build GRCh38
rs138214905 Chr.9: 14735608-14735608 on Build GRCh38
rs1044862953 Chr.9: 14735684-14735684 on Build GRCh38
rs974247467 Chr.9: 14735359-14735359 on Build GRCh38
rs886063752 Chr.9: 14735327-14735327 on Build GRCh38
rs922106636 Chr.9: 14735377-14735377 on Build GRCh38
rs1054316050 Chr.9: 14735538-14735538 on Build GRCh38
rs892401097 Chr.9: 14735544-14735544 on Build GRCh38
rs764201056 Chr.9: 14735531-14735554 on Build GRCh38
rs955375481 Chr.9: 14735297-14735297 on Build GRCh38
rs1045804803 Chr.9: 14735478-14735478 on Build GRCh38
rs572257381 Chr.9: 14735519-14735519 on Build GRCh38
rs886063754 Chr.9: 14735624-14735624 on Build GRCh38
rs905059340 Chr.9: 14735511-14735511 on Build GRCh38
rs146976330 Chr.9: 14735485-14735485 on Build GRCh38
rs1494343 Chr.9: 14735696-14735696 on Build GRCh38
rs1027122393 Chr.9: 14735663-14735663 on Build GRCh38
rs983191897 Chr.9: 14735420-14735420 on Build GRCh38
rs994590351 Chr.9: 14735494-14735494 on Build GRCh38
rs1056312185 Chr.9: 14735728-14735728 on Build GRCh38
rs556852600 Chr.9: 14735371-14735371 on Build GRCh38
rs760682289 Chr.9: 14735456-14735456 on Build GRCh38
rs988808846 Chr.9: 14735327-14735327 on Build GRCh38
rs932167179 Chr.9: 14735379-14735379 on Build GRCh38
rs536610674 Chr.9: 14735364-14735364 on Build GRCh38
rs1010308669 Chr.9: 14735279-14735279 on Build GRCh38
rs987640611 Chr.9: 14735599-14735599 on Build GRCh38
rs772916234 Chr.9: 14735606-14735606 on Build GRCh38
rs571246149 Chr.9: 14735336-14735336 on Build GRCh38
rs1052480140 Chr.9: 14735380-14735380 on Build GRCh38
rs533397720 Chr.9: 14735574-14735574 on Build GRCh38
rs987469189 Chr.9: 14735298-14735298 on Build GRCh38
rs560870187 Chr.9: 14735573-14735573 on Build GRCh38
rs898898775 Chr.9: 14735275-14735275 on Build GRCh38
rs1034933389 Chr.9: 14735314-14735314 on Build GRCh38

View Full Product Details