Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 19033140-19033670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAXO1
Entrez Gene ID
158297
Gene Name
stabilizer of axonemal microtubules 1
Gene Aliases
C9orf138, FAM154A
UniGene
Hs.731593
Gene Chromosome Location
Chr.9: 18927660-19049354 on Build GRCh38
Transcripts
NM_001287049.1 NM_001287050.1 NM_153707.3 XM_011517744.1 XM_011517745.1 XM_017014314.1 XM_017014315.1

SNP Detail

SNP ID Chromosome Location
rs568657062 Chr.9: 19033321-19033321 on Build GRCh38
rs770015324 Chr.9: 19033630-19033630 on Build GRCh38
rs897570310 Chr.9: 19033278-19033278 on Build GRCh38
rs950775405 Chr.9: 19033354-19033354 on Build GRCh38
rs900531046 Chr.9: 19033558-19033558 on Build GRCh38
rs1027634042 Chr.9: 19033564-19033564 on Build GRCh38
rs372528222 Chr.9: 19033637-19033637 on Build GRCh38
rs546842581 Chr.9: 19033248-19033248 on Build GRCh38
rs753547090 Chr.9: 19033229-19033229 on Build GRCh38
rs950589219 Chr.9: 19033228-19033228 on Build GRCh38
rs893996772 Chr.9: 19033266-19033266 on Build GRCh38
rs772499204 Chr.9: 19033396-19033396 on Build GRCh38
rs116329746 Chr.9: 19033309-19033309 on Build GRCh38
rs971201911 Chr.9: 19033604-19033604 on Build GRCh38
rs777633380 Chr.9: 19033202-19033202 on Build GRCh38
rs570858758 Chr.9: 19033292-19033292 on Build GRCh38
rs945753154 Chr.9: 19033476-19033476 on Build GRCh38
rs567963751 Chr.9: 19033265-19033265 on Build GRCh38
rs570997458 Chr.9: 19033437-19033437 on Build GRCh38
rs58614427 Chr.9: 19033584-19033584 on Build GRCh38
rs964835489 Chr.9: 19033211-19033211 on Build GRCh38
rs918943404 Chr.9: 19033645-19033645 on Build GRCh38
rs7036182 Chr.9: 19033644-19033644 on Build GRCh38
rs535009538 Chr.9: 19033505-19033505 on Build GRCh38
rs866790150 Chr.9: 19033272-19033272 on Build GRCh38
rs917178210 Chr.9: 19033230-19033230 on Build GRCh38
rs935799705 Chr.9: 19033262-19033262 on Build GRCh38
rs982204488 Chr.9: 19033376-19033376 on Build GRCh38
rs926918230 Chr.9: 19033260-19033260 on Build GRCh38
rs893958153 Chr.9: 19033164-19033164 on Build GRCh38
rs920481119 Chr.9: 19033221-19033221 on Build GRCh38
rs1017868103 Chr.9: 19033341-19033341 on Build GRCh38
rs1007788163 Chr.9: 19033163-19033163 on Build GRCh38
rs902493729 Chr.9: 19033574-19033574 on Build GRCh38
rs370372806 Chr.9: 19033635-19033635 on Build GRCh38
rs552432125 Chr.9: 19033179-19033179 on Build GRCh38
rs1020916009 Chr.9: 19033357-19033358 on Build GRCh38
rs369503596 Chr.9: 19033517-19033517 on Build GRCh38
rs915834082 Chr.9: 19033419-19033419 on Build GRCh38
rs915614858 Chr.9: 19033234-19033234 on Build GRCh38
rs1008913763 Chr.9: 19033201-19033201 on Build GRCh38
rs890727560 Chr.9: 19033291-19033291 on Build GRCh38
rs62563561 Chr.9: 19033227-19033227 on Build GRCh38
rs7036118 Chr.9: 19033378-19033378 on Build GRCh38
rs575024443 Chr.9: 19033566-19033566 on Build GRCh38
rs983266445 Chr.9: 19033368-19033368 on Build GRCh38
rs564260145 Chr.9: 19033194-19033194 on Build GRCh38
rs973494601 Chr.9: 19033435-19033435 on Build GRCh38
rs979741413 Chr.9: 19033259-19033259 on Build GRCh38
rs943510939 Chr.9: 19033501-19033501 on Build GRCh38
rs920987308 Chr.9: 19033362-19033362 on Build GRCh38
rs890623123 Chr.9: 19033518-19033518 on Build GRCh38
rs1051506076 Chr.9: 19033462-19033462 on Build GRCh38
rs10963923 Chr.9: 19033606-19033606 on Build GRCh38
rs1009112274 Chr.9: 19033300-19033300 on Build GRCh38
rs954720497 Chr.9: 19033182-19033182 on Build GRCh38
rs991397608 Chr.9: 19033397-19033397 on Build GRCh38
rs912984644 Chr.9: 19033456-19033456 on Build GRCh38
rs894066242 Chr.9: 19033550-19033550 on Build GRCh38
rs1042579766 Chr.9: 19033507-19033507 on Build GRCh38
rs991346981 Chr.9: 19033233-19033233 on Build GRCh38
rs528838527 Chr.9: 19033308-19033308 on Build GRCh38
rs1042549969 Chr.9: 19033273-19033273 on Build GRCh38
rs1021450325 Chr.9: 19033186-19033186 on Build GRCh38
rs780592834 Chr.9: 19033438-19033438 on Build GRCh38
rs1028142570 Chr.9: 19033335-19033335 on Build GRCh38
rs979980182 Chr.9: 19033395-19033395 on Build GRCh38
rs957280420 Chr.9: 19033628-19033628 on Build GRCh38
rs971295366 Chr.9: 19033391-19033391 on Build GRCh38
rs944982589 Chr.9: 19033524-19033524 on Build GRCh38
rs868836577 Chr.9: 19033619-19033619 on Build GRCh38
rs193100926 Chr.9: 19033429-19033429 on Build GRCh38
rs769007861 Chr.9: 19033570-19033570 on Build GRCh38
rs554268931 Chr.9: 19033507-19033507 on Build GRCh38
rs553590951 Chr.9: 19033559-19033559 on Build GRCh38
rs60251915 Chr.9: 19033379-19033379 on Build GRCh38
rs762366702 Chr.9: 19033613-19033614 on Build GRCh38
rs1009860779 Chr.9: 19033168-19033168 on Build GRCh38
rs776920669 Chr.9: 19033594-19033594 on Build GRCh38
rs890618201 Chr.9: 19033323-19033323 on Build GRCh38
rs893846267 Chr.9: 19033347-19033347 on Build GRCh38
rs557371899 Chr.9: 19033600-19033600 on Build GRCh38
rs959862512 Chr.9: 19033389-19033389 on Build GRCh38
rs1007840486 Chr.9: 19033336-19033336 on Build GRCh38
rs747434155 Chr.9: 19033553-19033553 on Build GRCh38
rs773201855 Chr.9: 19033642-19033642 on Build GRCh38
rs1039677303 Chr.9: 19033541-19033541 on Build GRCh38
rs973890673 Chr.9: 19033215-19033215 on Build GRCh38
rs531026017 Chr.9: 19033169-19033169 on Build GRCh38

View Full Product Details