Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 19058265-19058777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAUS6
Entrez Gene ID
54801
Gene Name
HAUS augmin like complex subunit 6
Gene Aliases
Dgt6, FAM29A
UniGene
Hs.533468
Gene Chromosome Location
Chr.9: 19053137-19102942 on Build GRCh38
Transcripts
NM_001270890.1 NM_017645.4 XM_011517935.2 XM_017014830.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7149337 c.2437C>G p.L813V Chr.9: 19058330-19058330 on Build GRCh38
1700975 c.2375C>T p.S792F Chr.9: 19058392-19058392 on Build GRCh38
4918292 c.2298G>T p.K766N Chr.9: 19058469-19058469 on Build GRCh38
6013449 c.2404G>C p.E802Q Chr.9: 19058363-19058363 on Build GRCh38
1107602 c.2443G>T p.D815Y Chr.9: 19058324-19058324 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377309779 Chr.9: 19058428-19058428 on Build GRCh38
rs770193757 Chr.9: 19058304-19058305 on Build GRCh38
rs756887821 Chr.9: 19058429-19058429 on Build GRCh38
rs567621193 Chr.9: 19058544-19058544 on Build GRCh38
rs763219876 Chr.9: 19058681-19058681 on Build GRCh38
rs374948281 Chr.9: 19058658-19058658 on Build GRCh38
rs781522340 Chr.9: 19058592-19058592 on Build GRCh38
rs762842123 Chr.9: 19058486-19058486 on Build GRCh38
rs756480163 Chr.9: 19058374-19058374 on Build GRCh38
rs764389944 Chr.9: 19058426-19058428 on Build GRCh38
rs140508604 Chr.9: 19058389-19058389 on Build GRCh38
rs374074360 Chr.9: 19058704-19058704 on Build GRCh38
rs764332656 Chr.9: 19058749-19058749 on Build GRCh38
rs773856977 Chr.9: 19058500-19058500 on Build GRCh38
rs998915585 Chr.9: 19058463-19058463 on Build GRCh38
rs141308291 Chr.9: 19058436-19058436 on Build GRCh38
rs10511670 Chr.9: 19058745-19058745 on Build GRCh38
rs757268773 Chr.9: 19058575-19058575 on Build GRCh38
rs776427843 Chr.9: 19058422-19058422 on Build GRCh38
rs574489899 Chr.9: 19058659-19058659 on Build GRCh38
rs773565485 Chr.9: 19058315-19058315 on Build GRCh38
rs753748231 Chr.9: 19058322-19058322 on Build GRCh38
rs199727248 Chr.9: 19058476-19058476 on Build GRCh38
rs751984422 Chr.9: 19058688-19058688 on Build GRCh38
rs372663494 Chr.9: 19058743-19058743 on Build GRCh38
rs4977493 Chr.9: 19058485-19058485 on Build GRCh38
rs759679187 Chr.9: 19058302-19058305 on Build GRCh38
rs565164551 Chr.9: 19058742-19058742 on Build GRCh38
rs751893733 Chr.9: 19058467-19058467 on Build GRCh38
rs749784720 Chr.9: 19058380-19058380 on Build GRCh38
rs765656040 Chr.9: 19058535-19058535 on Build GRCh38
rs143374581 Chr.9: 19058736-19058736 on Build GRCh38
rs760910824 Chr.9: 19058416-19058416 on Build GRCh38
rs947654102 Chr.9: 19058671-19058671 on Build GRCh38
rs368360263 Chr.9: 19058304-19058304 on Build GRCh38
rs147144205 Chr.9: 19058744-19058744 on Build GRCh38
rs748633507 Chr.9: 19058445-19058445 on Build GRCh38
rs775389859 Chr.9: 19058426-19058426 on Build GRCh38
rs768573168 Chr.9: 19058616-19058616 on Build GRCh38
rs755443632 Chr.9: 19058526-19058526 on Build GRCh38
rs527919595 Chr.9: 19058473-19058473 on Build GRCh38
rs141501503 Chr.9: 19058431-19058431 on Build GRCh38
rs771461771 Chr.9: 19058639-19058639 on Build GRCh38
rs184378901 Chr.9: 19058450-19058450 on Build GRCh38
rs201410896 Chr.9: 19058439-19058439 on Build GRCh38
rs778960278 Chr.9: 19058731-19058731 on Build GRCh38
rs62619766 Chr.9: 19058638-19058638 on Build GRCh38
rs554017283 Chr.9: 19058701-19058701 on Build GRCh38
rs375476349 Chr.9: 19058393-19058393 on Build GRCh38
rs181118909 Chr.9: 19058721-19058721 on Build GRCh38
rs770652422 Chr.9: 19058421-19058421 on Build GRCh38
rs762141378 Chr.9: 19058297-19058297 on Build GRCh38
rs201374475 Chr.9: 19058613-19058613 on Build GRCh38
rs761378315 Chr.9: 19058508-19058508 on Build GRCh38
rs538119546 Chr.9: 19058571-19058571 on Build GRCh38
rs142781641 Chr.9: 19058715-19058715 on Build GRCh38
rs763198448 Chr.9: 19058374-19058374 on Build GRCh38
rs113547478 Chr.9: 19058600-19058600 on Build GRCh38
rs775826721 Chr.9: 19058317-19058319 on Build GRCh38
rs774152882 Chr.9: 19058623-19058623 on Build GRCh38
rs757894954 Chr.9: 19058357-19058357 on Build GRCh38
rs750669925 Chr.9: 19058710-19058710 on Build GRCh38
rs763537717 Chr.9: 19058490-19058490 on Build GRCh38
rs554778233 Chr.9: 19058725-19058725 on Build GRCh38
rs370946839 Chr.9: 19058329-19058329 on Build GRCh38
rs750031100 Chr.9: 19058517-19058517 on Build GRCh38
rs779622045 Chr.9: 19058440-19058440 on Build GRCh38
rs561181097 Chr.9: 19058379-19058379 on Build GRCh38
rs145639916 Chr.9: 19058398-19058398 on Build GRCh38
rs769550922 Chr.9: 19058483-19058483 on Build GRCh38
rs757394902 Chr.9: 19058699-19058699 on Build GRCh38
rs777456054 Chr.9: 19058365-19058365 on Build GRCh38
rs746062566 Chr.9: 19058595-19058595 on Build GRCh38
rs146530083 Chr.9: 19058366-19058366 on Build GRCh38
rs752136153 Chr.9: 19058750-19058750 on Build GRCh38
rs11515236 Chr.9: 19058427-19058427 on Build GRCh38
rs764717616 Chr.9: 19058685-19058685 on Build GRCh38
rs1023848823 Chr.9: 19058522-19058522 on Build GRCh38
rs747208152 Chr.9: 19058472-19058472 on Build GRCh38
rs951008205 Chr.9: 19058606-19058606 on Build GRCh38
rs542826832 Chr.9: 19058356-19058356 on Build GRCh38
rs764737576 Chr.9: 19058332-19058332 on Build GRCh38
rs191418569 Chr.9: 19058424-19058424 on Build GRCh38
rs753278112 Chr.9: 19058539-19058539 on Build GRCh38
rs919556447 Chr.9: 19058382-19058382 on Build GRCh38
rs748306154 Chr.9: 19058392-19058392 on Build GRCh38
rs767268752 Chr.9: 19058513-19058513 on Build GRCh38
rs753042827 Chr.9: 19058661-19058661 on Build GRCh38
rs768525992 Chr.9: 19058410-19058410 on Build GRCh38
rs138541318 Chr.9: 19058735-19058735 on Build GRCh38
rs755390338 Chr.9: 19058437-19058437 on Build GRCh38
rs758915364 Chr.9: 19058540-19058540 on Build GRCh38
rs749240428 Chr.9: 19058614-19058614 on Build GRCh38
rs767602625 Chr.9: 19058754-19058754 on Build GRCh38
rs762181341 Chr.9: 19058752-19058752 on Build GRCh38
rs762232786 Chr.9: 19058599-19058620 on Build GRCh38
rs189673127 Chr.9: 19058458-19058458 on Build GRCh38
rs142674219 Chr.9: 19058318-19058318 on Build GRCh38
rs368700958 Chr.9: 19058724-19058724 on Build GRCh38

View Full Product Details