Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35829224-35829718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM8B
Entrez Gene ID
51754
Gene Name
transmembrane protein 8B
Gene Aliases
C9orf127, NAG-5, NAG5, NGX6
UniGene
Hs.493808
Gene Chromosome Location
Chr.9: 35825817-35865515 on Build GRCh38
Transcripts
NM_001042589.2 NM_001042590.2 NM_016446.3 XM_005251483.4 XM_011517901.2 XM_011517902.2 XM_011517903.2 XM_011517904.2 XM_011517905.1 XM_011517910.2 XM_011517911.2 XM_011517915.2 XM_017014806.1

SNP Detail

SNP ID Chromosome Location
rs907308915 Chr.9: 35829584-35829584 on Build GRCh38
rs572287590 Chr.9: 35829334-35829334 on Build GRCh38
rs891934858 Chr.9: 35829289-35829289 on Build GRCh38
rs997577564 Chr.9: 35829282-35829282 on Build GRCh38
rs1039163102 Chr.9: 35829249-35829249 on Build GRCh38
rs952104586 Chr.9: 35829261-35829261 on Build GRCh38
rs914280110 Chr.9: 35829406-35829406 on Build GRCh38
rs553876550 Chr.9: 35829560-35829560 on Build GRCh38
rs989825832 Chr.9: 35829399-35829399 on Build GRCh38
rs867023653 Chr.9: 35829369-35829369 on Build GRCh38
rs1010522922 Chr.9: 35829293-35829293 on Build GRCh38
rs1032291578 Chr.9: 35829605-35829605 on Build GRCh38
rs1023061556 Chr.9: 35829274-35829274 on Build GRCh38
rs1031903839 Chr.9: 35829322-35829322 on Build GRCh38
rs1003029377 Chr.9: 35829595-35829595 on Build GRCh38
rs370232891 Chr.9: 35829474-35829474 on Build GRCh38
rs956314630 Chr.9: 35829610-35829610 on Build GRCh38
rs887741413 Chr.9: 35829536-35829536 on Build GRCh38
rs1028103181 Chr.9: 35829252-35829252 on Build GRCh38
rs1022292426 Chr.9: 35829569-35829569 on Build GRCh38
rs897058951 Chr.9: 35829503-35829503 on Build GRCh38
rs956517318 Chr.9: 35829329-35829329 on Build GRCh38
rs900600955 Chr.9: 35829266-35829266 on Build GRCh38
rs186013232 Chr.9: 35829415-35829415 on Build GRCh38
rs912327252 Chr.9: 35829423-35829424 on Build GRCh38
rs779470623 Chr.9: 35829522-35829522 on Build GRCh38
rs932322429 Chr.9: 35829410-35829410 on Build GRCh38
rs2381409 Chr.9: 35829393-35829393 on Build GRCh38
rs1017166127 Chr.9: 35829438-35829438 on Build GRCh38
rs1051935556 Chr.9: 35829286-35829286 on Build GRCh38
rs982034886 Chr.9: 35829322-35829333 on Build GRCh38
rs1023032161 Chr.9: 35829558-35829558 on Build GRCh38
rs527695038 Chr.9: 35829548-35829548 on Build GRCh38
rs1005811929 Chr.9: 35829422-35829422 on Build GRCh38
rs533701131 Chr.9: 35829559-35829559 on Build GRCh38
rs564301843 Chr.9: 35829535-35829535 on Build GRCh38
rs113663575 Chr.9: 35829500-35829500 on Build GRCh38
rs943852405 Chr.9: 35829425-35829425 on Build GRCh38

View Full Product Details