Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 37917151-37917658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHB
Entrez Gene ID
6461
Gene Name
SH2 domain containing adaptor protein B
Gene Aliases
bA3J10.2
UniGene
Hs.521482
Gene Chromosome Location
Chr.9: 37915898-38069213 on Build GRCh38
Transcripts
NM_003028.2

SNP Detail

SNP ID Chromosome Location
rs149274311 Chr.9: 37917476-37917476 on Build GRCh38
rs900086095 Chr.9: 37917195-37917195 on Build GRCh38
rs1029801686 Chr.9: 37917243-37917243 on Build GRCh38
rs373792099 Chr.9: 37917448-37917448 on Build GRCh38
rs762364208 Chr.9: 37917326-37917326 on Build GRCh38
rs531965302 Chr.9: 37917557-37917557 on Build GRCh38
rs917154274 Chr.9: 37917347-37917347 on Build GRCh38
rs111733894 Chr.9: 37917385-37917385 on Build GRCh38
rs1051314110 Chr.9: 37917491-37917491 on Build GRCh38
rs559684824 Chr.9: 37917544-37917544 on Build GRCh38
rs976412037 Chr.9: 37917483-37917483 on Build GRCh38
rs958664371 Chr.9: 37917342-37917342 on Build GRCh38
rs531053258 Chr.9: 37917627-37917627 on Build GRCh38
rs932433403 Chr.9: 37917518-37917518 on Build GRCh38
rs891370114 Chr.9: 37917492-37917492 on Build GRCh38
rs553358654 Chr.9: 37917187-37917187 on Build GRCh38
rs941567073 Chr.9: 37917568-37917568 on Build GRCh38
rs932795301 Chr.9: 37917192-37917192 on Build GRCh38
rs918949139 Chr.9: 37917325-37917325 on Build GRCh38
rs549443742 Chr.9: 37917181-37917181 on Build GRCh38
rs554375511 Chr.9: 37917478-37917478 on Build GRCh38
rs991382047 Chr.9: 37917343-37917343 on Build GRCh38
rs751009731 Chr.9: 37917553-37917553 on Build GRCh38
rs574145996 Chr.9: 37917522-37917522 on Build GRCh38
rs888669050 Chr.9: 37917257-37917257 on Build GRCh38
rs575393084 Chr.9: 37917459-37917459 on Build GRCh38
rs138556078 Chr.9: 37917271-37917271 on Build GRCh38
rs780527447 Chr.9: 37917576-37917576 on Build GRCh38
rs548019837 Chr.9: 37917561-37917561 on Build GRCh38
rs749216816 Chr.9: 37917356-37917356 on Build GRCh38
rs1024381033 Chr.9: 37917334-37917334 on Build GRCh38
rs994967111 Chr.9: 37917556-37917556 on Build GRCh38
rs7028246 Chr.9: 37917605-37917605 on Build GRCh38
rs1037277422 Chr.9: 37917602-37917602 on Build GRCh38
rs1033530058 Chr.9: 37917406-37917406 on Build GRCh38
rs545406028 Chr.9: 37917592-37917592 on Build GRCh38
rs977247499 Chr.9: 37917407-37917408 on Build GRCh38
rs1036997705 Chr.9: 37917283-37917283 on Build GRCh38
rs1049944003 Chr.9: 37917228-37917228 on Build GRCh38
rs997477301 Chr.9: 37917209-37917209 on Build GRCh38
rs753999397 Chr.9: 37917315-37917315 on Build GRCh38
rs922333751 Chr.9: 37917510-37917510 on Build GRCh38
rs891316035 Chr.9: 37917248-37917248 on Build GRCh38
rs765821487 Chr.9: 37917338-37917338 on Build GRCh38
rs764009399 Chr.9: 37917173-37917173 on Build GRCh38
rs77388885 Chr.9: 37917457-37917457 on Build GRCh38
rs377748296 Chr.9: 37917285-37917285 on Build GRCh38
rs1020862465 Chr.9: 37917474-37917474 on Build GRCh38
rs1037566677 Chr.9: 37917525-37917525 on Build GRCh38
rs79534838 Chr.9: 37917455-37917455 on Build GRCh38
rs936045803 Chr.9: 37917447-37917447 on Build GRCh38
rs539862478 Chr.9: 37917524-37917524 on Build GRCh38
rs76942206 Chr.9: 37917458-37917458 on Build GRCh38
rs989359449 Chr.9: 37917473-37917473 on Build GRCh38
rs924760569 Chr.9: 37917413-37917413 on Build GRCh38

View Full Product Details