Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15331390-15331900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIGA
Entrez Gene ID
5277
Gene Name
phosphatidylinositol glycan anchor biosynthesis class A
Gene Aliases
GPI3, MCAHS2, PIG-A, PNH1
UniGene
Hs.137154
Gene Chromosome Location
Chr.X: 15319451-15335554 on Build GRCh38
Transcripts
NM_002641.3 NM_020473.3 NR_033835.1 NR_033836.1 XM_017029581.1

SNP Detail

SNP ID Chromosome Location
rs143565005 Chr.X: 15331493-15331493 on Build GRCh38
rs772668943 Chr.X: 15331718-15331718 on Build GRCh38
rs797044924 Chr.X: 15331833-15331833 on Build GRCh38
rs764210969 Chr.X: 15331757-15331757 on Build GRCh38
rs769478872 Chr.X: 15331714-15331714 on Build GRCh38
rs776453997 Chr.X: 15331475-15331475 on Build GRCh38
rs1006798135 Chr.X: 15331749-15331749 on Build GRCh38
rs769125619 Chr.X: 15331878-15331878 on Build GRCh38
rs762748091 Chr.X: 15331733-15331733 on Build GRCh38
rs777795093 Chr.X: 15331624-15331624 on Build GRCh38
rs1057518239 Chr.X: 15331869-15331869 on Build GRCh38
rs4830937 Chr.X: 15331619-15331619 on Build GRCh38
rs750784049 Chr.X: 15331422-15331423 on Build GRCh38
rs587776727 Chr.X: 15331500-15331500 on Build GRCh38
rs146801580 Chr.X: 15331645-15331645 on Build GRCh38
rs775206996 Chr.X: 15331432-15331432 on Build GRCh38
rs760682734 Chr.X: 15331511-15331511 on Build GRCh38
rs199422233 Chr.X: 15331768-15331768 on Build GRCh38
rs756212857 Chr.X: 15331623-15331623 on Build GRCh38
rs200475470 Chr.X: 15331811-15331811 on Build GRCh38
rs375707235 Chr.X: 15331417-15331417 on Build GRCh38
rs765502766 Chr.X: 15331830-15331830 on Build GRCh38
rs772695695 Chr.X: 15331426-15331426 on Build GRCh38
rs587777400 Chr.X: 15331653-15331653 on Build GRCh38
rs764988988 Chr.X: 15331504-15331504 on Build GRCh38
rs762862377 Chr.X: 15331561-15331561 on Build GRCh38
rs113204630 Chr.X: 15331604-15331604 on Build GRCh38
rs776182358 Chr.X: 15331699-15331699 on Build GRCh38
rs779308669 Chr.X: 15331424-15331424 on Build GRCh38
rs751436894 Chr.X: 15331603-15331603 on Build GRCh38
rs879255543 Chr.X: 15331862-15331863 on Build GRCh38
rs368684591 Chr.X: 15331445-15331445 on Build GRCh38
rs753941641 Chr.X: 15331799-15331799 on Build GRCh38
rs61730284 Chr.X: 15331658-15331658 on Build GRCh38
rs760266545 Chr.X: 15331438-15331438 on Build GRCh38
rs768231310 Chr.X: 15331439-15331439 on Build GRCh38
rs199422232 Chr.X: 15331637-15331637 on Build GRCh38
rs587777398 Chr.X: 15331701-15331701 on Build GRCh38
rs766304305 Chr.X: 15331562-15331562 on Build GRCh38
rs768321389 Chr.X: 15331681-15331681 on Build GRCh38
rs587776724 Chr.X: 15331471-15331472 on Build GRCh38
rs34422225 Chr.X: 15331876-15331876 on Build GRCh38
rs780532806 Chr.X: 15331840-15331840 on Build GRCh38
rs864309579 Chr.X: 15331852-15331852 on Build GRCh38
rs758769627 Chr.X: 15331651-15331651 on Build GRCh38
rs747478493 Chr.X: 15331666-15331666 on Build GRCh38
rs766109827 Chr.X: 15331742-15331742 on Build GRCh38
rs757482806 Chr.X: 15331631-15331631 on Build GRCh38
rs759443446 Chr.X: 15331756-15331756 on Build GRCh38
rs779205409 Chr.X: 15331640-15331640 on Build GRCh38
rs750216116 Chr.X: 15331547-15331547 on Build GRCh38
rs866734779 Chr.X: 15331644-15331644 on Build GRCh38
rs140606572 Chr.X: 15331855-15331855 on Build GRCh38
rs754050083 Chr.X: 15331626-15331626 on Build GRCh38
rs774306554 Chr.X: 15331746-15331746 on Build GRCh38
rs587777397 Chr.X: 15331855-15331856 on Build GRCh38
rs587776726 Chr.X: 15331681-15331682 on Build GRCh38
rs746412962 Chr.X: 15331425-15331425 on Build GRCh38
rs750555194 Chr.X: 15331835-15331835 on Build GRCh38
rs375401655 Chr.X: 15331870-15331870 on Build GRCh38
rs587777396 Chr.X: 15331576-15331576 on Build GRCh38
rs747568374 Chr.X: 15331848-15331848 on Build GRCh38
rs758863767 Chr.X: 15331839-15331839 on Build GRCh38

View Full Product Details