Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 20006436-20006941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP7D2
Entrez Gene ID
256714
Gene Name
MAP7 domain containing 2
Gene Aliases
-
UniGene
Hs.127951
Gene Chromosome Location
Chr.X: 20006713-20116996 on Build GRCh38
Transcripts
NM_001168465.1 NM_001168466.1 NM_001168467.1 NM_152780.3 XM_005274478.2 XM_005274479.2 XM_005274480.3 XM_006724481.2 XM_006724482.2 XM_011545483.1 XM_011545484.1 XM_011545485.1 XM_017029393.1 XM_017029394.1 XM_017029395.1 XM_017029396.1 XM_017029397.1

SNP Detail

SNP ID Chromosome Location
rs374753096 Chr.X: 20006723-20006723 on Build GRCh38
rs778801073 Chr.X: 20006821-20006821 on Build GRCh38
rs897789000 Chr.X: 20006596-20006596 on Build GRCh38
rs995242287 Chr.X: 20006837-20006837 on Build GRCh38
rs963826032 Chr.X: 20006509-20006509 on Build GRCh38
rs1043931126 Chr.X: 20006494-20006494 on Build GRCh38
rs762796577 Chr.X: 20006664-20006664 on Build GRCh38
rs766423871 Chr.X: 20006674-20006674 on Build GRCh38
rs926489895 Chr.X: 20006777-20006777 on Build GRCh38
rs866491802 Chr.X: 20006603-20006603 on Build GRCh38
rs1048049285 Chr.X: 20006852-20006852 on Build GRCh38
rs1018222848 Chr.X: 20006502-20006502 on Build GRCh38
rs980624872 Chr.X: 20006679-20006679 on Build GRCh38
rs982463747 Chr.X: 20006643-20006643 on Build GRCh38
rs751455915 Chr.X: 20006757-20006757 on Build GRCh38
rs914004263 Chr.X: 20006883-20006883 on Build GRCh38
rs569265283 Chr.X: 20006611-20006611 on Build GRCh38
rs1056921817 Chr.X: 20006817-20006817 on Build GRCh38
rs998365537 Chr.X: 20006549-20006549 on Build GRCh38
rs1007691919 Chr.X: 20006497-20006497 on Build GRCh38
rs767051707 Chr.X: 20006908-20006908 on Build GRCh38
rs775638243 Chr.X: 20006495-20006495 on Build GRCh38
rs1036841907 Chr.X: 20006822-20006822 on Build GRCh38
rs951000302 Chr.X: 20006565-20006565 on Build GRCh38
rs949218698 Chr.X: 20006738-20006738 on Build GRCh38
rs1055419197 Chr.X: 20006480-20006480 on Build GRCh38
rs1006559482 Chr.X: 20006905-20006905 on Build GRCh38
rs896899331 Chr.X: 20006826-20006826 on Build GRCh38
rs371471294 Chr.X: 20006478-20006478 on Build GRCh38
rs397895244 Chr.X: 20006604-20006604 on Build GRCh38
rs993926410 Chr.X: 20006624-20006624 on Build GRCh38
rs1016959705 Chr.X: 20006587-20006587 on Build GRCh38
rs181588551 Chr.X: 20006778-20006778 on Build GRCh38
rs745691983 Chr.X: 20006832-20006832 on Build GRCh38
rs55887754 Chr.X: 20006789-20006789 on Build GRCh38
rs889415039 Chr.X: 20006877-20006877 on Build GRCh38
rs533004249 Chr.X: 20006603-20006604 on Build GRCh38
rs189018327 Chr.X: 20006661-20006661 on Build GRCh38
rs765171703 Chr.X: 20006649-20006649 on Build GRCh38
rs1029320934 Chr.X: 20006552-20006552 on Build GRCh38
rs56243063 Chr.X: 20006786-20006786 on Build GRCh38

View Full Product Details