Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 23001823-23002344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX53
Entrez Gene ID
168400
Gene Name
DEAD-box helicase 53
Gene Aliases
CAGE, CT26
UniGene
Hs.434416
Gene Chromosome Location
Chr.X: 22999961-23002089 on Build GRCh38
Transcripts
NM_182699.3 NR_073010.2

Gene Symbol
PTCHD1-AS
Entrez Gene ID
100873065
Gene Name
PTCHD1 antisense RNA (head to head)
Gene Aliases
DDX53-AS1, PTCHD1AS1, PTCHD1AS2
UniGene
Hs.628459
Gene Chromosome Location
Chr.X: 22193005-23293146 on Build GRCh38
Transcripts
NM_182699.3 NR_073010.2

SNP Detail

SNP ID Chromosome Location
rs867763402 Chr.X: 23002313-23002313 on Build GRCh38
rs759627854 Chr.X: 23001848-23001848 on Build GRCh38
rs78745205 Chr.X: 23002146-23002146 on Build GRCh38
rs916135989 Chr.X: 23002283-23002283 on Build GRCh38
rs200573414 Chr.X: 23001903-23001903 on Build GRCh38
rs750037498 Chr.X: 23001893-23001893 on Build GRCh38
rs77234118 Chr.X: 23002290-23002290 on Build GRCh38
rs952310795 Chr.X: 23002272-23002272 on Build GRCh38
rs894515536 Chr.X: 23001963-23001963 on Build GRCh38
rs1028653237 Chr.X: 23002187-23002187 on Build GRCh38
rs779768341 Chr.X: 23002176-23002176 on Build GRCh38
rs773011947 Chr.X: 23002028-23002028 on Build GRCh38
rs199977337 Chr.X: 23001920-23001920 on Build GRCh38
rs141481850 Chr.X: 23001904-23001904 on Build GRCh38
rs1056412179 Chr.X: 23001951-23001951 on Build GRCh38
rs80018002 Chr.X: 23002227-23002227 on Build GRCh38
rs750999649 Chr.X: 23001908-23001908 on Build GRCh38
rs777075751 Chr.X: 23001881-23001881 on Build GRCh38
rs764486395 Chr.X: 23001855-23001855 on Build GRCh38
rs776935454 Chr.X: 23001999-23001999 on Build GRCh38
rs929844146 Chr.X: 23001966-23001966 on Build GRCh38
rs762280181 Chr.X: 23002003-23002003 on Build GRCh38
rs76205219 Chr.X: 23002169-23002169 on Build GRCh38
rs776154426 Chr.X: 23002319-23002320 on Build GRCh38
rs772154862 Chr.X: 23001977-23001977 on Build GRCh38
rs764984044 Chr.X: 23001890-23001890 on Build GRCh38
rs746295041 Chr.X: 23002164-23002164 on Build GRCh38
rs150859492 Chr.X: 23001913-23001913 on Build GRCh38
rs1045879799 Chr.X: 23002093-23002093 on Build GRCh38
rs942488901 Chr.X: 23002123-23002124 on Build GRCh38
rs78647628 Chr.X: 23002158-23002158 on Build GRCh38
rs991790083 Chr.X: 23002276-23002276 on Build GRCh38
rs1007835076 Chr.X: 23002199-23002199 on Build GRCh38
rs202047830 Chr.X: 23001991-23001991 on Build GRCh38
rs746157266 Chr.X: 23001956-23001956 on Build GRCh38
rs148316221 Chr.X: 23001852-23001852 on Build GRCh38
rs779685021 Chr.X: 23001926-23001926 on Build GRCh38
rs370317544 Chr.X: 23001849-23001849 on Build GRCh38
rs79341555 Chr.X: 23002255-23002255 on Build GRCh38
rs373801952 Chr.X: 23001911-23001911 on Build GRCh38
rs961128879 Chr.X: 23002223-23002223 on Build GRCh38
rs768469251 Chr.X: 23002154-23002154 on Build GRCh38
rs747012036 Chr.X: 23001987-23001987 on Build GRCh38
rs76023255 Chr.X: 23002194-23002194 on Build GRCh38
rs762227181 Chr.X: 23001884-23001884 on Build GRCh38
rs187384365 Chr.X: 23002170-23002170 on Build GRCh38
rs78965413 Chr.X: 23002101-23002101 on Build GRCh38
rs892923672 Chr.X: 23002173-23002173 on Build GRCh38
rs982082014 Chr.X: 23001916-23001916 on Build GRCh38
rs777533584 Chr.X: 23001927-23001927 on Build GRCh38
rs950024745 Chr.X: 23002039-23002039 on Build GRCh38
rs909644031 Chr.X: 23002006-23002006 on Build GRCh38
rs973763624 Chr.X: 23002251-23002251 on Build GRCh38
rs768244190 Chr.X: 23002277-23002277 on Build GRCh38
rs1048176432 Chr.X: 23001970-23001970 on Build GRCh38
rs112634260 Chr.X: 23002189-23002189 on Build GRCh38
rs79801205 Chr.X: 23002161-23002161 on Build GRCh38
rs755852799 Chr.X: 23001862-23001870 on Build GRCh38
rs192125531 Chr.X: 23002205-23002205 on Build GRCh38

View Full Product Details