Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 24558975-24559518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCYT1B
Entrez Gene ID
9468
Gene Name
phosphate cytidylyltransferase 1, choline, beta
Gene Aliases
CCTB, CTB
UniGene
Hs.660708
Gene Chromosome Location
Chr.X: 24558087-24672862 on Build GRCh38
Transcripts
NM_001163264.1 NM_001163265.1 NM_004845.4 XM_017029977.1

SNP Detail

SNP ID Chromosome Location
rs767613191 Chr.X: 24559026-24559026 on Build GRCh38
rs386416761 Chr.X: 24559421-24559422 on Build GRCh38
rs866750308 Chr.X: 24559445-24559445 on Build GRCh38
rs891917581 Chr.X: 24559120-24559120 on Build GRCh38
rs752562223 Chr.X: 24559027-24559027 on Build GRCh38
rs61756165 Chr.X: 24559479-24559479 on Build GRCh38
rs977313732 Chr.X: 24559001-24559001 on Build GRCh38
rs982693728 Chr.X: 24559310-24559310 on Build GRCh38
rs762122606 Chr.X: 24559437-24559437 on Build GRCh38
rs868844359 Chr.X: 24559439-24559439 on Build GRCh38
rs879164772 Chr.X: 24559442-24559443 on Build GRCh38
rs766823659 Chr.X: 24559424-24559426 on Build GRCh38
rs905716898 Chr.X: 24559085-24559085 on Build GRCh38
rs55813228 Chr.X: 24559216-24559216 on Build GRCh38
rs199723851 Chr.X: 24559426-24559427 on Build GRCh38
rs3180097 Chr.X: 24559087-24559087 on Build GRCh38
rs386416762 Chr.X: 24559425-24559426 on Build GRCh38
rs61756166 Chr.X: 24559477-24559477 on Build GRCh38
rs753551064 Chr.X: 24559317-24559317 on Build GRCh38
rs56367400 Chr.X: 24559318-24559318 on Build GRCh38
rs202168053 Chr.X: 24559433-24559433 on Build GRCh38
rs976988120 Chr.X: 24559191-24559191 on Build GRCh38
rs765585748 Chr.X: 24559435-24559436 on Build GRCh38
rs766625539 Chr.X: 24558998-24558998 on Build GRCh38
rs929048023 Chr.X: 24559034-24559034 on Build GRCh38
rs200641368 Chr.X: 24559430-24559430 on Build GRCh38
rs919732086 Chr.X: 24559007-24559007 on Build GRCh38
rs745551927 Chr.X: 24559145-24559145 on Build GRCh38
rs965603232 Chr.X: 24559180-24559180 on Build GRCh38
rs1010728621 Chr.X: 24559142-24559142 on Build GRCh38
rs141424149 Chr.X: 24559000-24559000 on Build GRCh38
rs576039960 Chr.X: 24559456-24559457 on Build GRCh38
rs56166969 Chr.X: 24559465-24559465 on Build GRCh38
rs941231508 Chr.X: 24559101-24559103 on Build GRCh38
rs778662590 Chr.X: 24559107-24559107 on Build GRCh38
rs5901755 Chr.X: 24559428-24559429 on Build GRCh38
rs867547154 Chr.X: 24559416-24559416 on Build GRCh38
rs56116914 Chr.X: 24559467-24559467 on Build GRCh38
rs983564637 Chr.X: 24559405-24559405 on Build GRCh38
rs777341737 Chr.X: 24559440-24559441 on Build GRCh38
rs143567620 Chr.X: 24559146-24559146 on Build GRCh38
rs908898914 Chr.X: 24559409-24559409 on Build GRCh38
rs1006921547 Chr.X: 24559483-24559483 on Build GRCh38
rs921120001 Chr.X: 24559153-24559153 on Build GRCh38
rs756837726 Chr.X: 24559088-24559088 on Build GRCh38
rs759564017 Chr.X: 24559002-24559002 on Build GRCh38
rs748587714 Chr.X: 24559270-24559270 on Build GRCh38
rs756136435 Chr.X: 24559042-24559042 on Build GRCh38
rs148040646 Chr.X: 24559336-24559336 on Build GRCh38
rs912462501 Chr.X: 24559317-24559317 on Build GRCh38
rs1039725838 Chr.X: 24559497-24559497 on Build GRCh38
rs796738989 Chr.X: 24559332-24559332 on Build GRCh38
rs190552352 Chr.X: 24559214-24559214 on Build GRCh38
rs979723589 Chr.X: 24559420-24559420 on Build GRCh38
rs199704520 Chr.X: 24559429-24559429 on Build GRCh38
rs756755918 Chr.X: 24559420-24559421 on Build GRCh38
rs55732431 Chr.X: 24559457-24559457 on Build GRCh38
rs1038230835 Chr.X: 24559138-24559139 on Build GRCh38
rs5986619 Chr.X: 24559441-24559441 on Build GRCh38
rs2178608 Chr.X: 24559474-24559474 on Build GRCh38
rs756744505 Chr.X: 24559456-24559461 on Build GRCh38
rs540793084 Chr.X: 24559442-24559443 on Build GRCh38
rs570509560 Chr.X: 24559331-24559332 on Build GRCh38
rs55957731 Chr.X: 24559050-24559050 on Build GRCh38
rs200420724 Chr.X: 24559435-24559435 on Build GRCh38
rs371076538 Chr.X: 24559083-24559083 on Build GRCh38
rs200405997 Chr.X: 24559390-24559393 on Build GRCh38
rs1007162327 Chr.X: 24559152-24559152 on Build GRCh38
rs201092921 Chr.X: 24559424-24559425 on Build GRCh38
rs753707939 Chr.X: 24559082-24559082 on Build GRCh38
rs763794359 Chr.X: 24559079-24559079 on Build GRCh38
rs960692064 Chr.X: 24559055-24559055 on Build GRCh38
rs932485447 Chr.X: 24559167-24559167 on Build GRCh38
rs773435017 Chr.X: 24559418-24559419 on Build GRCh38
rs374459473 Chr.X: 24559230-24559230 on Build GRCh38
rs375662642 Chr.X: 24559441-24559442 on Build GRCh38
rs983174006 Chr.X: 24559036-24559036 on Build GRCh38
rs752356119 Chr.X: 24559242-24559242 on Build GRCh38
rs759861990 Chr.X: 24559434-24559435 on Build GRCh38
rs774753388 Chr.X: 24559317-24559318 on Build GRCh38
rs950506366 Chr.X: 24559393-24559393 on Build GRCh38
rs745671471 Chr.X: 24559168-24559169 on Build GRCh38
rs371418457 Chr.X: 24559418-24559418 on Build GRCh38
rs1018567581 Chr.X: 24559169-24559169 on Build GRCh38

View Full Product Details