Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 44148380-44148896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFHC2
Entrez Gene ID
80258
Gene Name
EF-hand domain containing 2
Gene Aliases
MRX74, dJ1158H2.1
UniGene
Hs.521953
Gene Chromosome Location
Chr.X: 44146254-44343677 on Build GRCh38
Transcripts
NM_025184.3 XM_006724562.2

SNP Detail

SNP ID Chromosome Location
rs768439834 Chr.X: 44148871-44148871 on Build GRCh38
rs370246792 Chr.X: 44148804-44148804 on Build GRCh38
rs780135262 Chr.X: 44148768-44148768 on Build GRCh38
rs1002815723 Chr.X: 44148723-44148723 on Build GRCh38
rs758866914 Chr.X: 44148816-44148816 on Build GRCh38
rs751334977 Chr.X: 44148614-44148614 on Build GRCh38
rs991573549 Chr.X: 44148509-44148509 on Build GRCh38
rs765759198 Chr.X: 44148746-44148746 on Build GRCh38
rs771672756 Chr.X: 44148852-44148852 on Build GRCh38
rs777476496 Chr.X: 44148805-44148805 on Build GRCh38
rs750610150 Chr.X: 44148665-44148665 on Build GRCh38
rs755898109 Chr.X: 44148790-44148790 on Build GRCh38
rs887532739 Chr.X: 44148560-44148560 on Build GRCh38
rs34729789 Chr.X: 44148853-44148853 on Build GRCh38
rs916979748 Chr.X: 44148512-44148512 on Build GRCh38
rs1050875891 Chr.X: 44148793-44148793 on Build GRCh38
rs781246906 Chr.X: 44148769-44148769 on Build GRCh38
rs905777586 Chr.X: 44148636-44148636 on Build GRCh38
rs370446144 Chr.X: 44148620-44148620 on Build GRCh38
rs996163434 Chr.X: 44148777-44148777 on Build GRCh38
rs778950139 Chr.X: 44148827-44148827 on Build GRCh38
rs749093591 Chr.X: 44148818-44148818 on Build GRCh38
rs949823505 Chr.X: 44148578-44148578 on Build GRCh38
rs867085679 Chr.X: 44148835-44148835 on Build GRCh38
rs73481049 Chr.X: 44148765-44148765 on Build GRCh38
rs1036098081 Chr.X: 44148740-44148740 on Build GRCh38
rs775319468 Chr.X: 44148492-44148492 on Build GRCh38
rs374637588 Chr.X: 44148831-44148831 on Build GRCh38
rs753368892 Chr.X: 44148766-44148766 on Build GRCh38
rs886936245 Chr.X: 44148616-44148616 on Build GRCh38
rs764578894 Chr.X: 44148819-44148819 on Build GRCh38
rs763841814 Chr.X: 44148611-44148611 on Build GRCh38
rs373258547 Chr.X: 44148753-44148753 on Build GRCh38
rs751978799 Chr.X: 44148850-44148850 on Build GRCh38
rs186129923 Chr.X: 44148615-44148615 on Build GRCh38
rs1038212280 Chr.X: 44148675-44148675 on Build GRCh38
rs762896687 Chr.X: 44148527-44148529 on Build GRCh38
rs745349172 Chr.X: 44148839-44148839 on Build GRCh38
rs752972883 Chr.X: 44148771-44148771 on Build GRCh38
rs755480992 Chr.X: 44148862-44148862 on Build GRCh38

View Full Product Details