Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49033634-49034141 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFE3
Entrez Gene ID
7030
Gene Name
transcription factor binding to IGHM enhancer 3
Gene Aliases
RCCP2, RCCX1, TFEA, bHLHe33
UniGene
Hs.730740
Gene Chromosome Location
Chr.X: 49028726-49043517 on Build GRCh38
Transcripts
NM_001282142.1 NM_006521.5

SNP Detail

SNP ID Chromosome Location
rs782164118 Chr.X: 49033965-49033965 on Build GRCh38
rs782816369 Chr.X: 49033990-49033990 on Build GRCh38
rs369475777 Chr.X: 49033711-49033711 on Build GRCh38
rs782436702 Chr.X: 49034092-49034092 on Build GRCh38
rs782521367 Chr.X: 49034079-49034079 on Build GRCh38
rs782358989 Chr.X: 49033793-49033793 on Build GRCh38
rs781929039 Chr.X: 49033796-49033796 on Build GRCh38
rs960063584 Chr.X: 49033837-49033837 on Build GRCh38
rs782555413 Chr.X: 49034113-49034113 on Build GRCh38
rs781835191 Chr.X: 49033830-49033830 on Build GRCh38
rs782115411 Chr.X: 49033778-49033778 on Build GRCh38
rs890548874 Chr.X: 49033655-49033655 on Build GRCh38
rs782524028 Chr.X: 49033968-49033968 on Build GRCh38
rs782076934 Chr.X: 49033712-49033712 on Build GRCh38
rs185587819 Chr.X: 49033705-49033705 on Build GRCh38
rs782034465 Chr.X: 49033768-49033768 on Build GRCh38
rs201168885 Chr.X: 49033699-49033699 on Build GRCh38
rs782004627 Chr.X: 49033777-49033777 on Build GRCh38
rs189222789 Chr.X: 49033825-49033825 on Build GRCh38
rs782369351 Chr.X: 49033766-49033766 on Build GRCh38
rs782656388 Chr.X: 49033709-49033709 on Build GRCh38
rs782379337 Chr.X: 49033720-49033720 on Build GRCh38
rs968351524 Chr.X: 49033951-49033951 on Build GRCh38
rs782538136 Chr.X: 49034095-49034096 on Build GRCh38
rs781846888 Chr.X: 49033824-49033824 on Build GRCh38
rs988063122 Chr.X: 49034026-49034026 on Build GRCh38
rs781863465 Chr.X: 49033693-49033693 on Build GRCh38
rs991221441 Chr.X: 49033863-49033863 on Build GRCh38
rs149741986 Chr.X: 49033923-49033923 on Build GRCh38
rs372314452 Chr.X: 49033694-49033694 on Build GRCh38
rs782793485 Chr.X: 49034034-49034034 on Build GRCh38
rs782635252 Chr.X: 49033688-49033688 on Build GRCh38
rs373906296 Chr.X: 49033997-49033997 on Build GRCh38
rs3027471 Chr.X: 49033821-49033821 on Build GRCh38
rs192304551 Chr.X: 49033964-49033964 on Build GRCh38
rs924850248 Chr.X: 49034013-49034013 on Build GRCh38
rs782613018 Chr.X: 49033698-49033698 on Build GRCh38
rs886969467 Chr.X: 49033786-49033786 on Build GRCh38
rs782593756 Chr.X: 49034082-49034082 on Build GRCh38
rs782716693 Chr.X: 49033826-49033826 on Build GRCh38
rs145602217 Chr.X: 49034018-49034018 on Build GRCh38
rs782584600 Chr.X: 49034093-49034093 on Build GRCh38
rs782075514 Chr.X: 49033816-49033816 on Build GRCh38
rs912024196 Chr.X: 49034031-49034031 on Build GRCh38
rs782228674 Chr.X: 49034114-49034114 on Build GRCh38
rs781888101 Chr.X: 49034097-49034097 on Build GRCh38

View Full Product Details