Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 107067052-107067562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM41
Entrez Gene ID
55285
Gene Name
RNA binding motif protein 41
Gene Aliases
-
UniGene
Hs.139053
Gene Chromosome Location
Chr.X: 107061885-107118827 on Build GRCh38
Transcripts
NM_001171080.1 NM_001324242.1 NM_001324243.1 NM_001324244.1 NM_018301.4 NR_136728.1 NR_136729.1 NR_136730.1 NR_136731.1 NR_136732.1 XM_011530980.2 XM_011530981.2

SNP Detail

SNP ID Chromosome Location
rs932733730 Chr.X: 107067415-107067415 on Build GRCh38
rs894033420 Chr.X: 107067086-107067086 on Build GRCh38
rs961365621 Chr.X: 107067352-107067352 on Build GRCh38
rs749711213 Chr.X: 107067495-107067495 on Build GRCh38
rs996259413 Chr.X: 107067293-107067293 on Build GRCh38
rs773559637 Chr.X: 107067519-107067519 on Build GRCh38
rs763221762 Chr.X: 107067517-107067518 on Build GRCh38
rs746251164 Chr.X: 107067472-107067472 on Build GRCh38
rs113278330 Chr.X: 107067382-107067382 on Build GRCh38
rs142775579 Chr.X: 107067240-107067240 on Build GRCh38
rs757821981 Chr.X: 107067145-107067145 on Build GRCh38
rs780411576 Chr.X: 107067483-107067483 on Build GRCh38
rs942301688 Chr.X: 107067235-107067235 on Build GRCh38
rs779204935 Chr.X: 107067470-107067470 on Build GRCh38
rs1016005047 Chr.X: 107067327-107067327 on Build GRCh38
rs146046356 Chr.X: 107067513-107067513 on Build GRCh38
rs922656225 Chr.X: 107067414-107067414 on Build GRCh38
rs952613821 Chr.X: 107067247-107067247 on Build GRCh38
rs768372674 Chr.X: 107067297-107067297 on Build GRCh38
rs747235400 Chr.X: 107067535-107067535 on Build GRCh38
rs998934837 Chr.X: 107067174-107067174 on Build GRCh38
rs985612751 Chr.X: 107067443-107067443 on Build GRCh38
rs900696802 Chr.X: 107067267-107067267 on Build GRCh38
rs902887308 Chr.X: 107067166-107067166 on Build GRCh38
rs772335736 Chr.X: 107067515-107067515 on Build GRCh38
rs1007975151 Chr.X: 107067394-107067394 on Build GRCh38
rs977158205 Chr.X: 107067375-107067375 on Build GRCh38
rs185856723 Chr.X: 107067081-107067081 on Build GRCh38
rs915434596 Chr.X: 107067459-107067459 on Build GRCh38
rs1048227950 Chr.X: 107067105-107067105 on Build GRCh38
rs781731303 Chr.X: 107067175-107067175 on Build GRCh38
rs1031776971 Chr.X: 107067489-107067489 on Build GRCh38
rs764494150 Chr.X: 107067111-107067111 on Build GRCh38
rs748235606 Chr.X: 107067514-107067514 on Build GRCh38
rs757705767 Chr.X: 107067437-107067437 on Build GRCh38
rs895589294 Chr.X: 107067215-107067215 on Build GRCh38
rs1012675509 Chr.X: 107067222-107067222 on Build GRCh38
rs371858336 Chr.X: 107067504-107067504 on Build GRCh38

View Full Product Details