Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 108156306-108156810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A6
Entrez Gene ID
1288
Gene Name
collagen type IV alpha 6 chain
Gene Aliases
CXDELq22.3, DELXq22.3, DFNX6
UniGene
Hs.145586
Gene Chromosome Location
Chr.X: 108155607-108439497 on Build GRCh38
Transcripts
NM_001287758.1 NM_001287759.1 NM_001287760.1 NM_001847.3 NM_033641.3 XM_006724617.3 XM_011530852.2 XM_011530853.2

SNP Detail

SNP ID Chromosome Location
rs888617352 Chr.X: 108156722-108156722 on Build GRCh38
rs1006164773 Chr.X: 108156466-108156466 on Build GRCh38
rs189587894 Chr.X: 108156535-108156535 on Build GRCh38
rs1031371355 Chr.X: 108156687-108156687 on Build GRCh38
rs1015864511 Chr.X: 108156743-108156743 on Build GRCh38
rs888617903 Chr.X: 108156465-108156465 on Build GRCh38
rs1039747716 Chr.X: 108156506-108156506 on Build GRCh38
rs930253035 Chr.X: 108156334-108156334 on Build GRCh38
rs999447301 Chr.X: 108156667-108156667 on Build GRCh38
rs748749238 Chr.X: 108156530-108156530 on Build GRCh38
rs745381116 Chr.X: 108156729-108156729 on Build GRCh38
rs899950925 Chr.X: 108156545-108156545 on Build GRCh38
rs954751371 Chr.X: 108156649-108156649 on Build GRCh38
rs979536987 Chr.X: 108156368-108156368 on Build GRCh38
rs576629835 Chr.X: 108156595-108156595 on Build GRCh38
rs998117623 Chr.X: 108156584-108156584 on Build GRCh38
rs1029629890 Chr.X: 108156640-108156640 on Build GRCh38
rs957884121 Chr.X: 108156689-108156689 on Build GRCh38
rs1057225607 Chr.X: 108156337-108156337 on Build GRCh38
rs181344454 Chr.X: 108156677-108156677 on Build GRCh38
rs948509165 Chr.X: 108156442-108156442 on Build GRCh38
rs1005782508 Chr.X: 108156727-108156727 on Build GRCh38
rs780831255 Chr.X: 108156346-108156346 on Build GRCh38
rs916311572 Chr.X: 108156739-108156739 on Build GRCh38
rs969142528 Chr.X: 108156771-108156771 on Build GRCh38
rs112048474 Chr.X: 108156528-108156528 on Build GRCh38
rs1044535761 Chr.X: 108156448-108156448 on Build GRCh38

View Full Product Details