Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 183250824-183251329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NMNAT2
Entrez Gene ID
23057
Gene Name
nicotinamide nucleotide adenylyltransferase 2
Gene Aliases
C1orf15, PNAT2
UniGene
Hs.497123
Gene Chromosome Location
Chr.1: 183248237-183418499 on Build GRCh38
Transcripts
NM_015039.3 NM_170706.3

SNP Detail

SNP ID Chromosome Location
rs201645009 Chr.1: 183251169-183251169 on Build GRCh38
rs1001554483 Chr.1: 183251165-183251165 on Build GRCh38
rs547298370 Chr.1: 183251016-183251016 on Build GRCh38
rs567308559 Chr.1: 183250880-183250880 on Build GRCh38
rs886660178 Chr.1: 183251069-183251069 on Build GRCh38
rs200886873 Chr.1: 183250993-183250993 on Build GRCh38
rs188772255 Chr.1: 183251223-183251223 on Build GRCh38
rs376757381 Chr.1: 183250909-183250909 on Build GRCh38
rs893222202 Chr.1: 183251186-183251186 on Build GRCh38
rs753179227 Chr.1: 183250867-183250867 on Build GRCh38
rs1032201275 Chr.1: 183251250-183251250 on Build GRCh38
rs375065076 Chr.1: 183250850-183250850 on Build GRCh38
rs201583277 Chr.1: 183251048-183251048 on Build GRCh38
rs897841635 Chr.1: 183251106-183251106 on Build GRCh38
rs202098419 Chr.1: 183250980-183250980 on Build GRCh38
rs913598581 Chr.1: 183250891-183250891 on Build GRCh38
rs200073270 Chr.1: 183250928-183250928 on Build GRCh38
rs200476043 Chr.1: 183250921-183250921 on Build GRCh38
rs943089186 Chr.1: 183250959-183250959 on Build GRCh38
rs200202879 Chr.1: 183251062-183251062 on Build GRCh38
rs200274043 Chr.1: 183251300-183251300 on Build GRCh38
rs202215431 Chr.1: 183251295-183251295 on Build GRCh38
rs200820741 Chr.1: 183251184-183251184 on Build GRCh38
rs201648687 Chr.1: 183250948-183250948 on Build GRCh38
rs199832981 Chr.1: 183251170-183251170 on Build GRCh38
rs1033032812 Chr.1: 183251174-183251174 on Build GRCh38
rs199644713 Chr.1: 183250901-183250901 on Build GRCh38
rs1014462556 Chr.1: 183251073-183251073 on Build GRCh38
rs201900425 Chr.1: 183251247-183251247 on Build GRCh38
rs780849816 Chr.1: 183251244-183251244 on Build GRCh38
rs1009501065 Chr.1: 183251278-183251278 on Build GRCh38
rs974426358 Chr.1: 183251303-183251303 on Build GRCh38
rs911598398 Chr.1: 183250953-183250953 on Build GRCh38
rs957980745 Chr.1: 183250852-183250852 on Build GRCh38
rs201002390 Chr.1: 183251257-183251257 on Build GRCh38
rs200495940 Chr.1: 183251020-183251020 on Build GRCh38
rs201252307 Chr.1: 183251063-183251063 on Build GRCh38
rs200268149 Chr.1: 183250981-183250981 on Build GRCh38
rs1055921480 Chr.1: 183251046-183251046 on Build GRCh38
rs199844681 Chr.1: 183250950-183250950 on Build GRCh38
rs199756683 Chr.1: 183251110-183251110 on Build GRCh38
rs867057724 Chr.1: 183251145-183251145 on Build GRCh38
rs202240373 Chr.1: 183250995-183250995 on Build GRCh38
rs201225774 Chr.1: 183251285-183251285 on Build GRCh38
rs905907631 Chr.1: 183251129-183251129 on Build GRCh38
rs201514500 Chr.1: 183250898-183250898 on Build GRCh38
rs200575240 Chr.1: 183251114-183251114 on Build GRCh38
rs200061144 Chr.1: 183251254-183251254 on Build GRCh38
rs989737791 Chr.1: 183250926-183250926 on Build GRCh38
rs201988685 Chr.1: 183251201-183251201 on Build GRCh38
rs142219834 Chr.1: 183250951-183250951 on Build GRCh38
rs200242551 Chr.1: 183251284-183251284 on Build GRCh38
rs1045706243 Chr.1: 183251081-183251081 on Build GRCh38
rs199592014 Chr.1: 183251050-183251050 on Build GRCh38
rs758510767 Chr.1: 183250868-183250868 on Build GRCh38
rs201204799 Chr.1: 183250944-183250944 on Build GRCh38
rs201600321 Chr.1: 183250922-183250922 on Build GRCh38
rs201994443 Chr.1: 183251265-183251265 on Build GRCh38
rs146734847 Chr.1: 183250982-183250982 on Build GRCh38
rs199939062 Chr.1: 183251207-183251207 on Build GRCh38

View Full Product Details