Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 129908036-129908547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UTP14A
Entrez Gene ID
10813
Gene Name
UTP14A small subunit processome component
Gene Aliases
NYCO16, SDCCAG16, dJ537K23.3
UniGene
Hs.458598
Gene Chromosome Location
Chr.X: 129906121-129929762 on Build GRCh38
Transcripts
NM_001166221.1 NM_006649.3 XM_017029237.1 XM_017029238.1

SNP Detail

SNP ID Chromosome Location
rs904794568 Chr.X: 129908417-129908417 on Build GRCh38
rs751264780 Chr.X: 129908346-129908346 on Build GRCh38
rs938280342 Chr.X: 129908308-129908308 on Build GRCh38
rs748141404 Chr.X: 129908126-129908126 on Build GRCh38
rs143067310 Chr.X: 129908172-129908172 on Build GRCh38
rs1057462208 Chr.X: 129908303-129908303 on Build GRCh38
rs757565993 Chr.X: 129908069-129908069 on Build GRCh38
rs1020571446 Chr.X: 129908400-129908400 on Build GRCh38
rs1046638552 Chr.X: 129908204-129908204 on Build GRCh38
rs972187330 Chr.X: 129908109-129908109 on Build GRCh38
rs746012078 Chr.X: 129908080-129908080 on Build GRCh38
rs766223559 Chr.X: 129908331-129908331 on Build GRCh38
rs774205938 Chr.X: 129908091-129908091 on Build GRCh38
rs779153657 Chr.X: 129908154-129908160 on Build GRCh38
rs1054423419 Chr.X: 129908321-129908321 on Build GRCh38
rs1010807313 Chr.X: 129908356-129908356 on Build GRCh38
rs756504889 Chr.X: 129908415-129908415 on Build GRCh38
rs773178664 Chr.X: 129908093-129908093 on Build GRCh38
rs750790804 Chr.X: 129908385-129908385 on Build GRCh38
rs771727520 Chr.X: 129908128-129908128 on Build GRCh38
rs781407385 Chr.X: 129908078-129908078 on Build GRCh38
rs201734952 Chr.X: 129908079-129908079 on Build GRCh38
rs747132410 Chr.X: 129908063-129908063 on Build GRCh38
rs148845598 Chr.X: 129908090-129908090 on Build GRCh38
rs370624137 Chr.X: 129908136-129908136 on Build GRCh38
rs780259509 Chr.X: 129908480-129908480 on Build GRCh38
rs905328191 Chr.X: 129908260-129908260 on Build GRCh38
rs773067456 Chr.X: 129908135-129908135 on Build GRCh38
rs776084700 Chr.X: 129908142-129908142 on Build GRCh38
rs754623183 Chr.X: 129908443-129908443 on Build GRCh38
rs764745939 Chr.X: 129908173-129908173 on Build GRCh38
rs57312277 Chr.X: 129908343-129908343 on Build GRCh38
rs754122316 Chr.X: 129908499-129908499 on Build GRCh38
rs1019890817 Chr.X: 129908410-129908410 on Build GRCh38

View Full Product Details