Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 135287112-135287618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF75D
Entrez Gene ID
7626
Gene Name
zinc finger protein 75D
Gene Aliases
D8C6, ZKSCAN24, ZNF75, ZNF82, ZSCAN28
UniGene
Hs.371717
Gene Chromosome Location
Chr.X: 135248589-135344109 on Build GRCh38
Transcripts
NM_001185063.2 NM_007131.4 NR_110381.1 XM_005262469.1 XM_005262471.3 XM_011531391.1 XM_011531395.2 XM_011531396.2 XM_011531397.2 XM_017029818.1 XM_017029819.1 XM_017029820.1

SNP Detail

SNP ID Chromosome Location
rs782373490 Chr.X: 135287339-135287339 on Build GRCh38
rs781996329 Chr.X: 135287343-135287343 on Build GRCh38
rs782065716 Chr.X: 135287205-135287205 on Build GRCh38
rs200077305 Chr.X: 135287348-135287348 on Build GRCh38
rs782487677 Chr.X: 135287281-135287281 on Build GRCh38
rs374016610 Chr.X: 135287362-135287362 on Build GRCh38
rs781912304 Chr.X: 135287204-135287204 on Build GRCh38
rs782539023 Chr.X: 135287570-135287570 on Build GRCh38
rs782638541 Chr.X: 135287196-135287196 on Build GRCh38
rs782616981 Chr.X: 135287579-135287579 on Build GRCh38
rs370506070 Chr.X: 135287511-135287511 on Build GRCh38
rs782222579 Chr.X: 135287276-135287276 on Build GRCh38
rs138085112 Chr.X: 135287201-135287201 on Build GRCh38
rs782291799 Chr.X: 135287144-135287144 on Build GRCh38
rs782660699 Chr.X: 135287142-135287142 on Build GRCh38
rs782315760 Chr.X: 135287327-135287327 on Build GRCh38
rs781963375 Chr.X: 135287216-135287216 on Build GRCh38
rs781846273 Chr.X: 135287403-135287403 on Build GRCh38
rs781859573 Chr.X: 135287295-135287295 on Build GRCh38
rs782117518 Chr.X: 135287411-135287411 on Build GRCh38
rs782365684 Chr.X: 135287193-135287193 on Build GRCh38
rs782622244 Chr.X: 135287558-135287558 on Build GRCh38
rs782269479 Chr.X: 135287200-135287200 on Build GRCh38
rs1129093 Chr.X: 135287236-135287236 on Build GRCh38
rs782223348 Chr.X: 135287188-135287188 on Build GRCh38
rs782729498 Chr.X: 135287401-135287401 on Build GRCh38
rs146364929 Chr.X: 135287378-135287378 on Build GRCh38
rs782533016 Chr.X: 135287296-135287296 on Build GRCh38
rs782495381 Chr.X: 135287522-135287522 on Build GRCh38
rs782254803 Chr.X: 135287569-135287569 on Build GRCh38
rs782707041 Chr.X: 135287271-135287271 on Build GRCh38
rs782652504 Chr.X: 135287399-135287399 on Build GRCh38
rs140155206 Chr.X: 135287338-135287338 on Build GRCh38
rs782432955 Chr.X: 135287152-135287152 on Build GRCh38
rs782609409 Chr.X: 135287330-135287330 on Build GRCh38
rs782169549 Chr.X: 135287321-135287321 on Build GRCh38
rs377434762 Chr.X: 135287300-135287300 on Build GRCh38
rs367866888 Chr.X: 135287479-135287479 on Build GRCh38
rs782704185 Chr.X: 135287166-135287166 on Build GRCh38
rs782423250 Chr.X: 135287376-135287376 on Build GRCh38
rs782162507 Chr.X: 135287269-135287269 on Build GRCh38
rs371378374 Chr.X: 135287172-135287172 on Build GRCh38
rs200181900 Chr.X: 135287433-135287433 on Build GRCh38
rs782783133 Chr.X: 135287421-135287421 on Build GRCh38
rs201770071 Chr.X: 135287237-135287237 on Build GRCh38
rs782622883 Chr.X: 135287287-135287287 on Build GRCh38
rs782114578 Chr.X: 135287227-135287227 on Build GRCh38

View Full Product Details