Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 135967276-135967800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMGT1
Entrez Gene ID
93380
Gene Name
membrane magnesium transporter 1
Gene Aliases
EMC5, TMEM32
UniGene
Hs.110702
Gene Chromosome Location
Chr.X: 135962072-135973975 on Build GRCh38
Transcripts
NM_173470.1

SNP Detail

SNP ID Chromosome Location
rs781853438 Chr.X: 135967419-135967419 on Build GRCh38
rs201063329 Chr.X: 135967428-135967428 on Build GRCh38
rs367822678 Chr.X: 135967452-135967452 on Build GRCh38
rs782035841 Chr.X: 135967592-135967592 on Build GRCh38
rs782729865 Chr.X: 135967442-135967442 on Build GRCh38
rs782464547 Chr.X: 135967342-135967342 on Build GRCh38
rs782612238 Chr.X: 135967364-135967364 on Build GRCh38
rs182059243 Chr.X: 135967334-135967334 on Build GRCh38
rs782519902 Chr.X: 135967525-135967525 on Build GRCh38
rs990339816 Chr.X: 135967537-135967537 on Build GRCh38
rs1004069630 Chr.X: 135967748-135967748 on Build GRCh38
rs782791839 Chr.X: 135967418-135967418 on Build GRCh38
rs782063283 Chr.X: 135967381-135967381 on Build GRCh38
rs981720229 Chr.X: 135967305-135967305 on Build GRCh38
rs1039825290 Chr.X: 135967567-135967567 on Build GRCh38
rs375909449 Chr.X: 135967407-135967407 on Build GRCh38
rs373366045 Chr.X: 135967365-135967365 on Build GRCh38
rs374214640 Chr.X: 135967571-135967571 on Build GRCh38
rs781943312 Chr.X: 135967358-135967358 on Build GRCh38
rs782031034 Chr.X: 135967383-135967383 on Build GRCh38
rs782302502 Chr.X: 135967382-135967382 on Build GRCh38
rs199809391 Chr.X: 135967453-135967453 on Build GRCh38
rs887057145 Chr.X: 135967734-135967734 on Build GRCh38
rs1051036803 Chr.X: 135967646-135967646 on Build GRCh38
rs368903410 Chr.X: 135967523-135967523 on Build GRCh38
rs782353547 Chr.X: 135967349-135967349 on Build GRCh38
rs946480947 Chr.X: 135967562-135967562 on Build GRCh38

View Full Product Details