Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 186308163-186308672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRG4
Entrez Gene ID
10216
Gene Name
proteoglycan 4
Gene Aliases
CACP, HAPO, JCAP, MSF, SZP
UniGene
Hs.647723
Gene Chromosome Location
Chr.1: 186296273-186314562 on Build GRCh38
Transcripts
NM_001127708.2 NM_001127709.2 NM_001127710.2 NM_001303232.1 NM_005807.4 XM_017000002.1 XM_017000003.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1728188 c.2875C>G p.Q959E Chr.1: 186308594-186308594 on Build GRCh38
7095339 c.2818G>C p.E940Q Chr.1: 186308537-186308537 on Build GRCh38
4913869 c.2532A>C p.P844P Chr.1: 186308251-186308251 on Build GRCh38
901100 c.2769C>A p.D923E Chr.1: 186308488-186308488 on Build GRCh38
4913944 c.2529T>A p.A843A Chr.1: 186308248-186308248 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760087679 Chr.1: 186308263-186308263 on Build GRCh38
rs763025365 Chr.1: 186308524-186308528 on Build GRCh38
rs879196581 Chr.1: 186308230-186308230 on Build GRCh38
rs747996260 Chr.1: 186308297-186308297 on Build GRCh38
rs371574559 Chr.1: 186308561-186308572 on Build GRCh38
rs868013528 Chr.1: 186308325-186308325 on Build GRCh38
rs1021634201 Chr.1: 186308587-186308587 on Build GRCh38
rs978288509 Chr.1: 186308276-186308276 on Build GRCh38
rs765323866 Chr.1: 186308191-186308191 on Build GRCh38
rs199605402 Chr.1: 186308558-186308558 on Build GRCh38
rs781580079 Chr.1: 186308470-186308470 on Build GRCh38
rs370601793 Chr.1: 186308492-186308492 on Build GRCh38
rs770347514 Chr.1: 186308480-186308480 on Build GRCh38
rs78918585 Chr.1: 186308532-186308532 on Build GRCh38
rs751196179 Chr.1: 186308535-186308535 on Build GRCh38
rs914554586 Chr.1: 186308397-186308397 on Build GRCh38
rs754513520 Chr.1: 186308223-186308223 on Build GRCh38
rs575773149 Chr.1: 186308606-186308608 on Build GRCh38
rs774423180 Chr.1: 186308511-186308511 on Build GRCh38
rs757878084 Chr.1: 186308514-186308514 on Build GRCh38
rs201850803 Chr.1: 186308602-186308602 on Build GRCh38
rs75928540 Chr.1: 186308395-186308395 on Build GRCh38
rs181303774 Chr.1: 186308551-186308551 on Build GRCh38
rs528241867 Chr.1: 186308581-186308581 on Build GRCh38
rs752608010 Chr.1: 186308508-186308508 on Build GRCh38
rs1047894704 Chr.1: 186308475-186308475 on Build GRCh38
rs34820948 Chr.1: 186308310-186308311 on Build GRCh38
rs765156915 Chr.1: 186308503-186308503 on Build GRCh38
rs954667660 Chr.1: 186308267-186308267 on Build GRCh38
rs749805360 Chr.1: 186308566-186308566 on Build GRCh38
rs763102061 Chr.1: 186308625-186308625 on Build GRCh38
rs1030694587 Chr.1: 186308252-186308252 on Build GRCh38
rs146935964 Chr.1: 186308235-186308235 on Build GRCh38
rs746835879 Chr.1: 186308231-186308232 on Build GRCh38
rs996220820 Chr.1: 186308521-186308521 on Build GRCh38
rs778605098 Chr.1: 186308226-186308226 on Build GRCh38
rs771814114 Chr.1: 186308229-186308229 on Build GRCh38
rs764826528 Chr.1: 186308418-186308418 on Build GRCh38
rs749344668 Chr.1: 186308484-186308484 on Build GRCh38
rs757596556 Chr.1: 186308452-186308452 on Build GRCh38
rs765420146 Chr.1: 186308466-186308470 on Build GRCh38
rs376770678 Chr.1: 186308448-186308448 on Build GRCh38
rs763075569 Chr.1: 186308321-186308321 on Build GRCh38
rs776625604 Chr.1: 186308499-186308499 on Build GRCh38
rs776884304 Chr.1: 186308429-186308429 on Build GRCh38
rs768388140 Chr.1: 186308424-186308425 on Build GRCh38
rs200451079 Chr.1: 186308549-186308549 on Build GRCh38
rs1053416542 Chr.1: 186308245-186308245 on Build GRCh38
rs955874664 Chr.1: 186308629-186308629 on Build GRCh38
rs75511330 Chr.1: 186308577-186308577 on Build GRCh38
rs552631244 Chr.1: 186308399-186308399 on Build GRCh38
rs777305605 Chr.1: 186308232-186308232 on Build GRCh38
rs769836301 Chr.1: 186308277-186308277 on Build GRCh38
rs777727804 Chr.1: 186308407-186308407 on Build GRCh38
rs368941961 Chr.1: 186308211-186308211 on Build GRCh38
rs767214166 Chr.1: 186308208-186308208 on Build GRCh38
rs763562501 Chr.1: 186308529-186308529 on Build GRCh38
rs761905657 Chr.1: 186308453-186308455 on Build GRCh38
rs768636935 Chr.1: 186308488-186308488 on Build GRCh38
rs575885397 Chr.1: 186308609-186308609 on Build GRCh38
rs780556356 Chr.1: 186308565-186308565 on Build GRCh38
rs753847010 Chr.1: 186308221-186308221 on Build GRCh38
rs78889600 Chr.1: 186308436-186308436 on Build GRCh38
rs970489739 Chr.1: 186308421-186308421 on Build GRCh38
rs200699648 Chr.1: 186308539-186308539 on Build GRCh38
rs373552428 Chr.1: 186308383-186308383 on Build GRCh38
rs369498738 Chr.1: 186308192-186308192 on Build GRCh38
rs978422870 Chr.1: 186308220-186308220 on Build GRCh38
rs772362320 Chr.1: 186308623-186308623 on Build GRCh38
rs957887709 Chr.1: 186308568-186308568 on Build GRCh38
rs774464069 Chr.1: 186308352-186308352 on Build GRCh38
rs142388523 Chr.1: 186308493-186308493 on Build GRCh38
rs759417504 Chr.1: 186308500-186308500 on Build GRCh38
rs773571923 Chr.1: 186308642-186308642 on Build GRCh38
rs886153188 Chr.1: 186308210-186308210 on Build GRCh38
rs146558489 Chr.1: 186308478-186308478 on Build GRCh38
rs780095231 Chr.1: 186308214-186308214 on Build GRCh38
rs775879212 Chr.1: 186308481-186308481 on Build GRCh38
rs1030787159 Chr.1: 186308567-186308567 on Build GRCh38
rs980473180 Chr.1: 186308603-186308603 on Build GRCh38
rs202211535 Chr.1: 186308222-186308222 on Build GRCh38
rs773183338 Chr.1: 186308513-186308514 on Build GRCh38
rs539584407 Chr.1: 186308586-186308586 on Build GRCh38
rs995965006 Chr.1: 186308185-186308185 on Build GRCh38
rs775709538 Chr.1: 186308312-186308312 on Build GRCh38
rs1030800112 Chr.1: 186308308-186308308 on Build GRCh38
rs541053554 Chr.1: 186308630-186308630 on Build GRCh38
rs752193399 Chr.1: 186308428-186308428 on Build GRCh38
rs760495240 Chr.1: 186308372-186308372 on Build GRCh38
rs777076056 Chr.1: 186308249-186308249 on Build GRCh38
rs1015929913 Chr.1: 186308218-186308218 on Build GRCh38
rs202232906 Chr.1: 186308444-186308444 on Build GRCh38
rs190199774 Chr.1: 186308469-186308469 on Build GRCh38
rs764307639 Chr.1: 186308323-186308323 on Build GRCh38
rs752065771 Chr.1: 186308607-186308608 on Build GRCh38
rs763280675 Chr.1: 186308196-186308196 on Build GRCh38
rs148222256 Chr.1: 186308228-186308228 on Build GRCh38

View Full Product Details