Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 200409841-200410346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF281
Entrez Gene ID
23528
Gene Name
zinc finger protein 281
Gene Aliases
ZBP-99, ZNP-99
UniGene
Hs.59757
Gene Chromosome Location
Chr.1: 200404947-200483604 on Build GRCh38
Transcripts
NM_001281293.1 NM_001281294.1 NM_012482.4 XM_017000887.1 XM_017000888.1

SNP Detail

SNP ID Chromosome Location
rs918337251 Chr.1: 200409982-200409982 on Build GRCh38
rs1048069622 Chr.1: 200409998-200409998 on Build GRCh38
rs562340128 Chr.1: 200410013-200410013 on Build GRCh38
rs879442675 Chr.1: 200410235-200410235 on Build GRCh38
rs933830453 Chr.1: 200410146-200410146 on Build GRCh38
rs772997705 Chr.1: 200410003-200410003 on Build GRCh38
rs1044951319 Chr.1: 200410187-200410187 on Build GRCh38
rs944911060 Chr.1: 200409958-200409958 on Build GRCh38
rs970359325 Chr.1: 200410084-200410084 on Build GRCh38
rs1048418389 Chr.1: 200409933-200409933 on Build GRCh38
rs947744608 Chr.1: 200410182-200410182 on Build GRCh38
rs1031071273 Chr.1: 200410029-200410029 on Build GRCh38
rs1034786739 Chr.1: 200410251-200410251 on Build GRCh38
rs992988977 Chr.1: 200410049-200410049 on Build GRCh38
rs543493598 Chr.1: 200409944-200409944 on Build GRCh38
rs902189076 Chr.1: 200410237-200410237 on Build GRCh38
rs565075620 Chr.1: 200409997-200409997 on Build GRCh38
rs922663838 Chr.1: 200410142-200410142 on Build GRCh38
rs972222947 Chr.1: 200410122-200410122 on Build GRCh38
rs989501976 Chr.1: 200409885-200409885 on Build GRCh38
rs889089333 Chr.1: 200410255-200410255 on Build GRCh38
rs548526206 Chr.1: 200410233-200410233 on Build GRCh38
rs144598379 Chr.1: 200410202-200410202 on Build GRCh38
rs1034733791 Chr.1: 200410039-200410039 on Build GRCh38
rs915452183 Chr.1: 200409892-200409892 on Build GRCh38
rs547004508 Chr.1: 200410241-200410241 on Build GRCh38
rs1040734996 Chr.1: 200409963-200409963 on Build GRCh38
rs540472021 Chr.1: 200409950-200409951 on Build GRCh38
rs912137748 Chr.1: 200410091-200410091 on Build GRCh38
rs953543547 Chr.1: 200410036-200410036 on Build GRCh38
rs956792094 Chr.1: 200410030-200410030 on Build GRCh38
rs1003381543 Chr.1: 200410018-200410018 on Build GRCh38
rs939989006 Chr.1: 200410130-200410130 on Build GRCh38
rs960198335 Chr.1: 200410043-200410043 on Build GRCh38
rs896210476 Chr.1: 200410324-200410324 on Build GRCh38
rs889679517 Chr.1: 200410006-200410006 on Build GRCh38
rs1011024113 Chr.1: 200410032-200410032 on Build GRCh38
rs1052256601 Chr.1: 200410231-200410231 on Build GRCh38
rs986392810 Chr.1: 200410070-200410070 on Build GRCh38
rs1022836324 Chr.1: 200410252-200410252 on Build GRCh38
rs10127913 Chr.1: 200410152-200410152 on Build GRCh38
rs912061973 Chr.1: 200409946-200409946 on Build GRCh38
rs1007974472 Chr.1: 200410267-200410267 on Build GRCh38
rs909545984 Chr.1: 200410178-200410178 on Build GRCh38
rs780208009 Chr.1: 200410286-200410286 on Build GRCh38
rs888137625 Chr.1: 200410002-200410002 on Build GRCh38
rs1045424991 Chr.1: 200410007-200410007 on Build GRCh38
rs909920216 Chr.1: 200410106-200410106 on Build GRCh38
rs559495128 Chr.1: 200410165-200410165 on Build GRCh38
rs1019168511 Chr.1: 200410279-200410279 on Build GRCh38
rs532398537 Chr.1: 200410035-200410035 on Build GRCh38
rs1003351815 Chr.1: 200410210-200410210 on Build GRCh38
rs371769987 Chr.1: 200410164-200410164 on Build GRCh38
rs367837317 Chr.1: 200410078-200410078 on Build GRCh38
rs918306238 Chr.1: 200410133-200410133 on Build GRCh38
rs1021986049 Chr.1: 200410023-200410023 on Build GRCh38
rs986812007 Chr.1: 200409938-200409938 on Build GRCh38
rs916163610 Chr.1: 200410082-200410082 on Build GRCh38
rs189973930 Chr.1: 200409934-200409934 on Build GRCh38
rs760582948 Chr.1: 200410168-200410168 on Build GRCh38
rs906334216 Chr.1: 200410012-200410012 on Build GRCh38
rs879634310 Chr.1: 200409922-200409922 on Build GRCh38
rs966142988 Chr.1: 200410107-200410107 on Build GRCh38
rs1037078688 Chr.1: 200410137-200410137 on Build GRCh38

View Full Product Details