Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 201194072-201194568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGFN1
Entrez Gene ID
91156
Gene Name
immunoglobulin-like and fibronectin type III domain containing 1
Gene Aliases
EEF1A2BP1
UniGene
Hs.519024
Gene Chromosome Location
Chr.1: 201190780-201228952 on Build GRCh38
Transcripts
NM_001164586.1 XM_005245580.3 XM_006711618.3 XM_011510120.2 XM_011510121.2 XM_017002787.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
902104 c.79A>C p.S27R Chr.1: 201194225-201194225 on Build GRCh38
7371893 c.16C>T p.R6W Chr.1: 201194162-201194162 on Build GRCh38
6052959 c.8-3C>A p.? Chr.1: 201194151-201194151 on Build GRCh38
5468303 c.14T>C p.L5P Chr.1: 201194160-201194160 on Build GRCh38
5546376 c.87G>A p.P29P Chr.1: 201194233-201194233 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115470024 Chr.1: 201194390-201194390 on Build GRCh38
rs576640863 Chr.1: 201194477-201194477 on Build GRCh38
rs907799523 Chr.1: 201194117-201194117 on Build GRCh38
rs187854226 Chr.1: 201194285-201194285 on Build GRCh38
rs529127329 Chr.1: 201194294-201194294 on Build GRCh38
rs755402018 Chr.1: 201194345-201194345 on Build GRCh38
rs563717809 Chr.1: 201194525-201194525 on Build GRCh38
rs550131357 Chr.1: 201194252-201194252 on Build GRCh38
rs946162952 Chr.1: 201194453-201194453 on Build GRCh38
rs542939183 Chr.1: 201194230-201194230 on Build GRCh38
rs761562477 Chr.1: 201194232-201194232 on Build GRCh38
rs569089050 Chr.1: 201194308-201194308 on Build GRCh38
rs41313898 Chr.1: 201194178-201194178 on Build GRCh38
rs996929334 Chr.1: 201194513-201194513 on Build GRCh38
rs983652497 Chr.1: 201194112-201194112 on Build GRCh38
rs1029659470 Chr.1: 201194350-201194350 on Build GRCh38
rs111943493 Chr.1: 201194491-201194491 on Build GRCh38
rs369077755 Chr.1: 201194200-201194200 on Build GRCh38
rs558151875 Chr.1: 201194346-201194346 on Build GRCh38
rs1000670373 Chr.1: 201194126-201194126 on Build GRCh38
rs116787657 Chr.1: 201194449-201194449 on Build GRCh38
rs776828401 Chr.1: 201194265-201194265 on Build GRCh38
rs1037618364 Chr.1: 201194159-201194159 on Build GRCh38
rs758717766 Chr.1: 201194305-201194305 on Build GRCh38
rs538165140 Chr.1: 201194323-201194323 on Build GRCh38
rs1052159810 Chr.1: 201194421-201194421 on Build GRCh38
rs776638291 Chr.1: 201194220-201194220 on Build GRCh38
rs189690117 Chr.1: 201194177-201194177 on Build GRCh38
rs964126609 Chr.1: 201194300-201194300 on Build GRCh38
rs548916837 Chr.1: 201194295-201194295 on Build GRCh38
rs375099833 Chr.1: 201194184-201194184 on Build GRCh38
rs751751999 Chr.1: 201194360-201194360 on Build GRCh38
rs937084765 Chr.1: 201194424-201194424 on Build GRCh38
rs75327835 Chr.1: 201194475-201194475 on Build GRCh38
rs955384307 Chr.1: 201194353-201194353 on Build GRCh38
rs749825277 Chr.1: 201194169-201194169 on Build GRCh38
rs751911173 Chr.1: 201194535-201194535 on Build GRCh38
rs1033486186 Chr.1: 201194298-201194298 on Build GRCh38
rs191973848 Chr.1: 201194404-201194404 on Build GRCh38
rs973264901 Chr.1: 201194403-201194403 on Build GRCh38
rs936127314 Chr.1: 201194476-201194476 on Build GRCh38
rs568244491 Chr.1: 201194360-201194360 on Build GRCh38
rs546318729 Chr.1: 201194281-201194281 on Build GRCh38
rs183565237 Chr.1: 201194489-201194489 on Build GRCh38
rs1568809 Chr.1: 201194456-201194456 on Build GRCh38
rs941958169 Chr.1: 201194123-201194123 on Build GRCh38
rs147277196 Chr.1: 201194251-201194251 on Build GRCh38
rs372720929 Chr.1: 201194173-201194173 on Build GRCh38
rs900033168 Chr.1: 201194501-201194501 on Build GRCh38
rs770934558 Chr.1: 201194209-201194209 on Build GRCh38
rs143808417 Chr.1: 201194118-201194118 on Build GRCh38
rs752679608 Chr.1: 201194389-201194394 on Build GRCh38
rs1055395926 Chr.1: 201194245-201194245 on Build GRCh38
rs563155391 Chr.1: 201194233-201194233 on Build GRCh38
rs532848908 Chr.1: 201194266-201194266 on Build GRCh38
rs773343563 Chr.1: 201194188-201194188 on Build GRCh38
rs932019087 Chr.1: 201194419-201194419 on Build GRCh38
rs563754393 Chr.1: 201194259-201194259 on Build GRCh38
rs1042332763 Chr.1: 201194459-201194459 on Build GRCh38
rs372970660 Chr.1: 201194229-201194229 on Build GRCh38
rs746792420 Chr.1: 201194236-201194236 on Build GRCh38
rs765313702 Chr.1: 201194355-201194355 on Build GRCh38
rs115956461 Chr.1: 201194162-201194162 on Build GRCh38
rs540748723 Chr.1: 201194163-201194163 on Build GRCh38
rs987375159 Chr.1: 201194354-201194354 on Build GRCh38
rs949749711 Chr.1: 201194458-201194458 on Build GRCh38
rs1010406643 Chr.1: 201194191-201194191 on Build GRCh38

View Full Product Details