Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 201368083-201368592 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNNT2
Entrez Gene ID
7139
Gene Name
troponin T2, cardiac type
Gene Aliases
CMD1D, CMH2, CMPD2, LVNC6, RCM3, TnTC, cTnT
UniGene
Hs.533613
Gene Chromosome Location
Chr.1: 201359008-201377828 on Build GRCh38
Transcripts
NM_000364.3 NM_001001430.2 NM_001001431.2 NM_001001432.2 NM_001276345.1 NM_001276346.1 NM_001276347.1 XM_006711508.3 XM_006711509.3 XM_011509938.2 XM_011509939.1 XM_011509940.2 XM_011509941.2 XM_011509942.2 XM_011509943.2 XM_011509944.2 XM_011509946.1 XM_017002216.1 XM_017002217.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
902183 c.84G>A p.A28A Chr.1: 201368211-201368211 on Build GRCh38
902182 c.114C>T p.T38T Chr.1: 201368181-201368181 on Build GRCh38
4026930 c.137C>T p.A46V Chr.1: 201368194-201368194 on Build GRCh38
4026931 c.86C>T p.A29V Chr.1: 201368194-201368194 on Build GRCh38
4872905 c.69G>A p.A23A Chr.1: 201368211-201368211 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745633544 Chr.1: 201368272-201368272 on Build GRCh38
rs774080530 Chr.1: 201368129-201368130 on Build GRCh38
rs907225035 Chr.1: 201368372-201368372 on Build GRCh38
rs766432591 Chr.1: 201368340-201368340 on Build GRCh38
rs745928510 Chr.1: 201368116-201368116 on Build GRCh38
rs867180029 Chr.1: 201368222-201368222 on Build GRCh38
rs756174289 Chr.1: 201368190-201368190 on Build GRCh38
rs757526942 Chr.1: 201368180-201368180 on Build GRCh38
rs1049178680 Chr.1: 201368319-201368319 on Build GRCh38
rs765718192 Chr.1: 201368137-201368137 on Build GRCh38
rs768621722 Chr.1: 201368118-201368118 on Build GRCh38
rs759076552 Chr.1: 201368242-201368245 on Build GRCh38
rs113983931 Chr.1: 201368230-201368230 on Build GRCh38
rs757361061 Chr.1: 201368115-201368115 on Build GRCh38
rs777845910 Chr.1: 201368352-201368352 on Build GRCh38
rs985899065 Chr.1: 201368455-201368455 on Build GRCh38
rs746492909 Chr.1: 201368181-201368181 on Build GRCh38
rs570070573 Chr.1: 201368303-201368303 on Build GRCh38
rs774827510 Chr.1: 201368412-201368412 on Build GRCh38
rs751742128 Chr.1: 201368111-201368111 on Build GRCh38
rs772769248 Chr.1: 201368123-201368123 on Build GRCh38
rs201414661 Chr.1: 201368122-201368122 on Build GRCh38
rs559011717 Chr.1: 201368334-201368334 on Build GRCh38
rs928958298 Chr.1: 201368563-201368563 on Build GRCh38
rs985868008 Chr.1: 201368291-201368291 on Build GRCh38
rs753185151 Chr.1: 201368140-201368140 on Build GRCh38
rs778730615 Chr.1: 201368194-201368194 on Build GRCh38
rs911587973 Chr.1: 201368292-201368292 on Build GRCh38
rs984511988 Chr.1: 201368568-201368568 on Build GRCh38
rs3753999 Chr.1: 201368358-201368358 on Build GRCh38
rs529140706 Chr.1: 201368539-201368539 on Build GRCh38
rs930819916 Chr.1: 201368309-201368309 on Build GRCh38
rs769274193 Chr.1: 201368403-201368403 on Build GRCh38
rs918945214 Chr.1: 201368241-201368241 on Build GRCh38
rs769213463 Chr.1: 201368367-201368367 on Build GRCh38
rs45576939 Chr.1: 201368308-201368308 on Build GRCh38
rs982080257 Chr.1: 201368128-201368128 on Build GRCh38
rs1036634301 Chr.1: 201368298-201368298 on Build GRCh38
rs753838494 Chr.1: 201368341-201368341 on Build GRCh38
rs1035876678 Chr.1: 201368516-201368516 on Build GRCh38
rs397516447 Chr.1: 201368189-201368189 on Build GRCh38
rs199730723 Chr.1: 201368236-201368236 on Build GRCh38
rs35229393 Chr.1: 201368362-201368362 on Build GRCh38
rs527486692 Chr.1: 201368149-201368149 on Build GRCh38
rs554778649 Chr.1: 201368439-201368439 on Build GRCh38
rs956259802 Chr.1: 201368133-201368133 on Build GRCh38
rs113051005 Chr.1: 201368161-201368161 on Build GRCh38
rs879475861 Chr.1: 201368540-201368540 on Build GRCh38
rs960423916 Chr.1: 201368523-201368523 on Build GRCh38
rs770683881 Chr.1: 201368129-201368129 on Build GRCh38
rs111683656 Chr.1: 201368160-201368160 on Build GRCh38
rs767803250 Chr.1: 201368440-201368440 on Build GRCh38
rs202215532 Chr.1: 201368121-201368121 on Build GRCh38
rs76929290 Chr.1: 201368518-201368518 on Build GRCh38
rs776406819 Chr.1: 201368215-201368215 on Build GRCh38
rs1049458233 Chr.1: 201368323-201368323 on Build GRCh38
rs911607144 Chr.1: 201368505-201368505 on Build GRCh38
rs376607276 Chr.1: 201368391-201368391 on Build GRCh38
rs1030933834 Chr.1: 201368385-201368385 on Build GRCh38
rs763236220 Chr.1: 201368145-201368145 on Build GRCh38
rs775895925 Chr.1: 201368231-201368231 on Build GRCh38
rs991910950 Chr.1: 201368538-201368538 on Build GRCh38
rs758399522 Chr.1: 201368342-201368342 on Build GRCh38
rs1022170015 Chr.1: 201368401-201368401 on Build GRCh38
rs730881120 Chr.1: 201368162-201368162 on Build GRCh38
rs200283086 Chr.1: 201368211-201368211 on Build GRCh38
rs762162586 Chr.1: 201368271-201368271 on Build GRCh38
rs375343614 Chr.1: 201368134-201368134 on Build GRCh38
rs777894361 Chr.1: 201368263-201368263 on Build GRCh38
rs944400054 Chr.1: 201368556-201368556 on Build GRCh38
rs200754249 Chr.1: 201368212-201368212 on Build GRCh38
rs770771962 Chr.1: 201368213-201368213 on Build GRCh38
rs891798584 Chr.1: 201368396-201368396 on Build GRCh38
rs944350429 Chr.1: 201368296-201368296 on Build GRCh38
rs746430628 Chr.1: 201368366-201368366 on Build GRCh38
rs746855269 Chr.1: 201368354-201368354 on Build GRCh38
rs775118066 Chr.1: 201368377-201368377 on Build GRCh38
rs45580032 Chr.1: 201368150-201368150 on Build GRCh38
rs576888252 Chr.1: 201368413-201368413 on Build GRCh38
rs953176159 Chr.1: 201368402-201368402 on Build GRCh38
rs3753998 Chr.1: 201368504-201368504 on Build GRCh38
rs943193497 Chr.1: 201368355-201368355 on Build GRCh38
rs767786507 Chr.1: 201368325-201368325 on Build GRCh38
rs750586455 Chr.1: 201368333-201368333 on Build GRCh38
rs956577127 Chr.1: 201368386-201368386 on Build GRCh38
rs931757029 Chr.1: 201368287-201368287 on Build GRCh38
rs989316733 Chr.1: 201368390-201368390 on Build GRCh38
rs2996498 Chr.1: 201368445-201368445 on Build GRCh38
rs727504254 Chr.1: 201368157-201368157 on Build GRCh38
rs370748496 Chr.1: 201368379-201368379 on Build GRCh38
rs760354030 Chr.1: 201368127-201368127 on Build GRCh38
rs367627730 Chr.1: 201368120-201368120 on Build GRCh38
rs999240712 Chr.1: 201368373-201368373 on Build GRCh38
rs748078123 Chr.1: 201368204-201368204 on Build GRCh38
rs1031661954 Chr.1: 201368126-201368126 on Build GRCh38

View Full Product Details