Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 201899835-201900337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMOD1
Entrez Gene ID
25802
Gene Name
leiomodin 1
Gene Aliases
1D, 64kD, D1, SM-LMOD, SMLMOD
UniGene
Hs.519075
Gene Chromosome Location
Chr.1: 201896456-201946588 on Build GRCh38
Transcripts
NM_012134.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3934394 c.816G>A p.K272K Chr.1: 201900197-201900197 on Build GRCh38
7195066 c.706G>A p.G236S Chr.1: 201900307-201900307 on Build GRCh38
5993407 c.1086G>A p.K362K Chr.1: 201899927-201899927 on Build GRCh38
4806858 c.1113C>T p.A371A Chr.1: 201899900-201899900 on Build GRCh38
6995981 c.826C>G p.P276A Chr.1: 201900187-201900187 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767379302 Chr.1: 201900279-201900279 on Build GRCh38
rs988887642 Chr.1: 201900182-201900182 on Build GRCh38
rs766682590 Chr.1: 201900187-201900187 on Build GRCh38
rs771378825 Chr.1: 201900249-201900249 on Build GRCh38
rs371488103 Chr.1: 201900114-201900114 on Build GRCh38
rs755855288 Chr.1: 201900220-201900220 on Build GRCh38
rs766631353 Chr.1: 201900089-201900089 on Build GRCh38
rs753423228 Chr.1: 201900222-201900222 on Build GRCh38
rs756949838 Chr.1: 201900034-201900034 on Build GRCh38
rs968309377 Chr.1: 201900168-201900168 on Build GRCh38
rs779712100 Chr.1: 201900221-201900221 on Build GRCh38
rs374097161 Chr.1: 201900083-201900083 on Build GRCh38
rs375832314 Chr.1: 201900005-201900005 on Build GRCh38
rs944314721 Chr.1: 201900299-201900299 on Build GRCh38
rs575923690 Chr.1: 201900029-201900029 on Build GRCh38
rs752738180 Chr.1: 201900022-201900022 on Build GRCh38
rs545021728 Chr.1: 201900127-201900127 on Build GRCh38
rs751891200 Chr.1: 201899978-201899978 on Build GRCh38
rs756222266 Chr.1: 201900014-201900014 on Build GRCh38
rs750669360 Chr.1: 201900095-201900095 on Build GRCh38
rs992229692 Chr.1: 201900269-201900269 on Build GRCh38
rs751574917 Chr.1: 201900209-201900209 on Build GRCh38
rs758404662 Chr.1: 201900023-201900023 on Build GRCh38
rs772006489 Chr.1: 201899900-201899900 on Build GRCh38
rs762270367 Chr.1: 201900072-201900072 on Build GRCh38
rs192215641 Chr.1: 201900218-201900218 on Build GRCh38
rs761862383 Chr.1: 201900272-201900272 on Build GRCh38
rs201119174 Chr.1: 201900285-201900285 on Build GRCh38
rs775405312 Chr.1: 201900266-201900266 on Build GRCh38
rs189201846 Chr.1: 201900186-201900186 on Build GRCh38
rs752025695 Chr.1: 201899904-201899904 on Build GRCh38
rs748544886 Chr.1: 201900147-201900147 on Build GRCh38
rs776940304 Chr.1: 201900190-201900190 on Build GRCh38
rs373771006 Chr.1: 201900016-201900016 on Build GRCh38
rs759717985 Chr.1: 201900198-201900198 on Build GRCh38
rs371011238 Chr.1: 201900056-201900056 on Build GRCh38
rs941293606 Chr.1: 201900297-201900297 on Build GRCh38
rs367728915 Chr.1: 201900139-201900139 on Build GRCh38
rs750571791 Chr.1: 201900013-201900013 on Build GRCh38
rs769355630 Chr.1: 201900052-201900052 on Build GRCh38
rs778272766 Chr.1: 201900230-201900230 on Build GRCh38
rs377022521 Chr.1: 201900113-201900113 on Build GRCh38
rs760795096 Chr.1: 201900085-201900085 on Build GRCh38
rs754498749 Chr.1: 201900229-201900229 on Build GRCh38
rs774792682 Chr.1: 201900065-201900065 on Build GRCh38
rs749837868 Chr.1: 201900055-201900055 on Build GRCh38
rs758455719 Chr.1: 201900096-201900096 on Build GRCh38
rs772543182 Chr.1: 201900150-201900150 on Build GRCh38
rs747396763 Chr.1: 201900244-201900244 on Build GRCh38
rs368890889 Chr.1: 201900015-201900015 on Build GRCh38
rs2820312 Chr.1: 201900129-201900129 on Build GRCh38
rs780925946 Chr.1: 201900040-201900040 on Build GRCh38
rs374052582 Chr.1: 201900238-201900238 on Build GRCh38
rs776995438 Chr.1: 201900256-201900256 on Build GRCh38
rs753545673 Chr.1: 201900308-201900308 on Build GRCh38
rs769986358 Chr.1: 201900260-201900260 on Build GRCh38
rs767708571 Chr.1: 201900080-201900080 on Build GRCh38
rs367984937 Chr.1: 201900281-201900281 on Build GRCh38
rs763440069 Chr.1: 201899965-201899965 on Build GRCh38
rs770583517 Chr.1: 201899909-201899909 on Build GRCh38
rs774928063 Chr.1: 201900140-201900140 on Build GRCh38
rs574701344 Chr.1: 201899906-201899906 on Build GRCh38
rs772426326 Chr.1: 201900234-201900234 on Build GRCh38
rs375079882 Chr.1: 201899891-201899891 on Build GRCh38
rs775994785 Chr.1: 201899964-201899964 on Build GRCh38
rs981981090 Chr.1: 201900099-201900099 on Build GRCh38
rs767389358 Chr.1: 201900217-201900217 on Build GRCh38
rs748818678 Chr.1: 201900138-201900138 on Build GRCh38
rs756098310 Chr.1: 201899885-201899885 on Build GRCh38
rs377506699 Chr.1: 201900053-201900053 on Build GRCh38
rs34071835 Chr.1: 201900185-201900185 on Build GRCh38
rs761991639 Chr.1: 201899989-201899989 on Build GRCh38
rs745456183 Chr.1: 201900044-201900044 on Build GRCh38
rs759823087 Chr.1: 201900259-201900259 on Build GRCh38
rs909800024 Chr.1: 201900286-201900286 on Build GRCh38
rs769123328 Chr.1: 201899937-201899937 on Build GRCh38
rs766105756 Chr.1: 201900307-201900307 on Build GRCh38
rs372721398 Chr.1: 201899920-201899920 on Build GRCh38
rs1002151471 Chr.1: 201900169-201900169 on Build GRCh38
rs745604964 Chr.1: 201900128-201900128 on Build GRCh38
rs779588952 Chr.1: 201900131-201900131 on Build GRCh38
rs777696417 Chr.1: 201899905-201899905 on Build GRCh38
rs199860756 Chr.1: 201899899-201899899 on Build GRCh38
rs565415486 Chr.1: 201900026-201900026 on Build GRCh38
rs754619674 Chr.1: 201900310-201900310 on Build GRCh38
rs547386916 Chr.1: 201900214-201900214 on Build GRCh38
rs759104821 Chr.1: 201899921-201899921 on Build GRCh38

View Full Product Details