Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202214075-202214544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LGR6
Entrez Gene ID
59352
Gene Name
leucine rich repeat containing G protein-coupled receptor 6
Gene Aliases
GPCR, VTS20631
UniGene
Hs.497402
Gene Chromosome Location
Chr.1: 202193990-202319761 on Build GRCh38
Transcripts
NM_001017403.1 NM_001017404.1 NM_021636.2 XM_005245404.3 XM_011509838.2 XM_011509839.2 XM_011509840.2 XM_017001996.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3750767 c.45C>T p.N15N Chr.1: 202214199-202214199 on Build GRCh38
3746727 c.26C>T p.S9L Chr.1: 202214180-202214180 on Build GRCh38
6411988 c.13G>T p.G5* Chr.1: 202214167-202214167 on Build GRCh38
6431014 c.4C>T p.R2C Chr.1: 202214158-202214158 on Build GRCh38
5422793 c.28G>C p.A10P Chr.1: 202214182-202214182 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962015529 Chr.1: 202214314-202214314 on Build GRCh38
rs775248579 Chr.1: 202214250-202214250 on Build GRCh38
rs370092675 Chr.1: 202214140-202214140 on Build GRCh38
rs952671555 Chr.1: 202214354-202214354 on Build GRCh38
rs966893174 Chr.1: 202214151-202214151 on Build GRCh38
rs918463756 Chr.1: 202214341-202214341 on Build GRCh38
rs367967876 Chr.1: 202214187-202214187 on Build GRCh38
rs995634952 Chr.1: 202214109-202214109 on Build GRCh38
rs374899792 Chr.1: 202214159-202214159 on Build GRCh38
rs752398460 Chr.1: 202214158-202214158 on Build GRCh38
rs1033003828 Chr.1: 202214471-202214471 on Build GRCh38
rs762676511 Chr.1: 202214263-202214263 on Build GRCh38
rs958265185 Chr.1: 202214472-202214472 on Build GRCh38
rs910931790 Chr.1: 202214132-202214132 on Build GRCh38
rs975281413 Chr.1: 202214480-202214480 on Build GRCh38
rs1035102360 Chr.1: 202214285-202214285 on Build GRCh38
rs900227187 Chr.1: 202214433-202214462 on Build GRCh38
rs751091875 Chr.1: 202214167-202214167 on Build GRCh38
rs140165672 Chr.1: 202214177-202214177 on Build GRCh38
rs911042086 Chr.1: 202214418-202214418 on Build GRCh38
rs768895294 Chr.1: 202214107-202214107 on Build GRCh38
rs921670113 Chr.1: 202214206-202214206 on Build GRCh38
rs992110249 Chr.1: 202214127-202214127 on Build GRCh38
rs768988876 Chr.1: 202214189-202214189 on Build GRCh38
rs200974439 Chr.1: 202214121-202214121 on Build GRCh38
rs1027671882 Chr.1: 202214525-202214525 on Build GRCh38
rs79821910 Chr.1: 202214199-202214199 on Build GRCh38
rs931856985 Chr.1: 202214489-202214518 on Build GRCh38
rs199949491 Chr.1: 202214266-202214266 on Build GRCh38
rs776657457 Chr.1: 202214236-202214236 on Build GRCh38
rs1001774097 Chr.1: 202214398-202214398 on Build GRCh38
rs978343055 Chr.1: 202214453-202214453 on Build GRCh38
rs559702002 Chr.1: 202214522-202214522 on Build GRCh38
rs536928069 Chr.1: 202214182-202214182 on Build GRCh38
rs182843779 Chr.1: 202214280-202214280 on Build GRCh38
rs895188610 Chr.1: 202214419-202214419 on Build GRCh38
rs374020393 Chr.1: 202214340-202214340 on Build GRCh38
rs966947253 Chr.1: 202214435-202214435 on Build GRCh38
rs1024909141 Chr.1: 202214424-202214424 on Build GRCh38
rs776367294 Chr.1: 202214122-202214122 on Build GRCh38
rs761408244 Chr.1: 202214274-202214274 on Build GRCh38
rs993429444 Chr.1: 202214524-202214524 on Build GRCh38
rs577582058 Chr.1: 202214342-202214342 on Build GRCh38
rs933040363 Chr.1: 202214500-202214500 on Build GRCh38
rs759361826 Chr.1: 202214136-202214136 on Build GRCh38
rs987146074 Chr.1: 202214510-202214510 on Build GRCh38
rs10920362 Chr.1: 202214180-202214180 on Build GRCh38
rs1051391791 Chr.1: 202214234-202214234 on Build GRCh38
rs756651501 Chr.1: 202214171-202214171 on Build GRCh38
rs772005735 Chr.1: 202214210-202214210 on Build GRCh38
rs201887207 Chr.1: 202214426-202214435 on Build GRCh38
rs1048473598 Chr.1: 202214467-202214467 on Build GRCh38
rs781736219 Chr.1: 202214173-202214173 on Build GRCh38
rs372072332 Chr.1: 202214202-202214202 on Build GRCh38
rs1037611774 Chr.1: 202214325-202214325 on Build GRCh38
rs745785601 Chr.1: 202214242-202214242 on Build GRCh38
rs577156669 Chr.1: 202214485-202214485 on Build GRCh38
rs972039687 Chr.1: 202214319-202214319 on Build GRCh38
rs368227880 Chr.1: 202214265-202214265 on Build GRCh38
rs984336173 Chr.1: 202214378-202214378 on Build GRCh38
rs74355478 Chr.1: 202214230-202214230 on Build GRCh38
rs962106871 Chr.1: 202214502-202214502 on Build GRCh38
rs558815389 Chr.1: 202214422-202214422 on Build GRCh38
rs78418110 Chr.1: 202214209-202214209 on Build GRCh38
rs760589808 Chr.1: 202214118-202214118 on Build GRCh38
rs1033804960 Chr.1: 202214412-202214412 on Build GRCh38
rs557527370 Chr.1: 202214333-202214333 on Build GRCh38
rs774486421 Chr.1: 202214116-202214116 on Build GRCh38
rs752927469 Chr.1: 202214377-202214377 on Build GRCh38
rs888492684 Chr.1: 202214494-202214494 on Build GRCh38

View Full Product Details