Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205145332-205145864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSTYK
Entrez Gene ID
25778
Gene Name
dual serine/threonine and tyrosine protein kinase
Gene Aliases
CAKUT1, DustyPK, HDCMD38P, RIP5, RIPK5
UniGene
Hs.6874
Gene Chromosome Location
Chr.1: 205142497-205211599 on Build GRCh38
Transcripts
NM_015375.2 NM_199462.2 XM_011509392.2 XM_011509393.2 XM_011509394.2

SNP Detail

SNP ID Chromosome Location
rs1046545686 Chr.1: 205145417-205145417 on Build GRCh38
rs928323088 Chr.1: 205145526-205145530 on Build GRCh38
rs780434780 Chr.1: 205145716-205145716 on Build GRCh38
rs1045812880 Chr.1: 205145523-205145525 on Build GRCh38
rs763422293 Chr.1: 205145663-205145663 on Build GRCh38
rs1019536611 Chr.1: 205145386-205145386 on Build GRCh38
rs966681657 Chr.1: 205145406-205145410 on Build GRCh38
rs948628094 Chr.1: 205145521-205145521 on Build GRCh38
rs765615425 Chr.1: 205145776-205145776 on Build GRCh38
rs995117449 Chr.1: 205145529-205145529 on Build GRCh38
rs928100280 Chr.1: 205145423-205145423 on Build GRCh38
rs753065732 Chr.1: 205145828-205145828 on Build GRCh38
rs61220458 Chr.1: 205145536-205145537 on Build GRCh38
rs141333991 Chr.1: 205145620-205145620 on Build GRCh38
rs989974612 Chr.1: 205145507-205145507 on Build GRCh38
rs533137408 Chr.1: 205145532-205145532 on Build GRCh38
rs1055308088 Chr.1: 205145526-205145526 on Build GRCh38
rs890039887 Chr.1: 205145541-205145541 on Build GRCh38
rs913880583 Chr.1: 205145590-205145590 on Build GRCh38
rs938078463 Chr.1: 205145652-205145652 on Build GRCh38
rs778197056 Chr.1: 205145525-205145525 on Build GRCh38
rs889997463 Chr.1: 205145778-205145778 on Build GRCh38
rs915825103 Chr.1: 205145515-205145516 on Build GRCh38
rs535166992 Chr.1: 205145574-205145574 on Build GRCh38
rs945425913 Chr.1: 205145406-205145406 on Build GRCh38
rs12137465 Chr.1: 205145537-205145537 on Build GRCh38
rs1033231653 Chr.1: 205145792-205145792 on Build GRCh38
rs886133350 Chr.1: 205145531-205145531 on Build GRCh38
rs957494523 Chr.1: 205145566-205145566 on Build GRCh38
rs756406959 Chr.1: 205145535-205145536 on Build GRCh38
rs553580489 Chr.1: 205145807-205145807 on Build GRCh38
rs990394511 Chr.1: 205145636-205145636 on Build GRCh38
rs1010552034 Chr.1: 205145829-205145829 on Build GRCh38
rs112877646 Chr.1: 205145786-205145786 on Build GRCh38
rs931039306 Chr.1: 205145757-205145758 on Build GRCh38
rs113671831 Chr.1: 205145787-205145787 on Build GRCh38
rs970293093 Chr.1: 205145690-205145690 on Build GRCh38
rs1036807889 Chr.1: 205145526-205145527 on Build GRCh38
rs67700841 Chr.1: 205145537-205145537 on Build GRCh38
rs913922155 Chr.1: 205145389-205145389 on Build GRCh38
rs189180628 Chr.1: 205145624-205145624 on Build GRCh38
rs112315860 Chr.1: 205145675-205145675 on Build GRCh38
rs140446140 Chr.1: 205145788-205145788 on Build GRCh38
rs1047818406 Chr.1: 205145530-205145530 on Build GRCh38
rs964509407 Chr.1: 205145534-205145534 on Build GRCh38
rs754053404 Chr.1: 205145715-205145715 on Build GRCh38
rs12041440 Chr.1: 205145525-205145525 on Build GRCh38
rs550906995 Chr.1: 205145504-205145504 on Build GRCh38
rs113377198 Chr.1: 205145607-205145607 on Build GRCh38
rs925359608 Chr.1: 205145407-205145407 on Build GRCh38
rs957454719 Chr.1: 205145479-205145479 on Build GRCh38
rs958116977 Chr.1: 205145634-205145634 on Build GRCh38
rs527679320 Chr.1: 205145650-205145650 on Build GRCh38
rs796146835 Chr.1: 205145536-205145537 on Build GRCh38
rs530784820 Chr.1: 205145505-205145505 on Build GRCh38
rs566030580 Chr.1: 205145565-205145565 on Build GRCh38
rs1008653325 Chr.1: 205145366-205145366 on Build GRCh38
rs766590479 Chr.1: 205145676-205145676 on Build GRCh38
rs1040025818 Chr.1: 205145744-205145744 on Build GRCh38
rs66966420 Chr.1: 205145366-205145368 on Build GRCh38
rs771578397 Chr.1: 205145365-205145366 on Build GRCh38
rs796558709 Chr.1: 205145528-205145529 on Build GRCh38
rs927999157 Chr.1: 205145633-205145633 on Build GRCh38
rs555030058 Chr.1: 205145525-205145526 on Build GRCh38
rs930875659 Chr.1: 205145707-205145707 on Build GRCh38
rs12048743 Chr.1: 205145745-205145745 on Build GRCh38
rs899373002 Chr.1: 205145527-205145527 on Build GRCh38
rs938194470 Chr.1: 205145524-205145524 on Build GRCh38
rs566871089 Chr.1: 205145706-205145710 on Build GRCh38
rs1047997996 Chr.1: 205145708-205145708 on Build GRCh38
rs1018687050 Chr.1: 205145533-205145533 on Build GRCh38
rs757588333 Chr.1: 205145365-205145367 on Build GRCh38
rs1029011435 Chr.1: 205145363-205145363 on Build GRCh38
rs989849919 Chr.1: 205145378-205145378 on Build GRCh38
rs200022188 Chr.1: 205145538-205145538 on Build GRCh38
rs973061773 Chr.1: 205145426-205145426 on Build GRCh38
rs192899888 Chr.1: 205145689-205145689 on Build GRCh38
rs898225007 Chr.1: 205145527-205145528 on Build GRCh38
rs902553359 Chr.1: 205145581-205145581 on Build GRCh38
rs36093419 Chr.1: 205145368-205145370 on Build GRCh38

View Full Product Details