Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206024000-206023496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSE
Entrez Gene ID
1510
Gene Name
cathepsin E
Gene Aliases
CATE
UniGene
Hs.644082
Gene Chromosome Location
Chr.1: 206009264-206023909 on Build GRCh38
Transcripts
NM_001317331.1 NM_001910.3 NM_148964.2 XM_011509244.1 XM_011509245.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1237080 c.22C>T p.L8L Chr.1: 206023770-206023770 on Build GRCh38
6124007 c.33_34CC>TT p.(=) Chr.1: 206023758-206023759 on Build GRCh38
7363272 c.67A>T p.R23W Chr.1: 206023725-206023725 on Build GRCh38
3482383 c.43G>A p.G15R Chr.1: 206023749-206023749 on Build GRCh38
1736307 c.8C>T p.T3M Chr.1: 206023784-206023784 on Build GRCh38
View More

View Full Product Details