Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206731647-206732163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPKAPK2
Entrez Gene ID
9261
Gene Name
mitogen-activated protein kinase-activated protein kinase 2
Gene Aliases
MAPKAP-K2, MK-2, MK2
UniGene
Hs.643566
Gene Chromosome Location
Chr.1: 206684912-206734285 on Build GRCh38
Transcripts
NM_004759.4 NM_032960.3 XM_005273353.3 XM_017002810.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5982528 c.1019G>A p.R340Q Chr.1: 206731879-206731879 on Build GRCh38
1491898 c.944C>A p.T315N Chr.1: 206731691-206731691 on Build GRCh38
5982529 c.1019G>A p.R340Q Chr.1: 206731879-206731879 on Build GRCh38
5705166 c.908G>A p.R303Q Chr.1: 206731655-206731655 on Build GRCh38
3750826 c.951C>T p.T317T Chr.1: 206731698-206731698 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs539375813 Chr.1: 206732118-206732118 on Build GRCh38
rs782592157 Chr.1: 206732128-206732128 on Build GRCh38
rs368050874 Chr.1: 206732037-206732037 on Build GRCh38
rs781906292 Chr.1: 206731993-206731993 on Build GRCh38
rs782072519 Chr.1: 206731842-206731842 on Build GRCh38
rs781860023 Chr.1: 206732072-206732072 on Build GRCh38
rs371877999 Chr.1: 206732090-206732090 on Build GRCh38
rs782491525 Chr.1: 206731895-206731895 on Build GRCh38
rs782425128 Chr.1: 206731839-206731839 on Build GRCh38
rs781790805 Chr.1: 206731802-206731802 on Build GRCh38
rs782434035 Chr.1: 206731722-206731722 on Build GRCh38
rs970977724 Chr.1: 206731849-206731849 on Build GRCh38
rs782749899 Chr.1: 206731903-206731903 on Build GRCh38
rs369480803 Chr.1: 206731747-206731747 on Build GRCh38
rs191198172 Chr.1: 206731971-206731971 on Build GRCh38
rs148086593 Chr.1: 206731853-206731853 on Build GRCh38
rs183160052 Chr.1: 206732050-206732050 on Build GRCh38
rs782450012 Chr.1: 206731740-206731740 on Build GRCh38
rs782262733 Chr.1: 206732027-206732027 on Build GRCh38
rs782384803 Chr.1: 206732048-206732048 on Build GRCh38
rs781837159 Chr.1: 206732129-206732129 on Build GRCh38
rs782713283 Chr.1: 206731884-206731884 on Build GRCh38
rs782309592 Chr.1: 206731960-206731960 on Build GRCh38
rs142025224 Chr.1: 206731878-206731878 on Build GRCh38
rs782780895 Chr.1: 206731862-206731862 on Build GRCh38
rs782597489 Chr.1: 206731679-206731679 on Build GRCh38
rs374110368 Chr.1: 206732021-206732021 on Build GRCh38
rs186406351 Chr.1: 206731733-206731733 on Build GRCh38
rs782285028 Chr.1: 206731831-206731831 on Build GRCh38
rs138882413 Chr.1: 206731841-206731841 on Build GRCh38
rs1047300594 Chr.1: 206731765-206731765 on Build GRCh38
rs782002200 Chr.1: 206731957-206731957 on Build GRCh38
rs782164664 Chr.1: 206731766-206731766 on Build GRCh38
rs782462168 Chr.1: 206732119-206732119 on Build GRCh38
rs781831820 Chr.1: 206732001-206732001 on Build GRCh38
rs994853586 Chr.1: 206731761-206731761 on Build GRCh38
rs376484035 Chr.1: 206731760-206731760 on Build GRCh38
rs782310577 Chr.1: 206732054-206732054 on Build GRCh38
rs782287868 Chr.1: 206731958-206731958 on Build GRCh38
rs782746203 Chr.1: 206731775-206731775 on Build GRCh38
rs782167091 Chr.1: 206732053-206732053 on Build GRCh38
rs782340351 Chr.1: 206731845-206731845 on Build GRCh38
rs782803294 Chr.1: 206731992-206731992 on Build GRCh38
rs782033903 Chr.1: 206731865-206731865 on Build GRCh38
rs782603179 Chr.1: 206731811-206731811 on Build GRCh38
rs782189785 Chr.1: 206731938-206731938 on Build GRCh38
rs781937063 Chr.1: 206731963-206731963 on Build GRCh38
rs782332601 Chr.1: 206731735-206731735 on Build GRCh38
rs782603935 Chr.1: 206731949-206731949 on Build GRCh38
rs782188773 Chr.1: 206732039-206732039 on Build GRCh38
rs781855787 Chr.1: 206731792-206731792 on Build GRCh38
rs782092001 Chr.1: 206732062-206732062 on Build GRCh38
rs371800749 Chr.1: 206732006-206732006 on Build GRCh38
rs782533777 Chr.1: 206731925-206731925 on Build GRCh38
rs147613858 Chr.1: 206731772-206731772 on Build GRCh38
rs1018321172 Chr.1: 206731773-206731773 on Build GRCh38
rs4073250 Chr.1: 206731698-206731698 on Build GRCh38
rs113044534 Chr.1: 206731726-206731726 on Build GRCh38
rs75615416 Chr.1: 206731752-206731752 on Build GRCh38
rs782082689 Chr.1: 206731964-206731964 on Build GRCh38
rs782241927 Chr.1: 206732046-206732046 on Build GRCh38
rs926328091 Chr.1: 206731744-206731744 on Build GRCh38
rs140988938 Chr.1: 206731918-206731918 on Build GRCh38
rs782531386 Chr.1: 206732028-206732028 on Build GRCh38
rs782192213 Chr.1: 206731734-206731734 on Build GRCh38
rs941786201 Chr.1: 206731879-206731879 on Build GRCh38
rs557263261 Chr.1: 206732020-206732020 on Build GRCh38
rs147497555 Chr.1: 206732058-206732058 on Build GRCh38
rs781819454 Chr.1: 206731889-206731889 on Build GRCh38
rs782458800 Chr.1: 206731944-206731944 on Build GRCh38
rs782572647 Chr.1: 206731942-206731942 on Build GRCh38
rs782279440 Chr.1: 206731705-206731705 on Build GRCh38
rs782440806 Chr.1: 206731995-206731995 on Build GRCh38
rs781971525 Chr.1: 206732044-206732044 on Build GRCh38
rs782225811 Chr.1: 206731952-206731952 on Build GRCh38
rs34526867 Chr.1: 206731695-206731695 on Build GRCh38
rs782749126 Chr.1: 206732064-206732064 on Build GRCh38
rs548381343 Chr.1: 206731970-206731970 on Build GRCh38
rs782345388 Chr.1: 206732040-206732040 on Build GRCh38
rs782689474 Chr.1: 206731927-206731927 on Build GRCh38
rs190801417 Chr.1: 206731804-206731804 on Build GRCh38
rs377553675 Chr.1: 206731815-206731815 on Build GRCh38
rs782395395 Chr.1: 206731754-206731754 on Build GRCh38
rs781947422 Chr.1: 206732061-206732061 on Build GRCh38
rs374876954 Chr.1: 206732098-206732098 on Build GRCh38
rs374478223 Chr.1: 206731847-206731847 on Build GRCh38
rs368347735 Chr.1: 206731998-206731998 on Build GRCh38

View Full Product Details