Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 220690168-220690698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf115
Entrez Gene ID
79762
Gene Name
chromosome 1 open reading frame 115
Gene Aliases
-
UniGene
Hs.519839
Gene Chromosome Location
Chr.1: 220690286-220699157 on Build GRCh38
Transcripts
NM_024709.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4855001 c.36G>C p.E12D Chr.1: 220690438-220690438 on Build GRCh38
6363296 c.160A>G p.S54G Chr.1: 220690562-220690562 on Build GRCh38
5969166 c.5C>T p.T2M Chr.1: 220690407-220690407 on Build GRCh38
4650297 c.274A>G p.K92E Chr.1: 220690676-220690676 on Build GRCh38
5992499 c.102C>A p.A34A Chr.1: 220690504-220690504 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766570284 Chr.1: 220690664-220690664 on Build GRCh38
rs981083621 Chr.1: 220690581-220690581 on Build GRCh38
rs199949758 Chr.1: 220690395-220690395 on Build GRCh38
rs1027123785 Chr.1: 220690299-220690299 on Build GRCh38
rs748048982 Chr.1: 220690361-220690361 on Build GRCh38
rs761203884 Chr.1: 220690663-220690663 on Build GRCh38
rs750453432 Chr.1: 220690570-220690570 on Build GRCh38
rs760647085 Chr.1: 220690627-220690627 on Build GRCh38
rs765182139 Chr.1: 220690636-220690636 on Build GRCh38
rs975725070 Chr.1: 220690234-220690234 on Build GRCh38
rs778809627 Chr.1: 220690588-220690588 on Build GRCh38
rs925046204 Chr.1: 220690250-220690250 on Build GRCh38
rs753734015 Chr.1: 220690578-220690578 on Build GRCh38
rs56959779 Chr.1: 220690284-220690284 on Build GRCh38
rs559559807 Chr.1: 220690190-220690190 on Build GRCh38
rs745538171 Chr.1: 220690657-220690657 on Build GRCh38
rs574239920 Chr.1: 220690613-220690613 on Build GRCh38
rs767644565 Chr.1: 220690558-220690558 on Build GRCh38
rs770667491 Chr.1: 220690601-220690601 on Build GRCh38
rs60340434 Chr.1: 220690280-220690280 on Build GRCh38
rs550973161 Chr.1: 220690304-220690304 on Build GRCh38
rs917819169 Chr.1: 220690318-220690318 on Build GRCh38
rs931191203 Chr.1: 220690326-220690326 on Build GRCh38
rs767903191 Chr.1: 220690618-220690618 on Build GRCh38
rs995994740 Chr.1: 220690197-220690197 on Build GRCh38
rs969261267 Chr.1: 220690429-220690429 on Build GRCh38
rs947876852 Chr.1: 220690224-220690224 on Build GRCh38
rs549438764 Chr.1: 220690519-220690519 on Build GRCh38
rs901212816 Chr.1: 220690360-220690360 on Build GRCh38
rs80017388 Chr.1: 220690241-220690241 on Build GRCh38
rs992113781 Chr.1: 220690306-220690306 on Build GRCh38
rs754841450 Chr.1: 220690584-220690584 on Build GRCh38
rs1032014819 Chr.1: 220690444-220690444 on Build GRCh38
rs748428279 Chr.1: 220690660-220690660 on Build GRCh38
rs562575200 Chr.1: 220690270-220690270 on Build GRCh38
rs951780609 Chr.1: 220690300-220690300 on Build GRCh38
rs773613083 Chr.1: 220690662-220690662 on Build GRCh38
rs530777958 Chr.1: 220690301-220690301 on Build GRCh38
rs775964993 Chr.1: 220690447-220690447 on Build GRCh38
rs1028907004 Chr.1: 220690235-220690235 on Build GRCh38
rs888586612 Chr.1: 220690434-220690434 on Build GRCh38
rs545212089 Chr.1: 220690656-220690656 on Build GRCh38
rs1004181717 Chr.1: 220690349-220690349 on Build GRCh38
rs546469178 Chr.1: 220690343-220690343 on Build GRCh38
rs568991746 Chr.1: 220690438-220690438 on Build GRCh38
rs769418928 Chr.1: 220690659-220690659 on Build GRCh38
rs780154748 Chr.1: 220690557-220690569 on Build GRCh38
rs376728254 Chr.1: 220690600-220690600 on Build GRCh38
rs1027094963 Chr.1: 220690209-220690209 on Build GRCh38
rs947962087 Chr.1: 220690319-220690319 on Build GRCh38
rs751772788 Chr.1: 220690581-220690582 on Build GRCh38
rs554579162 Chr.1: 220690572-220690572 on Build GRCh38
rs567425520 Chr.1: 220690305-220690305 on Build GRCh38
rs530038441 Chr.1: 220690324-220690324 on Build GRCh38
rs448634 Chr.1: 220690194-220690194 on Build GRCh38
rs774787613 Chr.1: 220690446-220690446 on Build GRCh38
rs984465940 Chr.1: 220690192-220690192 on Build GRCh38
rs936769527 Chr.1: 220690333-220690333 on Build GRCh38
rs573058187 Chr.1: 220690651-220690651 on Build GRCh38
rs1008506944 Chr.1: 220690257-220690257 on Build GRCh38
rs965625742 Chr.1: 220690231-220690231 on Build GRCh38
rs370724628 Chr.1: 220690239-220690239 on Build GRCh38
rs759881686 Chr.1: 220690606-220690606 on Build GRCh38
rs754142767 Chr.1: 220690630-220690630 on Build GRCh38
rs762436308 Chr.1: 220690615-220690615 on Build GRCh38
rs900039961 Chr.1: 220690274-220690274 on Build GRCh38
rs1024604064 Chr.1: 220690449-220690449 on Build GRCh38
rs537416758 Chr.1: 220690484-220690484 on Build GRCh38
rs552264612 Chr.1: 220690402-220690402 on Build GRCh38
rs758492110 Chr.1: 220690436-220690437 on Build GRCh38
rs752536191 Chr.1: 220690642-220690642 on Build GRCh38
rs903954779 Chr.1: 220690230-220690230 on Build GRCh38
rs755184006 Chr.1: 220690632-220690632 on Build GRCh38
rs763283894 Chr.1: 220690468-220690468 on Build GRCh38
rs1041448851 Chr.1: 220690582-220690582 on Build GRCh38
rs758255547 Chr.1: 220690595-220690595 on Build GRCh38
rs544004947 Chr.1: 220690218-220690218 on Build GRCh38
rs1056756835 Chr.1: 220690219-220690219 on Build GRCh38
rs917787563 Chr.1: 220690220-220690220 on Build GRCh38
rs756751823 Chr.1: 220690653-220690653 on Build GRCh38
rs371317519 Chr.1: 220690665-220690665 on Build GRCh38
rs1041334267 Chr.1: 220690375-220690375 on Build GRCh38
rs925093713 Chr.1: 220690329-220690329 on Build GRCh38
rs1052455285 Chr.1: 220690334-220690334 on Build GRCh38
rs746918853 Chr.1: 220690652-220690652 on Build GRCh38
rs892488905 Chr.1: 220690341-220690341 on Build GRCh38
rs1045031753 Chr.1: 220690322-220690322 on Build GRCh38
rs766476940 Chr.1: 220690574-220690574 on Build GRCh38
rs899810311 Chr.1: 220690590-220690590 on Build GRCh38
rs900093021 Chr.1: 220690376-220690376 on Build GRCh38
rs748100113 Chr.1: 220690591-220690591 on Build GRCh38
rs758478552 Chr.1: 220690476-220690476 on Build GRCh38
rs987770810 Chr.1: 220690300-220690300 on Build GRCh38
rs1020265960 Chr.1: 220690263-220690263 on Build GRCh38
rs745484837 Chr.1: 220690607-220690607 on Build GRCh38
rs1052402128 Chr.1: 220690248-220690248 on Build GRCh38
rs892436208 Chr.1: 220690255-220690255 on Build GRCh38
rs997416943 Chr.1: 220690296-220690296 on Build GRCh38
rs553252064 Chr.1: 220690646-220690646 on Build GRCh38
rs566719988 Chr.1: 220690383-220690383 on Build GRCh38
rs867181738 Chr.1: 220690501-220690501 on Build GRCh38
rs536997214 Chr.1: 220690628-220690628 on Build GRCh38
rs764396656 Chr.1: 220690516-220690516 on Build GRCh38
rs773378717 Chr.1: 220690622-220690622 on Build GRCh38
rs1019618636 Chr.1: 220690353-220690353 on Build GRCh38
rs937274317 Chr.1: 220690604-220690604 on Build GRCh38
rs1045185199 Chr.1: 220690229-220690229 on Build GRCh38
rs934972813 Chr.1: 220690253-220690253 on Build GRCh38
rs777409359 Chr.1: 220690599-220690599 on Build GRCh38
rs980682394 Chr.1: 220690500-220690500 on Build GRCh38
rs1027176505 Chr.1: 220690396-220690396 on Build GRCh38
rs1055608198 Chr.1: 220690608-220690608 on Build GRCh38
rs867080655 Chr.1: 220690442-220690442 on Build GRCh38
rs988222523 Chr.1: 220690571-220690571 on Build GRCh38
rs1045311328 Chr.1: 220690193-220690193 on Build GRCh38

View Full Product Details