Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235448622-235449168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GALNT2
Entrez Gene ID
148789
Gene Name
beta-1,3-N-acetylgalactosaminyltransferase 2
Gene Aliases
B3GalNAc-T2, MDDGA11
UniGene
Hs.498143
Gene Chromosome Location
Chr.1: 235440654-235504481 on Build GRCh38
Transcripts
NM_152490.4 XM_006711749.3 XM_017000394.1 XM_017000395.1 NM_001079515.2 NM_001287801.1 NM_001287802.1 NM_003193.4

Gene Symbol
TBCE
Entrez Gene ID
6905
Gene Name
tubulin folding cofactor E
Gene Aliases
HRD, KCS, KCS1, pac2
UniGene
Hs.744998
Gene Chromosome Location
Chr.1: 235367360-235448968 on Build GRCh38
Transcripts
NM_152490.4 XM_006711749.3 XM_017000394.1 XM_017000395.1 NM_001079515.2 NM_001287801.1 NM_001287802.1 NM_003193.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
300638 c.1535T>G p.L512* Chr.1: 235448713-235448713 on Build GRCh38
5987876 c.1712G>C p.G571A Chr.1: 235448737-235448737 on Build GRCh38
905841 c.1573G>A p.V525M Chr.1: 235448751-235448751 on Build GRCh38
6218207 c.1539G>C p.Q513H Chr.1: 235448717-235448717 on Build GRCh38
6218208 c.1692G>C p.Q564H Chr.1: 235448717-235448717 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746868782 Chr.1: 235448661-235448662 on Build GRCh38
rs974347125 Chr.1: 235449088-235449088 on Build GRCh38
rs1014748161 Chr.1: 235449091-235449091 on Build GRCh38
rs773529565 Chr.1: 235448693-235448693 on Build GRCh38
rs967920522 Chr.1: 235449103-235449103 on Build GRCh38
rs981317535 Chr.1: 235449136-235449136 on Build GRCh38
rs1027421692 Chr.1: 235448860-235448860 on Build GRCh38
rs952121633 Chr.1: 235448885-235448886 on Build GRCh38
rs781200142 Chr.1: 235448758-235448758 on Build GRCh38
rs769918946 Chr.1: 235448687-235448687 on Build GRCh38
rs369206654 Chr.1: 235449080-235449081 on Build GRCh38
rs753594563 Chr.1: 235448716-235448716 on Build GRCh38
rs757692186 Chr.1: 235448864-235448865 on Build GRCh38
rs946560126 Chr.1: 235448905-235448905 on Build GRCh38
rs112310923 Chr.1: 235448954-235448954 on Build GRCh38
rs535642426 Chr.1: 235449044-235449047 on Build GRCh38
rs886046152 Chr.1: 235448824-235448825 on Build GRCh38
rs952106324 Chr.1: 235449065-235449065 on Build GRCh38
rs756655831 Chr.1: 235448717-235448717 on Build GRCh38
rs771540713 Chr.1: 235448800-235448800 on Build GRCh38
rs556604625 Chr.1: 235448818-235448818 on Build GRCh38
rs774822889 Chr.1: 235448708-235448708 on Build GRCh38
rs773370448 Chr.1: 235448779-235448779 on Build GRCh38
rs566587741 Chr.1: 235449020-235449020 on Build GRCh38
rs776965318 Chr.1: 235448674-235448674 on Build GRCh38
rs964849747 Chr.1: 235448918-235448919 on Build GRCh38
rs907740419 Chr.1: 235448896-235448896 on Build GRCh38
rs766703830 Chr.1: 235448781-235448782 on Build GRCh38
rs369714783 Chr.1: 235448777-235448777 on Build GRCh38
rs746788701 Chr.1: 235448754-235448754 on Build GRCh38
rs575068441 Chr.1: 235448864-235448864 on Build GRCh38
rs139691464 Chr.1: 235448811-235448811 on Build GRCh38
rs776090134 Chr.1: 235448806-235448806 on Build GRCh38
rs180911749 Chr.1: 235449051-235449051 on Build GRCh38
rs996244211 Chr.1: 235448851-235448855 on Build GRCh38
rs754437381 Chr.1: 235448980-235448980 on Build GRCh38
rs1054064310 Chr.1: 235448966-235448967 on Build GRCh38
rs1028216424 Chr.1: 235449014-235449014 on Build GRCh38
rs902712992 Chr.1: 235449020-235449021 on Build GRCh38
rs911533241 Chr.1: 235448873-235448873 on Build GRCh38
rs35242859 Chr.1: 235449079-235449080 on Build GRCh38
rs528197044 Chr.1: 235448887-235448887 on Build GRCh38
rs776845780 Chr.1: 235449094-235449094 on Build GRCh38
rs140104308 Chr.1: 235448745-235448745 on Build GRCh38
rs746679827 Chr.1: 235448859-235448860 on Build GRCh38
rs758366722 Chr.1: 235448747-235448747 on Build GRCh38
rs771536610 Chr.1: 235449114-235449114 on Build GRCh38
rs540417673 Chr.1: 235449107-235449108 on Build GRCh38
rs148233223 Chr.1: 235448675-235448675 on Build GRCh38
rs749417404 Chr.1: 235448787-235448787 on Build GRCh38
rs763640897 Chr.1: 235448715-235448715 on Build GRCh38
rs748411410 Chr.1: 235448759-235448759 on Build GRCh38
rs904571818 Chr.1: 235448788-235448788 on Build GRCh38
rs767929260 Chr.1: 235448804-235448804 on Build GRCh38
rs763573622 Chr.1: 235448700-235448700 on Build GRCh38
rs886046153 Chr.1: 235448939-235448939 on Build GRCh38
rs560936483 Chr.1: 235448881-235448881 on Build GRCh38
rs550837792 Chr.1: 235448961-235448961 on Build GRCh38
rs950522785 Chr.1: 235449024-235449024 on Build GRCh38
rs535711932 Chr.1: 235449044-235449044 on Build GRCh38
rs1005005705 Chr.1: 235449069-235449070 on Build GRCh38
rs373300985 Chr.1: 235448801-235448801 on Build GRCh38
rs779655022 Chr.1: 235449021-235449023 on Build GRCh38
rs147400126 Chr.1: 235448656-235448656 on Build GRCh38
rs772808072 Chr.1: 235448734-235448749 on Build GRCh38
rs970219007 Chr.1: 235448817-235448817 on Build GRCh38
rs147519707 Chr.1: 235448744-235448744 on Build GRCh38
rs868603564 Chr.1: 235449130-235449130 on Build GRCh38
rs1027664898 Chr.1: 235449056-235449056 on Build GRCh38
rs747195091 Chr.1: 235448659-235448659 on Build GRCh38
rs780010345 Chr.1: 235448751-235448751 on Build GRCh38
rs751491289 Chr.1: 235448781-235448781 on Build GRCh38
rs564895195 Chr.1: 235448912-235448912 on Build GRCh38
rs905561514 Chr.1: 235448790-235448790 on Build GRCh38
rs926962008 Chr.1: 235448935-235448935 on Build GRCh38
rs965687744 Chr.1: 235449067-235449067 on Build GRCh38
rs944671621 Chr.1: 235448884-235448884 on Build GRCh38
rs1053160965 Chr.1: 235448960-235448960 on Build GRCh38
rs767215746 Chr.1: 235448816-235448816 on Build GRCh38
rs78867315 Chr.1: 235448870-235448870 on Build GRCh38
rs1005030702 Chr.1: 235449043-235449043 on Build GRCh38
rs534021496 Chr.1: 235449026-235449026 on Build GRCh38
rs973535871 Chr.1: 235448831-235448831 on Build GRCh38
rs367766528 Chr.1: 235449136-235449137 on Build GRCh38
rs7537 Chr.1: 235448904-235448904 on Build GRCh38
rs533888707 Chr.1: 235448672-235448672 on Build GRCh38
rs704705 Chr.1: 235449005-235449005 on Build GRCh38
rs768749834 Chr.1: 235448692-235448727 on Build GRCh38
rs1041048639 Chr.1: 235449026-235449027 on Build GRCh38
rs145256959 Chr.1: 235448991-235448991 on Build GRCh38
rs761424380 Chr.1: 235448706-235448707 on Build GRCh38
rs754180159 Chr.1: 235449023-235449024 on Build GRCh38
rs904630281 Chr.1: 235448923-235448923 on Build GRCh38
rs1014226363 Chr.1: 235448978-235448978 on Build GRCh38
rs761551269 Chr.1: 235448810-235448810 on Build GRCh38
rs140662460 Chr.1: 235448755-235448755 on Build GRCh38
rs764155861 Chr.1: 235448766-235448766 on Build GRCh38
rs535686495 Chr.1: 235448863-235448863 on Build GRCh38
rs189034185 Chr.1: 235448913-235448913 on Build GRCh38
rs766807109 Chr.1: 235448705-235448705 on Build GRCh38
rs944601368 Chr.1: 235448688-235448688 on Build GRCh38
rs950493106 Chr.1: 235448861-235448861 on Build GRCh38
rs556238424 Chr.1: 235449139-235449150 on Build GRCh38
rs971662481 Chr.1: 235449125-235449125 on Build GRCh38
rs372967513 Chr.1: 235448652-235448652 on Build GRCh38
rs771392750 Chr.1: 235449013-235449013 on Build GRCh38
rs769559799 Chr.1: 235448708-235448709 on Build GRCh38
rs934728694 Chr.1: 235448939-235448939 on Build GRCh38
rs763347083 Chr.1: 235448745-235448746 on Build GRCh38
rs562656404 Chr.1: 235448989-235448989 on Build GRCh38
rs776217784 Chr.1: 235448705-235448705 on Build GRCh38
rs202244855 Chr.1: 235448657-235448657 on Build GRCh38

View Full Product Details