Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 246945488-246946024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF670-ZNF695
Entrez Gene ID
100533111
Gene Name
ZNF670-ZNF695 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.732305
Gene Chromosome Location
Chr.1: 246945547-247078813 on Build GRCh38
Transcripts
NR_037894.1 NM_001204221.1 NR_037892.1

Gene Symbol
ZNF695
Entrez Gene ID
57116
Gene Name
zinc finger protein 695
Gene Aliases
SBZF3
UniGene
Hs.669893
Gene Chromosome Location
Chr.1: 246945547-247008093 on Build GRCh38
Transcripts
NR_037894.1 NM_001204221.1 NR_037892.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3741161 c.507C>G p.I169M Chr.1: 246945809-246945809 on Build GRCh38
5491325 c.489-4delT p.? Chr.1: 246945831-246945831 on Build GRCh38
1602058 c.491T>C p.V164A Chr.1: 246945825-246945825 on Build GRCh38
6577974 c.492C>G p.V164V Chr.1: 246945824-246945824 on Build GRCh38
4497258 c.492C>T p.V164V Chr.1: 246945824-246945824 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1045886718 Chr.1: 246945599-246945599 on Build GRCh38
rs767466574 Chr.1: 246945759-246945759 on Build GRCh38
rs951948980 Chr.1: 246945762-246945762 on Build GRCh38
rs536047782 Chr.1: 246945861-246945861 on Build GRCh38
rs6426191 Chr.1: 246945866-246945866 on Build GRCh38
rs376070027 Chr.1: 246945667-246945667 on Build GRCh38
rs370630789 Chr.1: 246945884-246945884 on Build GRCh38
rs766929840 Chr.1: 246945864-246945864 on Build GRCh38
rs535564327 Chr.1: 246945922-246945922 on Build GRCh38
rs866840110 Chr.1: 246945848-246945848 on Build GRCh38
rs756651607 Chr.1: 246945806-246945806 on Build GRCh38
rs1056414005 Chr.1: 246945654-246945654 on Build GRCh38
rs779523931 Chr.1: 246945972-246945972 on Build GRCh38
rs1000668029 Chr.1: 246945749-246945749 on Build GRCh38
rs966238237 Chr.1: 246945735-246945735 on Build GRCh38
rs951978796 Chr.1: 246945872-246945872 on Build GRCh38
rs111328919 Chr.1: 246945990-246945990 on Build GRCh38
rs567499163 Chr.1: 246945846-246945846 on Build GRCh38
rs550579788 Chr.1: 246945832-246945832 on Build GRCh38
rs751149453 Chr.1: 246945815-246945815 on Build GRCh38
rs773135492 Chr.1: 246945741-246945741 on Build GRCh38
rs907339257 Chr.1: 246945604-246945604 on Build GRCh38
rs185082300 Chr.1: 246945819-246945819 on Build GRCh38
rs41267541 Chr.1: 246945994-246945994 on Build GRCh38
rs765833870 Chr.1: 246945989-246945989 on Build GRCh38
rs996437048 Chr.1: 246945865-246945865 on Build GRCh38
rs776840588 Chr.1: 246945634-246945634 on Build GRCh38
rs947604829 Chr.1: 246945816-246945816 on Build GRCh38
rs756512217 Chr.1: 246945602-246945603 on Build GRCh38
rs760201635 Chr.1: 246945849-246945849 on Build GRCh38
rs768591835 Chr.1: 246945580-246945580 on Build GRCh38
rs551315725 Chr.1: 246945725-246945725 on Build GRCh38
rs180733755 Chr.1: 246945726-246945726 on Build GRCh38
rs981121130 Chr.1: 246945928-246945928 on Build GRCh38
rs200408255 Chr.1: 246945990-246945990 on Build GRCh38
rs984420931 Chr.1: 246945558-246945558 on Build GRCh38
rs766726374 Chr.1: 246945989-246945990 on Build GRCh38
rs1018783138 Chr.1: 246945734-246945734 on Build GRCh38
rs1014543516 Chr.1: 246945907-246945907 on Build GRCh38
rs1046701656 Chr.1: 246945708-246945708 on Build GRCh38
rs769658938 Chr.1: 246945723-246945723 on Build GRCh38
rs2642982 Chr.1: 246945712-246945712 on Build GRCh38
rs559412672 Chr.1: 246945669-246945669 on Build GRCh38
rs1011623816 Chr.1: 246945711-246945711 on Build GRCh38
rs928825145 Chr.1: 246945792-246945792 on Build GRCh38
rs984452021 Chr.1: 246945776-246945776 on Build GRCh38
rs752507505 Chr.1: 246945766-246945766 on Build GRCh38
rs930341050 Chr.1: 246945532-246945532 on Build GRCh38
rs753397612 Chr.1: 246945788-246945788 on Build GRCh38
rs767059038 Chr.1: 246945787-246945787 on Build GRCh38
rs937547983 Chr.1: 246945514-246945514 on Build GRCh38
rs2642983 Chr.1: 246945965-246945965 on Build GRCh38
rs528805814 Chr.1: 246945633-246945633 on Build GRCh38
rs907654536 Chr.1: 246945566-246945566 on Build GRCh38
rs866446496 Chr.1: 246945826-246945826 on Build GRCh38
rs1038579616 Chr.1: 246945533-246945533 on Build GRCh38
rs1055964574 Chr.1: 246945808-246945808 on Build GRCh38
rs893379993 Chr.1: 246945660-246945660 on Build GRCh38
rs974595874 Chr.1: 246945737-246945737 on Build GRCh38
rs927414250 Chr.1: 246945818-246945818 on Build GRCh38
rs1032178414 Chr.1: 246945754-246945754 on Build GRCh38
rs543156462 Chr.1: 246945612-246945612 on Build GRCh38
rs961653658 Chr.1: 246945925-246945925 on Build GRCh38

View Full Product Details