Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 5887756-5888268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD16
Entrez Gene ID
54522
Gene Name
ankyrin repeat domain 16
Gene Aliases
-
UniGene
Hs.289828
Gene Chromosome Location
Chr.10: 5861726-5889897 on Build GRCh38
Transcripts
NM_001009941.2 NM_001009943.2 NM_019046.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
397404 c.445G>A p.D149N Chr.10: 5887937-5887937 on Build GRCh38
1505562 c.373G>A p.E125K Chr.10: 5888009-5888009 on Build GRCh38
146871 c.383C>G p.A128G Chr.10: 5887999-5887999 on Build GRCh38
6461232 c.369G>C p.L123L Chr.10: 5888013-5888013 on Build GRCh38
6461231 c.370G>C p.V124L Chr.10: 5888012-5888012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764503814 Chr.10: 5888147-5888147 on Build GRCh38
rs958695813 Chr.10: 5887803-5887803 on Build GRCh38
rs746509178 Chr.10: 5887955-5887955 on Build GRCh38
rs758268673 Chr.10: 5888044-5888044 on Build GRCh38
rs1028080263 Chr.10: 5887794-5887794 on Build GRCh38
rs775860380 Chr.10: 5887853-5887853 on Build GRCh38
rs977301543 Chr.10: 5888226-5888226 on Build GRCh38
rs767754511 Chr.10: 5887892-5887892 on Build GRCh38
rs1013762451 Chr.10: 5888129-5888129 on Build GRCh38
rs932288252 Chr.10: 5887954-5887954 on Build GRCh38
rs373806653 Chr.10: 5887927-5887927 on Build GRCh38
rs991422995 Chr.10: 5887808-5887808 on Build GRCh38
rs543359172 Chr.10: 5888092-5888092 on Build GRCh38
rs756786475 Chr.10: 5888055-5888055 on Build GRCh38
rs377369009 Chr.10: 5887921-5887921 on Build GRCh38
rs769273469 Chr.10: 5887883-5887883 on Build GRCh38
rs1013543175 Chr.10: 5887780-5887780 on Build GRCh38
rs752666503 Chr.10: 5887946-5887946 on Build GRCh38
rs746626915 Chr.10: 5887847-5887847 on Build GRCh38
rs749667618 Chr.10: 5887992-5887992 on Build GRCh38
rs868520915 Chr.10: 5888145-5888145 on Build GRCh38
rs760681878 Chr.10: 5888013-5888013 on Build GRCh38
rs761953271 Chr.10: 5887890-5887890 on Build GRCh38
rs759292431 Chr.10: 5888017-5888017 on Build GRCh38
rs950875282 Chr.10: 5887796-5887796 on Build GRCh38
rs775065911 Chr.10: 5887888-5887888 on Build GRCh38
rs772571757 Chr.10: 5888076-5888076 on Build GRCh38
rs552596453 Chr.10: 5887831-5887832 on Build GRCh38
rs775209746 Chr.10: 5888114-5888114 on Build GRCh38
rs1025413441 Chr.10: 5888154-5888154 on Build GRCh38
rs547513286 Chr.10: 5887819-5887819 on Build GRCh38
rs148866244 Chr.10: 5887917-5887917 on Build GRCh38
rs200868116 Chr.10: 5888037-5888037 on Build GRCh38
rs1009058477 Chr.10: 5888072-5888072 on Build GRCh38
rs756879380 Chr.10: 5887968-5887968 on Build GRCh38
rs373191798 Chr.10: 5888097-5888097 on Build GRCh38
rs777300653 Chr.10: 5887953-5887953 on Build GRCh38
rs116747539 Chr.10: 5888241-5888241 on Build GRCh38
rs766608301 Chr.10: 5888014-5888014 on Build GRCh38
rs776808027 Chr.10: 5888087-5888087 on Build GRCh38
rs112121948 Chr.10: 5887924-5887924 on Build GRCh38
rs1054171104 Chr.10: 5887925-5887925 on Build GRCh38
rs779849725 Chr.10: 5887817-5887817 on Build GRCh38
rs770952763 Chr.10: 5888085-5888085 on Build GRCh38
rs767646701 Chr.10: 5887890-5887890 on Build GRCh38
rs763954169 Chr.10: 5888050-5888050 on Build GRCh38
rs778832643 Chr.10: 5887823-5887823 on Build GRCh38
rs773097564 Chr.10: 5887960-5887960 on Build GRCh38
rs763426235 Chr.10: 5887879-5887879 on Build GRCh38
rs767849040 Chr.10: 5887807-5887807 on Build GRCh38
rs370762685 Chr.10: 5887812-5887812 on Build GRCh38
rs780866869 Chr.10: 5888056-5888056 on Build GRCh38
rs745566361 Chr.10: 5888057-5888057 on Build GRCh38
rs2296136 Chr.10: 5887999-5887999 on Build GRCh38
rs770523835 Chr.10: 5887850-5887850 on Build GRCh38
rs777256459 Chr.10: 5887839-5887839 on Build GRCh38
rs774142230 Chr.10: 5888063-5888063 on Build GRCh38
rs537519126 Chr.10: 5888000-5888000 on Build GRCh38
rs74341195 Chr.10: 5887820-5887820 on Build GRCh38
rs370457689 Chr.10: 5888019-5888019 on Build GRCh38
rs772722804 Chr.10: 5887811-5887811 on Build GRCh38
rs745925041 Chr.10: 5887904-5887904 on Build GRCh38
rs1024253258 Chr.10: 5888185-5888185 on Build GRCh38
rs750489984 Chr.10: 5887897-5887897 on Build GRCh38
rs754822239 Chr.10: 5887922-5887922 on Build GRCh38
rs761010634 Chr.10: 5888116-5888116 on Build GRCh38
rs771633213 Chr.10: 5887837-5887837 on Build GRCh38
rs747641022 Chr.10: 5887834-5887834 on Build GRCh38
rs879359950 Chr.10: 5887852-5887852 on Build GRCh38
rs548724768 Chr.10: 5887937-5887937 on Build GRCh38
rs758358746 Chr.10: 5887952-5887952 on Build GRCh38
rs150555361 Chr.10: 5888078-5888078 on Build GRCh38
rs761937582 Chr.10: 5887879-5887882 on Build GRCh38
rs543841179 Chr.10: 5887835-5887835 on Build GRCh38
rs553492120 Chr.10: 5888069-5888069 on Build GRCh38
rs973464547 Chr.10: 5887782-5887782 on Build GRCh38
rs527592270 Chr.10: 5887893-5887893 on Build GRCh38
rs555175398 Chr.10: 5888220-5888220 on Build GRCh38
rs773274929 Chr.10: 5888010-5888010 on Build GRCh38
rs1012910497 Chr.10: 5888184-5888184 on Build GRCh38
rs748525342 Chr.10: 5888070-5888070 on Build GRCh38
rs538625336 Chr.10: 5888058-5888058 on Build GRCh38
rs377041437 Chr.10: 5888054-5888054 on Build GRCh38
rs776872410 Chr.10: 5888148-5888148 on Build GRCh38
rs377323749 Chr.10: 5887988-5887988 on Build GRCh38
rs772486885 Chr.10: 5888001-5888001 on Build GRCh38
rs747105504 Chr.10: 5888082-5888082 on Build GRCh38
rs920947153 Chr.10: 5887938-5887938 on Build GRCh38
rs752525069 Chr.10: 5888027-5888027 on Build GRCh38
rs377224855 Chr.10: 5887962-5887962 on Build GRCh38
rs769183503 Chr.10: 5887995-5887995 on Build GRCh38
rs776902514 Chr.10: 5888015-5888015 on Build GRCh38
rs886463920 Chr.10: 5888177-5888177 on Build GRCh38

View Full Product Details