Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 11951980-11952458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UPF2
Entrez Gene ID
26019
Gene Name
UPF2 regulator of nonsense transcripts homolog (yeast)
Gene Aliases
HUPF2, RENT2, smg-3
UniGene
Hs.370689
Gene Chromosome Location
Chr.10: 11920022-12043170 on Build GRCh38
Transcripts
NM_015542.3 NM_080599.2 XM_017016034.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4842843 c.2941G>C p.E981Q Chr.10: 11952159-11952159 on Build GRCh38
7076070 c.2870A>G p.K957R Chr.10: 11952230-11952230 on Build GRCh38
915278 c.2994G>A p.R998R Chr.10: 11952106-11952106 on Build GRCh38
1296983 c.2980G>A p.E994K Chr.10: 11952120-11952120 on Build GRCh38
7076662 c.2911G>C p.D971H Chr.10: 11952189-11952189 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775577196 Chr.10: 11952297-11952297 on Build GRCh38
rs113708407 Chr.10: 11952414-11952414 on Build GRCh38
rs112464709 Chr.10: 11952037-11952037 on Build GRCh38
rs138092895 Chr.10: 11952304-11952304 on Build GRCh38
rs756847596 Chr.10: 11952204-11952204 on Build GRCh38
rs1012272846 Chr.10: 11952314-11952314 on Build GRCh38
rs754121348 Chr.10: 11952206-11952206 on Build GRCh38
rs149547577 Chr.10: 11952353-11952353 on Build GRCh38
rs928434494 Chr.10: 11952316-11952316 on Build GRCh38
rs201617721 Chr.10: 11952280-11952280 on Build GRCh38
rs756508285 Chr.10: 11952057-11952057 on Build GRCh38
rs368084413 Chr.10: 11952285-11952285 on Build GRCh38
rs748595608 Chr.10: 11952267-11952267 on Build GRCh38
rs377274977 Chr.10: 11952102-11952102 on Build GRCh38
rs538307828 Chr.10: 11952416-11952416 on Build GRCh38
rs775522132 Chr.10: 11952172-11952172 on Build GRCh38
rs958714656 Chr.10: 11952328-11952328 on Build GRCh38
rs903395500 Chr.10: 11952031-11952031 on Build GRCh38
rs747612985 Chr.10: 11952133-11952133 on Build GRCh38
rs923838977 Chr.10: 11952279-11952279 on Build GRCh38
rs372581487 Chr.10: 11952086-11952086 on Build GRCh38
rs763632387 Chr.10: 11952192-11952192 on Build GRCh38
rs953604165 Chr.10: 11952434-11952434 on Build GRCh38
rs529447589 Chr.10: 11952309-11952309 on Build GRCh38
rs140801703 Chr.10: 11952082-11952082 on Build GRCh38
rs779417121 Chr.10: 11952244-11952244 on Build GRCh38
rs751116806 Chr.10: 11952038-11952038 on Build GRCh38
rs530494975 Chr.10: 11952142-11952142 on Build GRCh38
rs9663803 Chr.10: 11952290-11952290 on Build GRCh38
rs879580732 Chr.10: 11952319-11952319 on Build GRCh38
rs1007528906 Chr.10: 11952132-11952132 on Build GRCh38
rs973719695 Chr.10: 11952039-11952039 on Build GRCh38
rs755295504 Chr.10: 11952243-11952243 on Build GRCh38
rs531074827 Chr.10: 11952382-11952382 on Build GRCh38
rs1042525806 Chr.10: 11952166-11952166 on Build GRCh38
rs769657898 Chr.10: 11952294-11952294 on Build GRCh38
rs773493248 Chr.10: 11952112-11952112 on Build GRCh38
rs765300768 Chr.10: 11952157-11952157 on Build GRCh38
rs933531560 Chr.10: 11952412-11952412 on Build GRCh38
rs188546986 Chr.10: 11952362-11952362 on Build GRCh38
rs151248008 Chr.10: 11952200-11952200 on Build GRCh38
rs760519722 Chr.10: 11952432-11952432 on Build GRCh38
rs747130760 Chr.10: 11952134-11952134 on Build GRCh38
rs770960132 Chr.10: 11952276-11952276 on Build GRCh38
rs771032762 Chr.10: 11952288-11952288 on Build GRCh38
rs772088954 Chr.10: 11952110-11952110 on Build GRCh38
rs1046391079 Chr.10: 11952327-11952327 on Build GRCh38
rs766816918 Chr.10: 11952205-11952205 on Build GRCh38
rs144752256 Chr.10: 11952136-11952136 on Build GRCh38
rs563701298 Chr.10: 11952283-11952283 on Build GRCh38
rs762568543 Chr.10: 11952185-11952185 on Build GRCh38
rs759591095 Chr.10: 11952140-11952140 on Build GRCh38
rs963601800 Chr.10: 11952407-11952407 on Build GRCh38
rs1015419153 Chr.10: 11952371-11952371 on Build GRCh38
rs561425058 Chr.10: 11952135-11952135 on Build GRCh38
rs963871816 Chr.10: 11952016-11952016 on Build GRCh38
rs748256304 Chr.10: 11952103-11952103 on Build GRCh38
rs906617857 Chr.10: 11952339-11952339 on Build GRCh38
rs757946305 Chr.10: 11952030-11952030 on Build GRCh38
rs751294385 Chr.10: 11952202-11952202 on Build GRCh38
rs778426640 Chr.10: 11952100-11952100 on Build GRCh38
rs1041991274 Chr.10: 11952378-11952378 on Build GRCh38

View Full Product Details