Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 27028804-27029349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD26
Entrez Gene ID
22852
Gene Name
ankyrin repeat domain 26
Gene Aliases
THC2, bA145E8.1
UniGene
Hs.361041
Gene Chromosome Location
Chr.10: 26938195-27100498 on Build GRCh38
Transcripts
NM_001256053.1 NM_014915.2 XM_006717423.2 XM_006717425.3 XM_011519416.2 XM_017015928.1 XM_017015929.1 XM_017015930.1 XM_017015931.1 XM_017015932.1 XM_017015933.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6600509 c.3969G>A p.K1323K Chr.10: 27028903-27028903 on Build GRCh38
6600512 c.3970C>A p.Q1324K Chr.10: 27028902-27028902 on Build GRCh38
3997907 c.2643+3A>G p.? Chr.10: 27028849-27028849 on Build GRCh38
2134254 c.3817G>T p.A1273S Chr.10: 27029347-27029347 on Build GRCh38
3790706 c.2510C>G p.A837G Chr.10: 27029325-27029325 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750332409 Chr.10: 27028835-27028835 on Build GRCh38
rs926937632 Chr.10: 27028952-27028952 on Build GRCh38
rs535689758 Chr.10: 27029029-27029029 on Build GRCh38
rs10829163 Chr.10: 27028911-27028911 on Build GRCh38
rs766635392 Chr.10: 27028918-27028918 on Build GRCh38
rs780254821 Chr.10: 27028863-27028863 on Build GRCh38
rs577986400 Chr.10: 27029307-27029307 on Build GRCh38
rs943490446 Chr.10: 27029070-27029070 on Build GRCh38
rs1042989576 Chr.10: 27029049-27029049 on Build GRCh38
rs570137368 Chr.10: 27028986-27028986 on Build GRCh38
rs890436299 Chr.10: 27029082-27029082 on Build GRCh38
rs886046945 Chr.10: 27028884-27028884 on Build GRCh38
rs959708091 Chr.10: 27029271-27029271 on Build GRCh38
rs962215297 Chr.10: 27028848-27028848 on Build GRCh38
rs1043006435 Chr.10: 27029137-27029137 on Build GRCh38
rs749783019 Chr.10: 27028963-27028963 on Build GRCh38
rs771469274 Chr.10: 27028973-27028973 on Build GRCh38
rs959105865 Chr.10: 27029156-27029156 on Build GRCh38
rs201679608 Chr.10: 27028923-27028923 on Build GRCh38
rs775681582 Chr.10: 27029103-27029103 on Build GRCh38
rs554849202 Chr.10: 27028965-27028965 on Build GRCh38
rs755241312 Chr.10: 27028885-27028885 on Build GRCh38
rs748326243 Chr.10: 27028914-27028914 on Build GRCh38
rs750231011 Chr.10: 27029246-27029246 on Build GRCh38
rs762948559 Chr.10: 27029255-27029255 on Build GRCh38
rs200371774 Chr.10: 27029305-27029305 on Build GRCh38
rs572158989 Chr.10: 27029263-27029263 on Build GRCh38
rs909266110 Chr.10: 27029042-27029042 on Build GRCh38
rs1032522834 Chr.10: 27029154-27029154 on Build GRCh38
rs12268608 Chr.10: 27028944-27028944 on Build GRCh38
rs776277761 Chr.10: 27028991-27028991 on Build GRCh38
rs751725155 Chr.10: 27028865-27028865 on Build GRCh38
rs969628710 Chr.10: 27029226-27029226 on Build GRCh38
rs777777528 Chr.10: 27029141-27029141 on Build GRCh38
rs1025941982 Chr.10: 27029278-27029278 on Build GRCh38
rs754101571 Chr.10: 27029252-27029254 on Build GRCh38
rs766488085 Chr.10: 27029262-27029262 on Build GRCh38
rs72807627 Chr.10: 27028849-27028849 on Build GRCh38
rs763125135 Chr.10: 27029302-27029302 on Build GRCh38
rs756341370 Chr.10: 27029296-27029296 on Build GRCh38
rs370733209 Chr.10: 27029239-27029239 on Build GRCh38
rs368839645 Chr.10: 27029163-27029163 on Build GRCh38
rs765030276 Chr.10: 27029244-27029244 on Build GRCh38
rs945863720 Chr.10: 27029047-27029047 on Build GRCh38
rs771416156 Chr.10: 27029314-27029314 on Build GRCh38
rs778023428 Chr.10: 27028930-27028930 on Build GRCh38
rs748612731 Chr.10: 27029079-27029079 on Build GRCh38
rs931861170 Chr.10: 27029071-27029071 on Build GRCh38
rs185664036 Chr.10: 27029267-27029267 on Build GRCh38
rs760842664 Chr.10: 27028839-27028840 on Build GRCh38
rs755045164 Chr.10: 27029265-27029265 on Build GRCh38
rs374299408 Chr.10: 27028857-27028857 on Build GRCh38
rs774978340 Chr.10: 27028977-27028977 on Build GRCh38
rs142617940 Chr.10: 27029142-27029142 on Build GRCh38
rs752918983 Chr.10: 27029268-27029268 on Build GRCh38
rs781552855 Chr.10: 27029041-27029041 on Build GRCh38
rs1036432849 Chr.10: 27029132-27029132 on Build GRCh38
rs959115693 Chr.10: 27028842-27028842 on Build GRCh38
rs925869906 Chr.10: 27029056-27029056 on Build GRCh38
rs760234895 Chr.10: 27028980-27028980 on Build GRCh38
rs960066838 Chr.10: 27028979-27028979 on Build GRCh38
rs1027894319 Chr.10: 27028878-27028878 on Build GRCh38

View Full Product Details