Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73647331-73647826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNPO2L
Entrez Gene ID
79933
Gene Name
synaptopodin 2 like
Gene Aliases
-
UniGene
Hs.645273
Gene Chromosome Location
Chr.10: 73644881-73656105 on Build GRCh38
Transcripts
NM_001114133.2 NM_024875.4 XM_005270158.2 XM_005270159.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4731671 c.2139C>T p.T713T Chr.10: 73647513-73647513 on Build GRCh38
7269016 c.1552G>T p.A518S Chr.10: 73647428-73647428 on Build GRCh38
5969899 c.1981G>A p.E661K Chr.10: 73647671-73647671 on Build GRCh38
3747861 c.1476T>C p.P492P Chr.10: 73647504-73647504 on Build GRCh38
6795319 c.1844C>A p.S615Y Chr.10: 73647808-73647808 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200312652 Chr.10: 73647723-73647723 on Build GRCh38
rs1012102324 Chr.10: 73647712-73647712 on Build GRCh38
rs577750788 Chr.10: 73647358-73647358 on Build GRCh38
rs3812629 Chr.10: 73647532-73647532 on Build GRCh38
rs750008622 Chr.10: 73647593-73647593 on Build GRCh38
rs775794385 Chr.10: 73647552-73647552 on Build GRCh38
rs780472661 Chr.10: 73647488-73647489 on Build GRCh38
rs367989404 Chr.10: 73647596-73647596 on Build GRCh38
rs748622782 Chr.10: 73647744-73647744 on Build GRCh38
rs922037164 Chr.10: 73647377-73647377 on Build GRCh38
rs759446036 Chr.10: 73647782-73647782 on Build GRCh38
rs950862598 Chr.10: 73647504-73647504 on Build GRCh38
rs746109200 Chr.10: 73647633-73647633 on Build GRCh38
rs755526777 Chr.10: 73647397-73647397 on Build GRCh38
rs768127784 Chr.10: 73647483-73647483 on Build GRCh38
rs954745195 Chr.10: 73647717-73647717 on Build GRCh38
rs768606281 Chr.10: 73647531-73647532 on Build GRCh38
rs113374956 Chr.10: 73647729-73647729 on Build GRCh38
rs751344464 Chr.10: 73647602-73647602 on Build GRCh38
rs146498004 Chr.10: 73647666-73647666 on Build GRCh38
rs61737176 Chr.10: 73647383-73647383 on Build GRCh38
rs781274983 Chr.10: 73647616-73647616 on Build GRCh38
rs756499481 Chr.10: 73647462-73647462 on Build GRCh38
rs745581919 Chr.10: 73647451-73647451 on Build GRCh38
rs748289455 Chr.10: 73647532-73647533 on Build GRCh38
rs768668437 Chr.10: 73647533-73647533 on Build GRCh38
rs537837245 Chr.10: 73647719-73647719 on Build GRCh38
rs781510797 Chr.10: 73647379-73647380 on Build GRCh38
rs773594173 Chr.10: 73647744-73647749 on Build GRCh38
rs775835367 Chr.10: 73647653-73647653 on Build GRCh38
rs780246399 Chr.10: 73647393-73647393 on Build GRCh38
rs779237330 Chr.10: 73647632-73647632 on Build GRCh38
rs748283757 Chr.10: 73647466-73647466 on Build GRCh38
rs978401458 Chr.10: 73647745-73647745 on Build GRCh38
rs767848224 Chr.10: 73647413-73647413 on Build GRCh38
rs766065163 Chr.10: 73647693-73647693 on Build GRCh38
rs533607226 Chr.10: 73647496-73647496 on Build GRCh38
rs773543256 Chr.10: 73647379-73647379 on Build GRCh38
rs772295733 Chr.10: 73647539-73647539 on Build GRCh38
rs764517172 Chr.10: 73647557-73647557 on Build GRCh38
rs866590356 Chr.10: 73647614-73647614 on Build GRCh38
rs778016253 Chr.10: 73647534-73647534 on Build GRCh38
rs778426046 Chr.10: 73647524-73647524 on Build GRCh38
rs570603708 Chr.10: 73647669-73647669 on Build GRCh38
rs766197020 Chr.10: 73647771-73647771 on Build GRCh38
rs778882286 Chr.10: 73647715-73647715 on Build GRCh38
rs371822690 Chr.10: 73647658-73647658 on Build GRCh38
rs987505953 Chr.10: 73647404-73647404 on Build GRCh38
rs777096663 Chr.10: 73647662-73647662 on Build GRCh38
rs150403805 Chr.10: 73647783-73647783 on Build GRCh38
rs867479646 Chr.10: 73647514-73647514 on Build GRCh38
rs370617240 Chr.10: 73647457-73647457 on Build GRCh38
rs145644342 Chr.10: 73647454-73647454 on Build GRCh38
rs150532364 Chr.10: 73647366-73647366 on Build GRCh38
rs1035304636 Chr.10: 73647416-73647416 on Build GRCh38
rs773383473 Chr.10: 73647472-73647472 on Build GRCh38
rs747421994 Chr.10: 73647725-73647725 on Build GRCh38
rs760509432 Chr.10: 73647794-73647795 on Build GRCh38
rs377467415 Chr.10: 73647438-73647438 on Build GRCh38
rs772473138 Chr.10: 73647723-73647725 on Build GRCh38
rs769961395 Chr.10: 73647469-73647469 on Build GRCh38
rs758027011 Chr.10: 73647594-73647594 on Build GRCh38
rs749437804 Chr.10: 73647618-73647618 on Build GRCh38
rs753412836 Chr.10: 73647560-73647560 on Build GRCh38
rs758613243 Chr.10: 73647721-73647721 on Build GRCh38
rs140990304 Chr.10: 73647703-73647703 on Build GRCh38
rs763294332 Chr.10: 73647475-73647475 on Build GRCh38
rs1016528880 Chr.10: 73647677-73647677 on Build GRCh38
rs372058702 Chr.10: 73647773-73647773 on Build GRCh38
rs375335895 Chr.10: 73647781-73647781 on Build GRCh38
rs140374242 Chr.10: 73647482-73647482 on Build GRCh38
rs761182981 Chr.10: 73647554-73647554 on Build GRCh38
rs756007816 Chr.10: 73647805-73647805 on Build GRCh38
rs755607793 Chr.10: 73647423-73647423 on Build GRCh38
rs774786890 Chr.10: 73647480-73647480 on Build GRCh38
rs575283468 Chr.10: 73647468-73647468 on Build GRCh38
rs754810875 Chr.10: 73647699-73647699 on Build GRCh38
rs201306811 Chr.10: 73647375-73647375 on Build GRCh38
rs936548298 Chr.10: 73647506-73647506 on Build GRCh38
rs748910220 Chr.10: 73647428-73647428 on Build GRCh38
rs772277925 Chr.10: 73647652-73647652 on Build GRCh38
rs749813969 Chr.10: 73647530-73647531 on Build GRCh38
rs886108341 Chr.10: 73647634-73647634 on Build GRCh38
rs552756869 Chr.10: 73647537-73647537 on Build GRCh38
rs536226857 Chr.10: 73647479-73647479 on Build GRCh38
rs939132708 Chr.10: 73647610-73647610 on Build GRCh38
rs770893636 Chr.10: 73647626-73647626 on Build GRCh38
rs777453697 Chr.10: 73647426-73647426 on Build GRCh38
rs949047754 Chr.10: 73647562-73647562 on Build GRCh38
rs776937165 Chr.10: 73647736-73647736 on Build GRCh38
rs143723429 Chr.10: 73647763-73647763 on Build GRCh38
rs756852529 Chr.10: 73647432-73647432 on Build GRCh38
rs774125238 Chr.10: 73647779-73647779 on Build GRCh38
rs753285569 Chr.10: 73647511-73647511 on Build GRCh38
rs769208374 Chr.10: 73647654-73647654 on Build GRCh38
rs771255679 Chr.10: 73647536-73647536 on Build GRCh38
rs202160216 Chr.10: 73647402-73647402 on Build GRCh38
rs781327981 Chr.10: 73647702-73647702 on Build GRCh38
rs774557864 Chr.10: 73647532-73647532 on Build GRCh38
rs1042327624 Chr.10: 73647449-73647449 on Build GRCh38
rs778780364 Chr.10: 73647448-73647448 on Build GRCh38
rs899314285 Chr.10: 73647691-73647691 on Build GRCh38
rs987652062 Chr.10: 73647509-73647509 on Build GRCh38
rs767453923 Chr.10: 73647789-73647789 on Build GRCh38
rs148105482 Chr.10: 73647790-73647790 on Build GRCh38
rs199811689 Chr.10: 73647359-73647359 on Build GRCh38
rs770274901 Chr.10: 73647762-73647762 on Build GRCh38
rs762529167 Chr.10: 73647678-73647678 on Build GRCh38
rs1046012505 Chr.10: 73647617-73647617 on Build GRCh38
rs759995784 Chr.10: 73647538-73647538 on Build GRCh38
rs758102265 Chr.10: 73647530-73647530 on Build GRCh38
rs916422977 Chr.10: 73647764-73647764 on Build GRCh38
rs745729678 Chr.10: 73647527-73647527 on Build GRCh38
rs761214859 Chr.10: 73647578-73647578 on Build GRCh38
rs925249140 Chr.10: 73647756-73647756 on Build GRCh38
rs766884023 Chr.10: 73647387-73647387 on Build GRCh38
rs149659353 Chr.10: 73647418-73647418 on Build GRCh38
rs1029077322 Chr.10: 73647704-73647704 on Build GRCh38
rs371474010 Chr.10: 73647350-73647350 on Build GRCh38
rs769070146 Chr.10: 73647523-73647523 on Build GRCh38
rs753124975 Chr.10: 73647485-73647485 on Build GRCh38
rs746181467 Chr.10: 73647385-73647390 on Build GRCh38
rs764985644 Chr.10: 73647580-73647580 on Build GRCh38
rs749896968 Chr.10: 73647673-73647673 on Build GRCh38
rs376413443 Chr.10: 73647698-73647698 on Build GRCh38

View Full Product Details