Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 84139341-84139849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHITM
Entrez Gene ID
27069
Gene Name
growth hormone inducible transmembrane protein
Gene Aliases
DERP2, HSPC282, MICS1, My021, PTD010, TMBIM5
UniGene
Hs.352656
Gene Chromosome Location
Chr.10: 84139429-84153555 on Build GRCh38
Transcripts
NM_014394.2

SNP Detail

SNP ID Chromosome Location
rs890814125 Chr.10: 84139383-84139383 on Build GRCh38
rs1040731981 Chr.10: 84139706-84139706 on Build GRCh38
rs778960677 Chr.10: 84139441-84139441 on Build GRCh38
rs905716644 Chr.10: 84139570-84139570 on Build GRCh38
rs980456810 Chr.10: 84139461-84139461 on Build GRCh38
rs1007878154 Chr.10: 84139387-84139387 on Build GRCh38
rs898355648 Chr.10: 84139493-84139493 on Build GRCh38
rs969913030 Chr.10: 84139460-84139460 on Build GRCh38
rs1049905809 Chr.10: 84139693-84139693 on Build GRCh38
rs919723933 Chr.10: 84139666-84139666 on Build GRCh38
rs7923369 Chr.10: 84139495-84139495 on Build GRCh38
rs763265490 Chr.10: 84139599-84139599 on Build GRCh38
rs979955002 Chr.10: 84139748-84139748 on Build GRCh38
rs866802812 Chr.10: 84139803-84139803 on Build GRCh38
rs943818924 Chr.10: 84139700-84139700 on Build GRCh38
rs1026726505 Chr.10: 84139569-84139569 on Build GRCh38
rs960713707 Chr.10: 84139476-84139476 on Build GRCh38
rs1020952481 Chr.10: 84139720-84139720 on Build GRCh38
rs918288989 Chr.10: 84139823-84139823 on Build GRCh38
rs1036029969 Chr.10: 84139646-84139646 on Build GRCh38
rs180964266 Chr.10: 84139561-84139561 on Build GRCh38
rs538512357 Chr.10: 84139555-84139555 on Build GRCh38
rs766770969 Chr.10: 84139769-84139769 on Build GRCh38
rs930948407 Chr.10: 84139824-84139824 on Build GRCh38
rs972650799 Chr.10: 84139639-84139639 on Build GRCh38
rs959599389 Chr.10: 84139590-84139590 on Build GRCh38
rs548024401 Chr.10: 84139420-84139420 on Build GRCh38
rs889426046 Chr.10: 84139528-84139528 on Build GRCh38
rs536422315 Chr.10: 84139475-84139475 on Build GRCh38
rs761956292 Chr.10: 84139518-84139518 on Build GRCh38
rs561476414 Chr.10: 84139732-84139732 on Build GRCh38
rs992124939 Chr.10: 84139711-84139711 on Build GRCh38
rs1021168815 Chr.10: 84139369-84139369 on Build GRCh38
rs968536595 Chr.10: 84139403-84139403 on Build GRCh38
rs1009809631 Chr.10: 84139768-84139768 on Build GRCh38
rs565578673 Chr.10: 84139401-84139401 on Build GRCh38
rs938041171 Chr.10: 84139409-84139409 on Build GRCh38
rs568369732 Chr.10: 84139478-84139478 on Build GRCh38
rs1034147334 Chr.10: 84139809-84139809 on Build GRCh38
rs149473490 Chr.10: 84139789-84139789 on Build GRCh38
rs1050452828 Chr.10: 84139537-84139537 on Build GRCh38
rs1050004021 Chr.10: 84139365-84139365 on Build GRCh38
rs932821133 Chr.10: 84139514-84139514 on Build GRCh38
rs979486194 Chr.10: 84139496-84139496 on Build GRCh38
rs959479495 Chr.10: 84139812-84139812 on Build GRCh38
rs751916536 Chr.10: 84139559-84139559 on Build GRCh38
rs902087647 Chr.10: 84139543-84139543 on Build GRCh38
rs371944147 Chr.10: 84139547-84139547 on Build GRCh38
rs115032989 Chr.10: 84139408-84139408 on Build GRCh38
rs910981525 Chr.10: 84139698-84139698 on Build GRCh38
rs1042675655 Chr.10: 84139774-84139774 on Build GRCh38
rs995346293 Chr.10: 84139424-84139424 on Build GRCh38
rs912811666 Chr.10: 84139799-84139799 on Build GRCh38
rs943695956 Chr.10: 84139546-84139546 on Build GRCh38
rs944232445 Chr.10: 84139802-84139802 on Build GRCh38
rs1136944 Chr.10: 84139591-84139591 on Build GRCh38
rs925371555 Chr.10: 84139760-84139760 on Build GRCh38
rs1019173564 Chr.10: 84139542-84139542 on Build GRCh38
rs566234018 Chr.10: 84139438-84139438 on Build GRCh38
rs576610432 Chr.10: 84139737-84139737 on Build GRCh38
rs959914872 Chr.10: 84139679-84139679 on Build GRCh38
rs537494687 Chr.10: 84139487-84139487 on Build GRCh38
rs567240160 Chr.10: 84139484-84139484 on Build GRCh38
rs1013646343 Chr.10: 84139821-84139821 on Build GRCh38
rs891352454 Chr.10: 84139540-84139540 on Build GRCh38
rs1033790025 Chr.10: 84139575-84139575 on Build GRCh38
rs542443848 Chr.10: 84139708-84139708 on Build GRCh38
rs896909674 Chr.10: 84139551-84139551 on Build GRCh38
rs3829861 Chr.10: 84139503-84139503 on Build GRCh38

View Full Product Details