Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102743888-102744392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WBP1L
Entrez Gene ID
54838
Gene Name
WW domain binding protein 1-like
Gene Aliases
C10orf26, OPA1L, OPAL1
UniGene
Hs.500897
Gene Chromosome Location
Chr.10: 102742708-102816267 on Build GRCh38
Transcripts
NM_001083913.1 XM_017016360.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5765276 c.15G>C p.R5S Chr.10: 102744068-102744068 on Build GRCh38
4144316 c.54G>C p.L18L Chr.10: 102744107-102744107 on Build GRCh38
5497986 c.27C>T p.G9G Chr.10: 102744080-102744080 on Build GRCh38
4428380 c.40_42delCTC p.L15delL Chr.10: 102744093-102744095 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs180771793 Chr.10: 102744068-102744068 on Build GRCh38
rs200862123 Chr.10: 102744139-102744139 on Build GRCh38
rs968302231 Chr.10: 102744023-102744023 on Build GRCh38
rs765911465 Chr.10: 102744166-102744166 on Build GRCh38
rs1035554245 Chr.10: 102743971-102743971 on Build GRCh38
rs202146271 Chr.10: 102743997-102743998 on Build GRCh38
rs760241052 Chr.10: 102744030-102744030 on Build GRCh38
rs574482733 Chr.10: 102743949-102743949 on Build GRCh38
rs771833723 Chr.10: 102743970-102743970 on Build GRCh38
rs564326503 Chr.10: 102744121-102744121 on Build GRCh38
rs376002870 Chr.10: 102744031-102744031 on Build GRCh38
rs1012423324 Chr.10: 102743979-102743979 on Build GRCh38
rs896902289 Chr.10: 102744002-102744002 on Build GRCh38
rs979173132 Chr.10: 102744140-102744140 on Build GRCh38
rs567035376 Chr.10: 102744338-102744338 on Build GRCh38
rs771734577 Chr.10: 102744033-102744038 on Build GRCh38
rs999776999 Chr.10: 102744306-102744306 on Build GRCh38
rs368414528 Chr.10: 102744039-102744039 on Build GRCh38
rs758588293 Chr.10: 102744069-102744069 on Build GRCh38
rs534531041 Chr.10: 102744351-102744351 on Build GRCh38
rs754734159 Chr.10: 102744051-102744051 on Build GRCh38
rs773065189 Chr.10: 102743980-102743980 on Build GRCh38
rs1038455352 Chr.10: 102744369-102744369 on Build GRCh38
rs549910802 Chr.10: 102744207-102744207 on Build GRCh38
rs569897242 Chr.10: 102743996-102743997 on Build GRCh38
rs748061310 Chr.10: 102744032-102744033 on Build GRCh38
rs897177043 Chr.10: 102743956-102743956 on Build GRCh38
rs904043530 Chr.10: 102744297-102744297 on Build GRCh38
rs1016579580 Chr.10: 102743998-102744000 on Build GRCh38
rs763923912 Chr.10: 102744159-102744159 on Build GRCh38
rs939383164 Chr.10: 102744206-102744206 on Build GRCh38
rs746338103 Chr.10: 102744116-102744116 on Build GRCh38
rs1031703367 Chr.10: 102744354-102744354 on Build GRCh38
rs747073710 Chr.10: 102744078-102744078 on Build GRCh38
rs527620033 Chr.10: 102744112-102744112 on Build GRCh38
rs1006661727 Chr.10: 102744081-102744081 on Build GRCh38
rs772904618 Chr.10: 102744152-102744152 on Build GRCh38
rs1037048314 Chr.10: 102743947-102743947 on Build GRCh38
rs374917827 Chr.10: 102744189-102744189 on Build GRCh38
rs978772701 Chr.10: 102743928-102743928 on Build GRCh38
rs114804158 Chr.10: 102744370-102744370 on Build GRCh38
rs895281475 Chr.10: 102743977-102743977 on Build GRCh38
rs941584665 Chr.10: 102744344-102744344 on Build GRCh38
rs1053703871 Chr.10: 102743966-102743972 on Build GRCh38
rs189586966 Chr.10: 102743926-102743926 on Build GRCh38
rs545643421 Chr.10: 102744115-102744115 on Build GRCh38
rs978777005 Chr.10: 102744025-102744025 on Build GRCh38
rs775868740 Chr.10: 102744137-102744137 on Build GRCh38
rs112090972 Chr.10: 102744072-102744072 on Build GRCh38
rs1022456011 Chr.10: 102744273-102744273 on Build GRCh38
rs1022530701 Chr.10: 102744009-102744009 on Build GRCh38
rs549142102 Chr.10: 102744358-102744358 on Build GRCh38
rs943398489 Chr.10: 102743923-102743923 on Build GRCh38
rs73344501 Chr.10: 102744246-102744246 on Build GRCh38
rs775434681 Chr.10: 102743989-102743989 on Build GRCh38
rs770229144 Chr.10: 102744128-102744128 on Build GRCh38
rs529100477 Chr.10: 102744262-102744262 on Build GRCh38
rs771754799 Chr.10: 102744148-102744148 on Build GRCh38
rs934723022 Chr.10: 102743948-102743948 on Build GRCh38
rs888369522 Chr.10: 102743908-102743908 on Build GRCh38
rs906098724 Chr.10: 102743990-102743990 on Build GRCh38
rs186149703 Chr.10: 102744328-102744328 on Build GRCh38

View Full Product Details