Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 123164122-123164625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BUB3
Entrez Gene ID
9184
Gene Name
BUB3, mitotic checkpoint protein
Gene Aliases
BUB3L, hBUB3
UniGene
Hs.418533
Gene Chromosome Location
Chr.10: 123154244-123165370 on Build GRCh38
Transcripts
NM_001007793.2 NM_004725.3

SNP Detail

SNP ID Chromosome Location
rs1027001522 Chr.10: 123164537-123164537 on Build GRCh38
rs979091214 Chr.10: 123164364-123164365 on Build GRCh38
rs752784439 Chr.10: 123164589-123164589 on Build GRCh38
rs917353393 Chr.10: 123164472-123164473 on Build GRCh38
rs560338669 Chr.10: 123164177-123164177 on Build GRCh38
rs374121772 Chr.10: 123164303-123164303 on Build GRCh38
rs1046312635 Chr.10: 123164373-123164373 on Build GRCh38
rs879107527 Chr.10: 123164232-123164232 on Build GRCh38
rs978195158 Chr.10: 123164264-123164264 on Build GRCh38
rs572286922 Chr.10: 123164419-123164419 on Build GRCh38
rs777038313 Chr.10: 123164343-123164343 on Build GRCh38
rs894995005 Chr.10: 123164340-123164340 on Build GRCh38
rs954211929 Chr.10: 123164557-123164557 on Build GRCh38
rs996554911 Chr.10: 123164316-123164316 on Build GRCh38
rs576379747 Chr.10: 123164481-123164481 on Build GRCh38
rs567462989 Chr.10: 123164506-123164506 on Build GRCh38
rs527569482 Chr.10: 123164206-123164206 on Build GRCh38
rs936428447 Chr.10: 123164295-123164295 on Build GRCh38
rs943726135 Chr.10: 123164242-123164242 on Build GRCh38
rs1048461487 Chr.10: 123164566-123164566 on Build GRCh38
rs886626371 Chr.10: 123164169-123164169 on Build GRCh38
rs112003546 Chr.10: 123164498-123164498 on Build GRCh38
rs930871269 Chr.10: 123164542-123164542 on Build GRCh38
rs1053521535 Chr.10: 123164319-123164319 on Build GRCh38
rs535625670 Chr.10: 123164258-123164258 on Build GRCh38
rs923627889 Chr.10: 123164274-123164274 on Build GRCh38
rs1019404271 Chr.10: 123164229-123164229 on Build GRCh38
rs965025176 Chr.10: 123164305-123164305 on Build GRCh38
rs186611999 Chr.10: 123164474-123164474 on Build GRCh38
rs562196354 Chr.10: 123164585-123164585 on Build GRCh38
rs1004797223 Chr.10: 123164473-123164473 on Build GRCh38
rs553983272 Chr.10: 123164398-123164398 on Build GRCh38
rs968632520 Chr.10: 123164349-123164349 on Build GRCh38
rs992910655 Chr.10: 123164430-123164430 on Build GRCh38
rs546078340 Chr.10: 123164454-123164454 on Build GRCh38
rs146851508 Chr.10: 123164586-123164586 on Build GRCh38
rs369149974 Chr.10: 123164394-123164394 on Build GRCh38
rs539329253 Chr.10: 123164198-123164198 on Build GRCh38
rs897137468 Chr.10: 123164505-123164505 on Build GRCh38
rs912764627 Chr.10: 123164213-123164213 on Build GRCh38

View Full Product Details