Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 124789849-124790372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL10
Entrez Gene ID
399818
Gene Name
methyltransferase like 10
Gene Aliases
C10orf138
UniGene
Hs.720819
Gene Chromosome Location
Chr.10: 124757831-124791941 on Build GRCh38
Transcripts
NM_001304467.1 NM_001304468.1 NM_212554.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6768704 c.149G>A p.R50Q Chr.10: 124790300-124790300 on Build GRCh38
6418639 c.128A>G p.E43G Chr.10: 124790321-124790321 on Build GRCh38
1188080 c.166G>C p.G56R Chr.10: 124790283-124790283 on Build GRCh38
7194055 c.163A>G p.T55A Chr.10: 124790286-124790286 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777634555 Chr.10: 124790266-124790266 on Build GRCh38
rs1009675519 Chr.10: 124790018-124790018 on Build GRCh38
rs766429010 Chr.10: 124790324-124790324 on Build GRCh38
rs149135673 Chr.10: 124790298-124790298 on Build GRCh38
rs948608365 Chr.10: 124790204-124790205 on Build GRCh38
rs780645336 Chr.10: 124789958-124789958 on Build GRCh38
rs201104535 Chr.10: 124790301-124790301 on Build GRCh38
rs995498747 Chr.10: 124790042-124790042 on Build GRCh38
rs183291422 Chr.10: 124790151-124790151 on Build GRCh38
rs373121441 Chr.10: 124790219-124790219 on Build GRCh38
rs756162216 Chr.10: 124790233-124790233 on Build GRCh38
rs926464152 Chr.10: 124790040-124790040 on Build GRCh38
rs531737814 Chr.10: 124789955-124789955 on Build GRCh38
rs575765585 Chr.10: 124790178-124790178 on Build GRCh38
rs188070895 Chr.10: 124790232-124790232 on Build GRCh38
rs543006628 Chr.10: 124789940-124789940 on Build GRCh38
rs978385200 Chr.10: 124790150-124790150 on Build GRCh38
rs918025554 Chr.10: 124790144-124790144 on Build GRCh38
rs895666552 Chr.10: 124789977-124789977 on Build GRCh38
rs955048337 Chr.10: 124790015-124790015 on Build GRCh38
rs1042987851 Chr.10: 124789896-124789896 on Build GRCh38
rs774973995 Chr.10: 124790173-124790173 on Build GRCh38
rs1028912072 Chr.10: 124790333-124790333 on Build GRCh38
rs909428501 Chr.10: 124790166-124790166 on Build GRCh38
rs528676733 Chr.10: 124789953-124789953 on Build GRCh38
rs568341885 Chr.10: 124790114-124790114 on Build GRCh38
rs556940311 Chr.10: 124789985-124789985 on Build GRCh38
rs767774469 Chr.10: 124790317-124790317 on Build GRCh38
rs768976483 Chr.10: 124790311-124790311 on Build GRCh38
rs745413860 Chr.10: 124790294-124790294 on Build GRCh38
rs749815979 Chr.10: 124790308-124790308 on Build GRCh38
rs773389248 Chr.10: 124790318-124790318 on Build GRCh38
rs774809527 Chr.10: 124790316-124790316 on Build GRCh38
rs1020172307 Chr.10: 124790116-124790116 on Build GRCh38
rs577031762 Chr.10: 124790037-124790037 on Build GRCh38
rs1050851915 Chr.10: 124790191-124790191 on Build GRCh38
rs554051621 Chr.10: 124790145-124790145 on Build GRCh38
rs760808969 Chr.10: 124790319-124790319 on Build GRCh38
rs545674494 Chr.10: 124790097-124790097 on Build GRCh38
rs765195194 Chr.10: 124790238-124790238 on Build GRCh38
rs756996545 Chr.10: 124790273-124790273 on Build GRCh38
rs1019888422 Chr.10: 124790313-124790313 on Build GRCh38
rs1017789684 Chr.10: 124789954-124789954 on Build GRCh38
rs929335504 Chr.10: 124789898-124789898 on Build GRCh38
rs896475766 Chr.10: 124789893-124789893 on Build GRCh38
rs547704214 Chr.10: 124789931-124789931 on Build GRCh38
rs559249051 Chr.10: 124790099-124790099 on Build GRCh38
rs975549316 Chr.10: 124790129-124790129 on Build GRCh38
rs867213140 Chr.10: 124790189-124790189 on Build GRCh38
rs551535348 Chr.10: 124790035-124790035 on Build GRCh38
rs780972142 Chr.10: 124790282-124790282 on Build GRCh38
rs529569298 Chr.10: 124789930-124789930 on Build GRCh38
rs1047965279 Chr.10: 124789935-124789935 on Build GRCh38
rs1056954688 Chr.10: 124789963-124789963 on Build GRCh38
rs890882821 Chr.10: 124790202-124790202 on Build GRCh38
rs968372478 Chr.10: 124790146-124790146 on Build GRCh38
rs397774728 Chr.10: 124789909-124789910 on Build GRCh38
rs758181762 Chr.10: 124790248-124790248 on Build GRCh38
rs543587252 Chr.10: 124790201-124790201 on Build GRCh38
rs1029866464 Chr.10: 124789987-124789987 on Build GRCh38
rs557557796 Chr.10: 124790174-124790174 on Build GRCh38
rs146879084 Chr.10: 124790330-124790330 on Build GRCh38
rs763892974 Chr.10: 124790341-124790341 on Build GRCh38
rs951590275 Chr.10: 124790076-124790076 on Build GRCh38
rs965591524 Chr.10: 124790123-124790123 on Build GRCh38
rs746744382 Chr.10: 124790270-124790270 on Build GRCh38
rs11443011 Chr.10: 124789898-124789899 on Build GRCh38
rs535720902 Chr.10: 124790128-124790128 on Build GRCh38
rs375205453 Chr.10: 124790241-124790241 on Build GRCh38
rs545830638 Chr.10: 124790160-124790160 on Build GRCh38
rs556434296 Chr.10: 124790100-124790100 on Build GRCh38
rs1004428519 Chr.10: 124790102-124790102 on Build GRCh38
rs917052621 Chr.10: 124790192-124790192 on Build GRCh38
rs1039338588 Chr.10: 124790182-124790182 on Build GRCh38
rs138061690 Chr.10: 124790300-124790300 on Build GRCh38
rs748317012 Chr.10: 124790183-124790183 on Build GRCh38
rs1027189048 Chr.10: 124790054-124790054 on Build GRCh38
rs1033594462 Chr.10: 124790347-124790347 on Build GRCh38
rs888041151 Chr.10: 124789947-124789947 on Build GRCh38
rs867073585 Chr.10: 124789960-124789960 on Build GRCh38
rs757606059 Chr.10: 124790039-124790039 on Build GRCh38
rs758395582 Chr.10: 124790338-124790338 on Build GRCh38

View Full Product Details