Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 124829308-124829821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM175B
Entrez Gene ID
23172
Gene Name
family with sequence similarity 175 member B
Gene Aliases
ABRO1, KIAA0157
UniGene
Hs.280695
Gene Chromosome Location
Chr.10: 124801785-124836670 on Build GRCh38
Transcripts
NM_032182.3 XM_011539554.1 XM_017015986.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
915828 c.597G>T p.K199N Chr.10: 124829411-124829411 on Build GRCh38
1703218 c.599A>G p.D200G Chr.10: 124829413-124829413 on Build GRCh38
1346584 c.580A>G p.T194A Chr.10: 124829394-124829394 on Build GRCh38
4890985 c.602G>C p.G201A Chr.10: 124829416-124829416 on Build GRCh38
5654171 c.606G>A p.V202V Chr.10: 124829420-124829420 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762829423 Chr.10: 124829726-124829726 on Build GRCh38
rs182602143 Chr.10: 124829717-124829717 on Build GRCh38
rs922217011 Chr.10: 124829478-124829478 on Build GRCh38
rs370225126 Chr.10: 124829346-124829346 on Build GRCh38
rs570754641 Chr.10: 124829626-124829626 on Build GRCh38
rs746846486 Chr.10: 124829374-124829374 on Build GRCh38
rs372075955 Chr.10: 124829509-124829509 on Build GRCh38
rs558712170 Chr.10: 124829334-124829334 on Build GRCh38
rs933437294 Chr.10: 124829484-124829484 on Build GRCh38
rs917975087 Chr.10: 124829414-124829414 on Build GRCh38
rs903517243 Chr.10: 124829533-124829533 on Build GRCh38
rs372299645 Chr.10: 124829359-124829359 on Build GRCh38
rs950577228 Chr.10: 124829663-124829663 on Build GRCh38
rs1013342351 Chr.10: 124829642-124829642 on Build GRCh38
rs1039685443 Chr.10: 124829470-124829470 on Build GRCh38
rs773545398 Chr.10: 124829581-124829581 on Build GRCh38
rs138412356 Chr.10: 124829438-124829438 on Build GRCh38
rs374496414 Chr.10: 124829347-124829347 on Build GRCh38
rs750779698 Chr.10: 124829526-124829526 on Build GRCh38
rs958817236 Chr.10: 124829780-124829780 on Build GRCh38
rs566202502 Chr.10: 124829436-124829436 on Build GRCh38
rs1002008649 Chr.10: 124829768-124829768 on Build GRCh38
rs576780341 Chr.10: 124829460-124829460 on Build GRCh38
rs757640919 Chr.10: 124829349-124829349 on Build GRCh38
rs991954605 Chr.10: 124829386-124829386 on Build GRCh38
rs963673863 Chr.10: 124829757-124829757 on Build GRCh38
rs959432465 Chr.10: 124829367-124829367 on Build GRCh38
rs776350312 Chr.10: 124829384-124829384 on Build GRCh38
rs890850633 Chr.10: 124829493-124829493 on Build GRCh38
rs2303611 Chr.10: 124829420-124829420 on Build GRCh38
rs756340492 Chr.10: 124829356-124829356 on Build GRCh38
rs1024748368 Chr.10: 124829662-124829662 on Build GRCh38
rs979741312 Chr.10: 124829360-124829360 on Build GRCh38
rs764917002 Chr.10: 124829631-124829631 on Build GRCh38
rs529719130 Chr.10: 124829339-124829339 on Build GRCh38
rs1046287169 Chr.10: 124829647-124829647 on Build GRCh38
rs369869632 Chr.10: 124829361-124829361 on Build GRCh38
rs764043262 Chr.10: 124829727-124829727 on Build GRCh38
rs1051952837 Chr.10: 124829485-124829485 on Build GRCh38
rs906372298 Chr.10: 124829667-124829667 on Build GRCh38
rs762495232 Chr.10: 124829437-124829437 on Build GRCh38
rs368544409 Chr.10: 124829351-124829351 on Build GRCh38
rs376317263 Chr.10: 124829396-124829396 on Build GRCh38
rs748196596 Chr.10: 124829521-124829522 on Build GRCh38
rs983496672 Chr.10: 124829718-124829718 on Build GRCh38
rs777730841 Chr.10: 124829373-124829373 on Build GRCh38
rs752718975 Chr.10: 124829679-124829679 on Build GRCh38
rs771948525 Chr.10: 124829369-124829369 on Build GRCh38
rs568350281 Chr.10: 124829773-124829773 on Build GRCh38
rs766353425 Chr.10: 124829646-124829646 on Build GRCh38
rs750691535 Chr.10: 124829353-124829353 on Build GRCh38
rs1042164431 Chr.10: 124829519-124829519 on Build GRCh38
rs41300560 Chr.10: 124829573-124829573 on Build GRCh38
rs910269694 Chr.10: 124829570-124829570 on Build GRCh38
rs368389959 Chr.10: 124829447-124829447 on Build GRCh38
rs538155615 Chr.10: 124829715-124829715 on Build GRCh38
rs1006302712 Chr.10: 124829790-124829790 on Build GRCh38
rs1053418273 Chr.10: 124829611-124829611 on Build GRCh38
rs778841253 Chr.10: 124829506-124829509 on Build GRCh38
rs770745168 Chr.10: 124829380-124829380 on Build GRCh38

View Full Product Details